Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38839207-38839715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAGC
Entrez Gene ID
64121
Gene Name
Ras related GTP binding C
Gene Aliases
GTR2, RAGC, TIB929
UniGene
Hs.532461
Gene Chromosome Location
Chr.1: 38838197-38859823 on Build GRCh38
Transcripts
NM_001271851.1 NM_022157.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6847592 c.1078C>T p.R360* Chr.1: 38839675-38839675 on Build GRCh38
380742 c.1182G>T p.T394T Chr.1: 38839571-38839571 on Build GRCh38
187452 c.1187G>A p.R396Q Chr.1: 38839566-38839566 on Build GRCh38
6769750 c.1192G>A p.A398T Chr.1: 38839561-38839561 on Build GRCh38
5787560 c.1156C>G p.L386V Chr.1: 38839597-38839597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184780039 Chr.1: 38839272-38839272 on Build GRCh38
rs752648511 Chr.1: 38839536-38839536 on Build GRCh38
rs563228372 Chr.1: 38839375-38839375 on Build GRCh38
rs532270153 Chr.1: 38839407-38839407 on Build GRCh38
rs780315074 Chr.1: 38839561-38839561 on Build GRCh38
rs755057549 Chr.1: 38839563-38839563 on Build GRCh38
rs973220751 Chr.1: 38839309-38839309 on Build GRCh38
rs753629041 Chr.1: 38839540-38839540 on Build GRCh38
rs550985257 Chr.1: 38839501-38839501 on Build GRCh38
rs180757664 Chr.1: 38839242-38839242 on Build GRCh38
rs770548337 Chr.1: 38839588-38839588 on Build GRCh38
rs937309270 Chr.1: 38839443-38839443 on Build GRCh38
rs758550293 Chr.1: 38839558-38839558 on Build GRCh38
rs764045300 Chr.1: 38839282-38839282 on Build GRCh38
rs751840536 Chr.1: 38839691-38839691 on Build GRCh38
rs770232749 Chr.1: 38839572-38839572 on Build GRCh38
rs553068233 Chr.1: 38839674-38839674 on Build GRCh38
rs548454807 Chr.1: 38839478-38839478 on Build GRCh38
rs577096931 Chr.1: 38839331-38839332 on Build GRCh38
rs965783989 Chr.1: 38839581-38839581 on Build GRCh38
rs953044815 Chr.1: 38839248-38839248 on Build GRCh38
rs181882194 Chr.1: 38839491-38839491 on Build GRCh38
rs543453355 Chr.1: 38839304-38839304 on Build GRCh38
rs376494692 Chr.1: 38839648-38839648 on Build GRCh38
rs189687689 Chr.1: 38839422-38839422 on Build GRCh38
rs969315110 Chr.1: 38839645-38839645 on Build GRCh38
rs749660538 Chr.1: 38839580-38839580 on Build GRCh38
rs370932639 Chr.1: 38839500-38839500 on Build GRCh38
rs916538081 Chr.1: 38839321-38839321 on Build GRCh38
rs186958680 Chr.1: 38839528-38839528 on Build GRCh38
rs963490289 Chr.1: 38839623-38839623 on Build GRCh38
rs973676688 Chr.1: 38839625-38839625 on Build GRCh38
rs201546414 Chr.1: 38839349-38839349 on Build GRCh38
rs78358435 Chr.1: 38839689-38839689 on Build GRCh38
rs1055835247 Chr.1: 38839457-38839457 on Build GRCh38
rs539521742 Chr.1: 38839537-38839537 on Build GRCh38
rs766556826 Chr.1: 38839682-38839682 on Build GRCh38
rs548486349 Chr.1: 38839441-38839441 on Build GRCh38
rs370745456 Chr.1: 38839663-38839663 on Build GRCh38
rs954816061 Chr.1: 38839268-38839268 on Build GRCh38
rs1030276820 Chr.1: 38839267-38839267 on Build GRCh38
rs927596093 Chr.1: 38839368-38839368 on Build GRCh38
rs893520454 Chr.1: 38839455-38839455 on Build GRCh38
rs368045628 Chr.1: 38839649-38839649 on Build GRCh38
rs890472037 Chr.1: 38839566-38839566 on Build GRCh38
rs773571179 Chr.1: 38839578-38839578 on Build GRCh38
rs903341053 Chr.1: 38839533-38839533 on Build GRCh38
rs11549875 Chr.1: 38839289-38839289 on Build GRCh38
rs919088120 Chr.1: 38839331-38839334 on Build GRCh38
rs373907856 Chr.1: 38839611-38839611 on Build GRCh38
rs748679583 Chr.1: 38839567-38839567 on Build GRCh38
rs764704610 Chr.1: 38839539-38839539 on Build GRCh38
rs981799983 Chr.1: 38839330-38839330 on Build GRCh38
rs963554054 Chr.1: 38839305-38839305 on Build GRCh38
rs759007028 Chr.1: 38839590-38839590 on Build GRCh38
rs139732615 Chr.1: 38839633-38839633 on Build GRCh38
rs74914741 Chr.1: 38839333-38839333 on Build GRCh38
rs779604320 Chr.1: 38839644-38839644 on Build GRCh38
rs150676565 Chr.1: 38839562-38839562 on Build GRCh38
rs971897924 Chr.1: 38839331-38839333 on Build GRCh38
rs142105571 Chr.1: 38839589-38839589 on Build GRCh38
rs531664142 Chr.1: 38839408-38839408 on Build GRCh38
rs947941473 Chr.1: 38839327-38839327 on Build GRCh38
rs201062095 Chr.1: 38839331-38839331 on Build GRCh38

View Full Product Details