Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81122235-81122727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AATK
Entrez Gene ID
9625
Gene Name
apoptosis associated tyrosine kinase
Gene Aliases
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP
UniGene
Hs.514575
Gene Chromosome Location
Chr.17: 81117295-81166072 on Build GRCh38
Transcripts
NM_001080395.2 NM_004920.2 XM_006722193.2 XM_006722194.2 XM_006722195.2 XM_006722196.2 XM_011525509.2 XM_011525510.2 XM_011525511.2 XM_017025421.1 XM_017025422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404448 c.1696G>T p.G566C Chr.17: 81122240-81122240 on Build GRCh38
4656257 c.1654C>T p.P552S Chr.17: 81122282-81122282 on Build GRCh38
4070597 c.1213G>A p.E405K Chr.17: 81122723-81122723 on Build GRCh38
93835 c.1448C>G p.A483G Chr.17: 81122488-81122488 on Build GRCh38
5952845 c.1461G>A p.A487A Chr.17: 81122475-81122475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531974842 Chr.17: 81122599-81122599 on Build GRCh38
rs888195768 Chr.17: 81122518-81122518 on Build GRCh38
rs756104031 Chr.17: 81122611-81122612 on Build GRCh38
rs61738821 Chr.17: 81122315-81122315 on Build GRCh38
rs987978486 Chr.17: 81122448-81122448 on Build GRCh38
rs777514215 Chr.17: 81122566-81122566 on Build GRCh38
rs748836424 Chr.17: 81122334-81122334 on Build GRCh38
rs543997096 Chr.17: 81122610-81122610 on Build GRCh38
rs1046128576 Chr.17: 81122318-81122318 on Build GRCh38
rs1048120589 Chr.17: 81122514-81122514 on Build GRCh38
rs758349445 Chr.17: 81122301-81122301 on Build GRCh38
rs1036471112 Chr.17: 81122701-81122701 on Build GRCh38
rs749286232 Chr.17: 81122656-81122676 on Build GRCh38
rs766380495 Chr.17: 81122292-81122292 on Build GRCh38
rs776166038 Chr.17: 81122408-81122408 on Build GRCh38
rs754236546 Chr.17: 81122280-81122280 on Build GRCh38
rs778994530 Chr.17: 81122294-81122294 on Build GRCh38
rs750541119 Chr.17: 81122480-81122480 on Build GRCh38
rs535117206 Chr.17: 81122674-81122674 on Build GRCh38
rs529540810 Chr.17: 81122671-81122671 on Build GRCh38
rs777474990 Chr.17: 81122702-81122702 on Build GRCh38
rs766396092 Chr.17: 81122509-81122509 on Build GRCh38
rs751615190 Chr.17: 81122523-81122523 on Build GRCh38
rs572058783 Chr.17: 81122446-81122446 on Build GRCh38
rs936574596 Chr.17: 81122295-81122295 on Build GRCh38
rs577458342 Chr.17: 81122282-81122282 on Build GRCh38
rs997749616 Chr.17: 81122559-81122559 on Build GRCh38
rs753190135 Chr.17: 81122253-81122253 on Build GRCh38
rs865977066 Chr.17: 81122693-81122693 on Build GRCh38
rs752536287 Chr.17: 81122689-81122689 on Build GRCh38
rs953571091 Chr.17: 81122440-81122440 on Build GRCh38
rs766558218 Chr.17: 81122654-81122654 on Build GRCh38
rs758534588 Chr.17: 81122489-81122489 on Build GRCh38
rs776949776 Chr.17: 81122625-81122625 on Build GRCh38
rs756071269 Chr.17: 81122675-81122675 on Build GRCh38
rs772683166 Chr.17: 81122330-81122330 on Build GRCh38
rs754947879 Chr.17: 81122529-81122529 on Build GRCh38
rs777877832 Chr.17: 81122637-81122657 on Build GRCh38
rs887923050 Chr.17: 81122346-81122346 on Build GRCh38
rs1042143905 Chr.17: 81122322-81122322 on Build GRCh38
rs879641488 Chr.17: 81122627-81122627 on Build GRCh38
rs62073020 Chr.17: 81122300-81122300 on Build GRCh38
rs774053194 Chr.17: 81122406-81122406 on Build GRCh38
rs981754089 Chr.17: 81122474-81122474 on Build GRCh38
rs747864469 Chr.17: 81122324-81122324 on Build GRCh38
rs970618533 Chr.17: 81122638-81122639 on Build GRCh38
rs984200620 Chr.17: 81122666-81122666 on Build GRCh38
rs564169165 Chr.17: 81122568-81122568 on Build GRCh38
rs767493541 Chr.17: 81122683-81122683 on Build GRCh38
rs1001927743 Chr.17: 81122296-81122296 on Build GRCh38
rs986261954 Chr.17: 81122441-81122441 on Build GRCh38
rs113124233 Chr.17: 81122435-81122435 on Build GRCh38
rs997465637 Chr.17: 81122409-81122409 on Build GRCh38
rs765506632 Chr.17: 81122477-81122477 on Build GRCh38
rs770682190 Chr.17: 81122570-81122570 on Build GRCh38
rs191163566 Chr.17: 81122418-81122418 on Build GRCh38
rs764669060 Chr.17: 81122449-81122449 on Build GRCh38
rs1036523396 Chr.17: 81122553-81122553 on Build GRCh38
rs747989954 Chr.17: 81122535-81122535 on Build GRCh38
rs925375447 Chr.17: 81122443-81122443 on Build GRCh38
rs898028431 Chr.17: 81122706-81122706 on Build GRCh38
rs754992934 Chr.17: 81122680-81122680 on Build GRCh38
rs371390882 Chr.17: 81122472-81122472 on Build GRCh38
rs937651945 Chr.17: 81122665-81122666 on Build GRCh38
rs906322440 Chr.17: 81122670-81122670 on Build GRCh38
rs562385816 Chr.17: 81122651-81122651 on Build GRCh38
rs1030329205 Chr.17: 81122439-81122439 on Build GRCh38
rs541483093 Chr.17: 81122319-81122319 on Build GRCh38
rs779969279 Chr.17: 81122306-81122306 on Build GRCh38
rs948996619 Chr.17: 81122317-81122317 on Build GRCh38
rs769544119 Chr.17: 81122325-81122325 on Build GRCh38
rs761155830 Chr.17: 81122262-81122262 on Build GRCh38
rs765480787 Chr.17: 81122650-81122650 on Build GRCh38
rs898080685 Chr.17: 81122558-81122558 on Build GRCh38
rs764507597 Chr.17: 81122270-81122270 on Build GRCh38
rs374609689 Chr.17: 81122544-81122544 on Build GRCh38
rs1001982628 Chr.17: 81122321-81122321 on Build GRCh38
rs774041351 Chr.17: 81122580-81122580 on Build GRCh38
rs762067083 Chr.17: 81122476-81122476 on Build GRCh38
rs111964845 Chr.17: 81122272-81122272 on Build GRCh38
rs926551791 Chr.17: 81122649-81122650 on Build GRCh38
rs770526867 Chr.17: 81122374-81122374 on Build GRCh38
rs755958001 Chr.17: 81122692-81122692 on Build GRCh38
rs909624748 Chr.17: 81122688-81122688 on Build GRCh38
rs889095774 Chr.17: 81122567-81122567 on Build GRCh38
rs781102582 Chr.17: 81122531-81122531 on Build GRCh38
rs1006363050 Chr.17: 81122375-81122375 on Build GRCh38
rs761317304 Chr.17: 81122426-81122426 on Build GRCh38
rs1030735757 Chr.17: 81122564-81122564 on Build GRCh38
rs913810995 Chr.17: 81122462-81122462 on Build GRCh38
rs553682365 Chr.17: 81122397-81122397 on Build GRCh38
rs1032391052 Chr.17: 81122588-81122588 on Build GRCh38
rs939814145 Chr.17: 81122696-81122696 on Build GRCh38
rs769348971 Chr.17: 81122611-81122611 on Build GRCh38

View Full Product Details