Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81559102-81559602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPLOC4
Entrez Gene ID
55666
Gene Name
NPL4 homolog, ubiquitin recognition factor
Gene Aliases
NPL4
UniGene
Hs.464333
Gene Chromosome Location
Chr.17: 81556885-81637153 on Build GRCh38
Transcripts
NM_017921.3 NR_130139.1 NR_130140.1 XM_011524980.1 XM_011524981.1 XM_017024830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6728780 c.1700A>T p.H567L Chr.17: 81559386-81559386 on Build GRCh38
5363984 c.1758T>A p.C586* Chr.17: 81559328-81559328 on Build GRCh38
985887 c.1732A>G p.T578A Chr.17: 81559354-81559354 on Build GRCh38
3970439 c.1784C>T p.P595L Chr.17: 81559302-81559302 on Build GRCh38
5972504 c.1727C>T p.T576I Chr.17: 81559359-81559359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4073997 Chr.17: 81559376-81559376 on Build GRCh38
rs911783037 Chr.17: 81559346-81559346 on Build GRCh38
rs1033576731 Chr.17: 81559170-81559170 on Build GRCh38
rs201095089 Chr.17: 81559406-81559406 on Build GRCh38
rs185495039 Chr.17: 81559284-81559284 on Build GRCh38
rs780892360 Chr.17: 81559339-81559339 on Build GRCh38
rs764653543 Chr.17: 81559441-81559441 on Build GRCh38
rs200764596 Chr.17: 81559232-81559232 on Build GRCh38
rs760752376 Chr.17: 81559375-81559375 on Build GRCh38
rs372108702 Chr.17: 81559432-81559432 on Build GRCh38
rs915394361 Chr.17: 81559259-81559259 on Build GRCh38
rs565778782 Chr.17: 81559493-81559493 on Build GRCh38
rs201724783 Chr.17: 81559316-81559316 on Build GRCh38
rs939371876 Chr.17: 81559317-81559317 on Build GRCh38
rs963306166 Chr.17: 81559539-81559539 on Build GRCh38
rs765231711 Chr.17: 81559388-81559388 on Build GRCh38
rs754243833 Chr.17: 81559301-81559301 on Build GRCh38
rs113318717 Chr.17: 81559353-81559353 on Build GRCh38
rs995662756 Chr.17: 81559541-81559541 on Build GRCh38
rs763483716 Chr.17: 81559435-81559435 on Build GRCh38
rs978095554 Chr.17: 81559206-81559206 on Build GRCh38
rs756808590 Chr.17: 81559338-81559338 on Build GRCh38
rs943594611 Chr.17: 81559255-81559255 on Build GRCh38
rs1017452590 Chr.17: 81559179-81559179 on Build GRCh38
rs920344694 Chr.17: 81559291-81559291 on Build GRCh38
rs374727447 Chr.17: 81559228-81559228 on Build GRCh38
rs145477352 Chr.17: 81559240-81559240 on Build GRCh38
rs1012668419 Chr.17: 81559517-81559517 on Build GRCh38
rs375129128 Chr.17: 81559387-81559387 on Build GRCh38
rs929006755 Chr.17: 81559125-81559125 on Build GRCh38
rs553761278 Chr.17: 81559171-81559171 on Build GRCh38
rs779743562 Chr.17: 81559394-81559394 on Build GRCh38
rs556646071 Chr.17: 81559151-81559151 on Build GRCh38
rs755532885 Chr.17: 81559231-81559231 on Build GRCh38
rs752060168 Chr.17: 81559464-81559464 on Build GRCh38
rs189401802 Chr.17: 81559126-81559126 on Build GRCh38
rs535559042 Chr.17: 81559484-81559484 on Build GRCh38
rs4076819 Chr.17: 81559543-81559543 on Build GRCh38
rs1018685198 Chr.17: 81559576-81559576 on Build GRCh38
rs773520835 Chr.17: 81559161-81559161 on Build GRCh38
rs1055040024 Chr.17: 81559331-81559331 on Build GRCh38
rs80158493 Chr.17: 81559570-81559570 on Build GRCh38
rs989793858 Chr.17: 81559545-81559545 on Build GRCh38
rs371486676 Chr.17: 81559256-81559256 on Build GRCh38
rs974647404 Chr.17: 81559402-81559402 on Build GRCh38
rs951573764 Chr.17: 81559558-81559558 on Build GRCh38
rs558843959 Chr.17: 81559522-81559522 on Build GRCh38
rs768496713 Chr.17: 81559369-81559369 on Build GRCh38
rs760989644 Chr.17: 81559293-81559293 on Build GRCh38
rs780505200 Chr.17: 81559226-81559226 on Build GRCh38
rs17852305 Chr.17: 81559377-81559377 on Build GRCh38
rs765659882 Chr.17: 81559427-81559427 on Build GRCh38
rs537936874 Chr.17: 81559128-81559128 on Build GRCh38
rs1023847674 Chr.17: 81559528-81559528 on Build GRCh38
rs199791293 Chr.17: 81559286-81559286 on Build GRCh38
rs961441121 Chr.17: 81559201-81559201 on Build GRCh38
rs779644376 Chr.17: 81559364-81559364 on Build GRCh38
rs900363351 Chr.17: 81559186-81559186 on Build GRCh38
rs368004865 Chr.17: 81559370-81559370 on Build GRCh38
rs766605718 Chr.17: 81559296-81559296 on Build GRCh38
rs967332357 Chr.17: 81559559-81559559 on Build GRCh38
rs766562337 Chr.17: 81559486-81559486 on Build GRCh38
rs928080240 Chr.17: 81559417-81559417 on Build GRCh38
rs368492583 Chr.17: 81559424-81559424 on Build GRCh38
rs890377838 Chr.17: 81559157-81559157 on Build GRCh38
rs749100852 Chr.17: 81559366-81559366 on Build GRCh38
rs755373731 Chr.17: 81559305-81559305 on Build GRCh38
rs765719786 Chr.17: 81559139-81559139 on Build GRCh38
rs202133528 Chr.17: 81559415-81559415 on Build GRCh38
rs1038943924 Chr.17: 81559456-81559456 on Build GRCh38
rs773099693 Chr.17: 81559405-81559405 on Build GRCh38
rs547385419 Chr.17: 81559489-81559489 on Build GRCh38
rs775374294 Chr.17: 81559264-81559264 on Build GRCh38
rs900331475 Chr.17: 81559361-81559361 on Build GRCh38
rs755997193 Chr.17: 81559352-81559352 on Build GRCh38
rs1035247279 Chr.17: 81559223-81559223 on Build GRCh38
rs748031448 Chr.17: 81559400-81559400 on Build GRCh38
rs568502633 Chr.17: 81559469-81559469 on Build GRCh38
rs759007754 Chr.17: 81559385-81559385 on Build GRCh38
rs1024339446 Chr.17: 81559390-81559390 on Build GRCh38
rs773366241 Chr.17: 81559374-81559374 on Build GRCh38
rs966693889 Chr.17: 81559205-81559205 on Build GRCh38
rs770977635 Chr.17: 81559412-81559412 on Build GRCh38
rs755643109 Chr.17: 81559393-81559393 on Build GRCh38
rs200065610 Chr.17: 81559373-81559373 on Build GRCh38
rs1047602298 Chr.17: 81559144-81559144 on Build GRCh38
rs535424365 Chr.17: 81559166-81559166 on Build GRCh38
rs944415580 Chr.17: 81559455-81559455 on Build GRCh38
rs776413086 Chr.17: 81559243-81559243 on Build GRCh38
rs1029903996 Chr.17: 81559514-81559514 on Build GRCh38
rs1022549832 Chr.17: 81559549-81559549 on Build GRCh38
rs946532735 Chr.17: 81559470-81559470 on Build GRCh38
rs201622340 Chr.17: 81559396-81559396 on Build GRCh38
rs761777816 Chr.17: 81559372-81559372 on Build GRCh38
rs1043963925 Chr.17: 81559473-81559473 on Build GRCh38
rs181217938 Chr.17: 81559156-81559156 on Build GRCh38
rs756410877 Chr.17: 81559217-81559217 on Build GRCh38
rs1010974337 Chr.17: 81559198-81559198 on Build GRCh38
rs758536511 Chr.17: 81559243-81559249 on Build GRCh38
rs1039388197 Chr.17: 81559354-81559354 on Build GRCh38
rs1022841327 Chr.17: 81559199-81559199 on Build GRCh38
rs537605448 Chr.17: 81559516-81559516 on Build GRCh38
rs374544964 Chr.17: 81559454-81559454 on Build GRCh38
rs950487278 Chr.17: 81559410-81559410 on Build GRCh38
rs1005374673 Chr.17: 81559194-81559194 on Build GRCh38
rs749950444 Chr.17: 81559389-81559389 on Build GRCh38
rs750700616 Chr.17: 81559215-81559215 on Build GRCh38
rs373904474 Chr.17: 81559292-81559292 on Build GRCh38
rs989717632 Chr.17: 81559397-81559397 on Build GRCh38
rs765749847 Chr.17: 81559378-81559378 on Build GRCh38
rs770636858 Chr.17: 81559233-81559233 on Build GRCh38
rs972495896 Chr.17: 81559204-81559204 on Build GRCh38
rs748941511 Chr.17: 81559395-81559395 on Build GRCh38
rs895104009 Chr.17: 81559344-81559344 on Build GRCh38
rs753318257 Chr.17: 81559326-81559326 on Build GRCh38
rs762737271 Chr.17: 81559271-81559271 on Build GRCh38
rs139547131 Chr.17: 81559379-81559379 on Build GRCh38
rs915452182 Chr.17: 81559399-81559399 on Build GRCh38
rs369490885 Chr.17: 81559289-81559289 on Build GRCh38
rs769446676 Chr.17: 81559257-81559257 on Build GRCh38

View Full Product Details