Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81904640-81905140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYT2
Entrez Gene ID
5833
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
Gene Aliases
ET
UniGene
Hs.569843
Gene Chromosome Location
Chr.17: 81900965-81911477 on Build GRCh38
Transcripts
NM_001184917.2 NM_001256433.2 NM_001256434.2 NM_001256435.2 NM_001282203.1 NM_001282204.1 NM_002861.4 XM_005256386.3 XM_005256387.3 XM_006722287.3 XM_017024910.1 XM_017024911.1 XM_017024912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6742211 c.1117G>A p.A373T Chr.17: 81904886-81904886 on Build GRCh38
4130833 c.1116G>T p.E372D Chr.17: 81904887-81904887 on Build GRCh38
7213083 c.1128G>A p.A376A Chr.17: 81904929-81904929 on Build GRCh38
3523702 c.993C>T p.F331F Chr.17: 81905131-81905131 on Build GRCh38
4070749 c.1080C>T p.N360N Chr.17: 81904923-81904923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779170350 Chr.17: 81904993-81904993 on Build GRCh38
rs543643006 Chr.17: 81904706-81904706 on Build GRCh38
rs756660391 Chr.17: 81905062-81905062 on Build GRCh38
rs377278120 Chr.17: 81904928-81904928 on Build GRCh38
rs201073746 Chr.17: 81904873-81904873 on Build GRCh38
rs960117279 Chr.17: 81904707-81904707 on Build GRCh38
rs756107791 Chr.17: 81904987-81904987 on Build GRCh38
rs772470146 Chr.17: 81904995-81904995 on Build GRCh38
rs796583138 Chr.17: 81904893-81904893 on Build GRCh38
rs549185631 Chr.17: 81904816-81904816 on Build GRCh38
rs146242304 Chr.17: 81904870-81904870 on Build GRCh38
rs936968038 Chr.17: 81905002-81905002 on Build GRCh38
rs774727176 Chr.17: 81904820-81904820 on Build GRCh38
rs558120215 Chr.17: 81904662-81904662 on Build GRCh38
rs1040917091 Chr.17: 81905080-81905080 on Build GRCh38
rs899480363 Chr.17: 81904766-81904766 on Build GRCh38
rs757204451 Chr.17: 81904905-81904905 on Build GRCh38
rs374371929 Chr.17: 81904871-81904871 on Build GRCh38
rs749397926 Chr.17: 81904990-81904990 on Build GRCh38
rs776806846 Chr.17: 81904802-81904802 on Build GRCh38
rs759061730 Chr.17: 81904940-81904940 on Build GRCh38
rs146308482 Chr.17: 81905089-81905089 on Build GRCh38
rs1035816720 Chr.17: 81904824-81904824 on Build GRCh38
rs895566293 Chr.17: 81905012-81905012 on Build GRCh38
rs34655860 Chr.17: 81904911-81904911 on Build GRCh38
rs777955570 Chr.17: 81905100-81905100 on Build GRCh38
rs2230474 Chr.17: 81904728-81904728 on Build GRCh38
rs138132734 Chr.17: 81904917-81904917 on Build GRCh38
rs983342875 Chr.17: 81904710-81904710 on Build GRCh38
rs368920032 Chr.17: 81904814-81904814 on Build GRCh38
rs774169060 Chr.17: 81904958-81904959 on Build GRCh38
rs983538620 Chr.17: 81905057-81905057 on Build GRCh38
rs578101513 Chr.17: 81905115-81905115 on Build GRCh38
rs899762055 Chr.17: 81904952-81904952 on Build GRCh38
rs373373199 Chr.17: 81905026-81905026 on Build GRCh38
rs773645916 Chr.17: 81904930-81904930 on Build GRCh38
rs895657024 Chr.17: 81904830-81904830 on Build GRCh38
rs770992474 Chr.17: 81904957-81905006 on Build GRCh38
rs905541127 Chr.17: 81904967-81904967 on Build GRCh38
rs368308572 Chr.17: 81904849-81904849 on Build GRCh38
rs973374441 Chr.17: 81904731-81904731 on Build GRCh38
rs922046403 Chr.17: 81904736-81904736 on Build GRCh38
rs866765965 Chr.17: 81905060-81905060 on Build GRCh38
rs8069388 Chr.17: 81904948-81904948 on Build GRCh38
rs769823503 Chr.17: 81904913-81904913 on Build GRCh38
rs953415774 Chr.17: 81905065-81905065 on Build GRCh38
rs935458561 Chr.17: 81904759-81904759 on Build GRCh38
rs71367089 Chr.17: 81904669-81904670 on Build GRCh38
rs201803263 Chr.17: 81904848-81904848 on Build GRCh38
rs142625466 Chr.17: 81905088-81905088 on Build GRCh38
rs960228631 Chr.17: 81904831-81904831 on Build GRCh38
rs763855613 Chr.17: 81904958-81904958 on Build GRCh38
rs762647817 Chr.17: 81904956-81904956 on Build GRCh38
rs754208985 Chr.17: 81904712-81904712 on Build GRCh38
rs113217594 Chr.17: 81904822-81904822 on Build GRCh38
rs779946571 Chr.17: 81904989-81904989 on Build GRCh38
rs375092431 Chr.17: 81904992-81904992 on Build GRCh38
rs755105462 Chr.17: 81905052-81905052 on Build GRCh38
rs776536145 Chr.17: 81904780-81904780 on Build GRCh38
rs1001801927 Chr.17: 81904681-81904681 on Build GRCh38
rs570234782 Chr.17: 81904975-81904975 on Build GRCh38
rs376136979 Chr.17: 81905027-81905027 on Build GRCh38
rs777674049 Chr.17: 81904872-81904872 on Build GRCh38
rs111682906 Chr.17: 81905072-81905072 on Build GRCh38
rs146889521 Chr.17: 81905006-81905006 on Build GRCh38
rs370805286 Chr.17: 81905042-81905042 on Build GRCh38
rs184613479 Chr.17: 81905019-81905019 on Build GRCh38
rs751315617 Chr.17: 81905030-81905030 on Build GRCh38
rs777261885 Chr.17: 81905116-81905116 on Build GRCh38
rs748405697 Chr.17: 81905082-81905082 on Build GRCh38
rs778206205 Chr.17: 81904689-81904689 on Build GRCh38
rs1004708472 Chr.17: 81905017-81905017 on Build GRCh38
rs541542857 Chr.17: 81905043-81905043 on Build GRCh38
rs576622867 Chr.17: 81904685-81904685 on Build GRCh38
rs769994529 Chr.17: 81904806-81904806 on Build GRCh38
rs867069121 Chr.17: 81904882-81904882 on Build GRCh38
rs758181890 Chr.17: 81904788-81904788 on Build GRCh38
rs549688337 Chr.17: 81904857-81904857 on Build GRCh38
rs775642154 Chr.17: 81904813-81904813 on Build GRCh38
rs911932672 Chr.17: 81904916-81904916 on Build GRCh38
rs747418178 Chr.17: 81905113-81905113 on Build GRCh38
rs375641594 Chr.17: 81905039-81905039 on Build GRCh38
rs905632148 Chr.17: 81904793-81904793 on Build GRCh38
rs764925804 Chr.17: 81904869-81904869 on Build GRCh38
rs774639288 Chr.17: 81904927-81904927 on Build GRCh38
rs71166176 Chr.17: 81904676-81904677 on Build GRCh38
rs140709757 Chr.17: 81905018-81905018 on Build GRCh38
rs369979391 Chr.17: 81905063-81905063 on Build GRCh38
rs973448387 Chr.17: 81904904-81904904 on Build GRCh38
rs200834115 Chr.17: 81904929-81904929 on Build GRCh38
rs746826324 Chr.17: 81904883-81904883 on Build GRCh38
rs533319940 Chr.17: 81904779-81904779 on Build GRCh38
rs781281274 Chr.17: 81904908-81904908 on Build GRCh38
rs946931625 Chr.17: 81904931-81904931 on Build GRCh38
rs771781083 Chr.17: 81904753-81904753 on Build GRCh38
rs750254749 Chr.17: 81905037-81905037 on Build GRCh38
rs200148360 Chr.17: 81904953-81904953 on Build GRCh38
rs752287495 Chr.17: 81904951-81904951 on Build GRCh38
rs760359124 Chr.17: 81905054-81905054 on Build GRCh38
rs2230473 Chr.17: 81904789-81904789 on Build GRCh38
rs539825785 Chr.17: 81904742-81904742 on Build GRCh38
rs555878257 Chr.17: 81904865-81904865 on Build GRCh38
rs377163585 Chr.17: 81904976-81904976 on Build GRCh38
rs547048391 Chr.17: 81904926-81904926 on Build GRCh38
rs369032878 Chr.17: 81904969-81904969 on Build GRCh38
rs764482566 Chr.17: 81904819-81904819 on Build GRCh38
rs141017787 Chr.17: 81904850-81904850 on Build GRCh38
rs753907087 Chr.17: 81905047-81905047 on Build GRCh38
rs1004633224 Chr.17: 81904835-81904835 on Build GRCh38
rs745949719 Chr.17: 81904805-81904805 on Build GRCh38
rs771819598 Chr.17: 81905044-81905045 on Build GRCh38
rs7405872 Chr.17: 81904754-81904754 on Build GRCh38
rs371998898 Chr.17: 81905025-81905025 on Build GRCh38
rs759512345 Chr.17: 81904970-81905019 on Build GRCh38
rs960186520 Chr.17: 81905008-81905008 on Build GRCh38
rs775331364 Chr.17: 81905048-81905049 on Build GRCh38
rs77809114 Chr.17: 81905034-81905034 on Build GRCh38
rs35576786 Chr.17: 81904670-81904671 on Build GRCh38
rs375420216 Chr.17: 81904886-81904886 on Build GRCh38
rs763034365 Chr.17: 81904972-81904972 on Build GRCh38
rs529509290 Chr.17: 81904812-81904812 on Build GRCh38
rs192362456 Chr.17: 81904771-81904771 on Build GRCh38
rs74006128 Chr.17: 81905059-81905059 on Build GRCh38
rs771025633 Chr.17: 81904801-81904801 on Build GRCh38
rs767776918 Chr.17: 81905036-81905036 on Build GRCh38
rs374341998 Chr.17: 81904920-81904920 on Build GRCh38

View Full Product Details