Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9334445-9334974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TWSG1
Entrez Gene ID
57045
Gene Name
twisted gastrulation BMP signaling modulator 1
Gene Aliases
TSG
UniGene
Hs.514685
Gene Chromosome Location
Chr.18: 9334767-9402420 on Build GRCh38
Transcripts
NM_020648.5

SNP Detail

SNP ID Chromosome Location
rs944328282 Chr.18: 9334780-9334781 on Build GRCh38
rs933796903 Chr.18: 9334469-9334469 on Build GRCh38
rs574892877 Chr.18: 9334471-9334471 on Build GRCh38
rs546868843 Chr.18: 9334481-9334482 on Build GRCh38
rs1039089530 Chr.18: 9334915-9334915 on Build GRCh38
rs988742920 Chr.18: 9334825-9334825 on Build GRCh38
rs1015670258 Chr.18: 9334473-9334473 on Build GRCh38
rs943458235 Chr.18: 9334905-9334905 on Build GRCh38
rs889589465 Chr.18: 9334871-9334871 on Build GRCh38
rs1034217315 Chr.18: 9334939-9334939 on Build GRCh38
rs868181847 Chr.18: 9334567-9334567 on Build GRCh38
rs1004204015 Chr.18: 9334649-9334649 on Build GRCh38
rs1018026393 Chr.18: 9334503-9334503 on Build GRCh38
rs574477515 Chr.18: 9334735-9334735 on Build GRCh38
rs995511040 Chr.18: 9334756-9334756 on Build GRCh38
rs1027545881 Chr.18: 9334470-9334470 on Build GRCh38
rs966957814 Chr.18: 9334753-9334753 on Build GRCh38
rs979835943 Chr.18: 9334806-9334806 on Build GRCh38
rs867103489 Chr.18: 9334636-9334636 on Build GRCh38
rs912724052 Chr.18: 9334593-9334593 on Build GRCh38
rs895220675 Chr.18: 9334945-9334945 on Build GRCh38
rs541484067 Chr.18: 9334786-9334786 on Build GRCh38
rs1050914619 Chr.18: 9334870-9334870 on Build GRCh38
rs796624018 Chr.18: 9334536-9334536 on Build GRCh38
rs1033755759 Chr.18: 9334696-9334696 on Build GRCh38
rs556629113 Chr.18: 9334808-9334808 on Build GRCh38
rs886238745 Chr.18: 9334820-9334820 on Build GRCh38
rs1026501533 Chr.18: 9334763-9334763 on Build GRCh38
rs944443519 Chr.18: 9334558-9334558 on Build GRCh38
rs959828523 Chr.18: 9334700-9334700 on Build GRCh38
rs890538387 Chr.18: 9334475-9334475 on Build GRCh38
rs900348592 Chr.18: 9334618-9334620 on Build GRCh38
rs935649728 Chr.18: 9334811-9334811 on Build GRCh38
rs959538681 Chr.18: 9334484-9334484 on Build GRCh38
rs899180057 Chr.18: 9334938-9334938 on Build GRCh38
rs992942954 Chr.18: 9334777-9334777 on Build GRCh38
rs1049474299 Chr.18: 9334628-9334628 on Build GRCh38
rs569311267 Chr.18: 9334543-9334543 on Build GRCh38
rs1024041980 Chr.18: 9334781-9334781 on Build GRCh38
rs1050384244 Chr.18: 9334588-9334588 on Build GRCh38
rs950908663 Chr.18: 9334769-9334769 on Build GRCh38
rs534419451 Chr.18: 9334669-9334669 on Build GRCh38
rs1003596741 Chr.18: 9334676-9334676 on Build GRCh38
rs545373746 Chr.18: 9334950-9334950 on Build GRCh38
rs1040168052 Chr.18: 9334602-9334602 on Build GRCh38
rs558514008 Chr.18: 9334641-9334641 on Build GRCh38
rs1004152970 Chr.18: 9334472-9334472 on Build GRCh38
rs1042066621 Chr.18: 9334533-9334533 on Build GRCh38
rs1015550670 Chr.18: 9334660-9334660 on Build GRCh38
rs944464813 Chr.18: 9334600-9334600 on Build GRCh38
rs899131904 Chr.18: 9334505-9334505 on Build GRCh38
rs574942223 Chr.18: 9334845-9334845 on Build GRCh38
rs1045594188 Chr.18: 9334846-9334846 on Build GRCh38
rs567035425 Chr.18: 9334645-9334645 on Build GRCh38
rs28372752 Chr.18: 9334687-9334687 on Build GRCh38
rs1038780566 Chr.18: 9334568-9334568 on Build GRCh38
rs867283195 Chr.18: 9334499-9334499 on Build GRCh38
rs55759113 Chr.18: 9334656-9334656 on Build GRCh38
rs994667720 Chr.18: 9334619-9334619 on Build GRCh38
rs989658996 Chr.18: 9334723-9334723 on Build GRCh38
rs539836740 Chr.18: 9334565-9334565 on Build GRCh38
rs902905675 Chr.18: 9334606-9334606 on Build GRCh38
rs970779162 Chr.18: 9334474-9334474 on Build GRCh38
rs537405979 Chr.18: 9334943-9334943 on Build GRCh38
rs867710933 Chr.18: 9334523-9334523 on Build GRCh38
rs933847807 Chr.18: 9334860-9334860 on Build GRCh38
rs886356005 Chr.18: 9334630-9334630 on Build GRCh38

View Full Product Details