Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12446867-12447374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105371998
Entrez Gene ID
105371998
Gene Name
uncharacterized LOC105371998
Gene Aliases
-
UniGene
Hs.515283
Gene Chromosome Location
Chr.18: 12432774-12447798 on Build GRCh38
Transcripts
NR_136514.1 NM_001128626.1 NM_001128627.1 NM_020148.2 XM_005258122.4 XM_011525701.1 XM_011525702.1

Gene Symbol
SPIRE1
Entrez Gene ID
56907
Gene Name
spire type actin nucleation factor 1
Gene Aliases
Spir-1
UniGene
Hs.515283
Gene Chromosome Location
Chr.18: 12446512-12658102 on Build GRCh38
Transcripts
NR_136514.1 NM_001128626.1 NM_001128627.1 NM_020148.2 XM_005258122.4 XM_011525701.1 XM_011525702.1

SNP Detail

SNP ID Chromosome Location
rs993774593 Chr.18: 12447033-12447033 on Build GRCh38
rs979047298 Chr.18: 12447163-12447163 on Build GRCh38
rs765241254 Chr.18: 12447076-12447077 on Build GRCh38
rs549278390 Chr.18: 12446947-12446947 on Build GRCh38
rs898890481 Chr.18: 12447037-12447037 on Build GRCh38
rs1047167319 Chr.18: 12447263-12447263 on Build GRCh38
rs993679698 Chr.18: 12447223-12447223 on Build GRCh38
rs556029537 Chr.18: 12447219-12447219 on Build GRCh38
rs1014629160 Chr.18: 12447081-12447081 on Build GRCh38
rs978526557 Chr.18: 12447103-12447103 on Build GRCh38
rs1003365111 Chr.18: 12447322-12447322 on Build GRCh38
rs914172276 Chr.18: 12446958-12446958 on Build GRCh38
rs762512266 Chr.18: 12447236-12447236 on Build GRCh38
rs534148385 Chr.18: 12447155-12447155 on Build GRCh38
rs146489801 Chr.18: 12447075-12447075 on Build GRCh38
rs1034886443 Chr.18: 12447337-12447337 on Build GRCh38
rs544831780 Chr.18: 12447344-12447344 on Build GRCh38
rs760058370 Chr.18: 12447066-12447066 on Build GRCh38
rs942421732 Chr.18: 12446970-12446970 on Build GRCh38
rs947696526 Chr.18: 12446946-12446946 on Build GRCh38
rs569462679 Chr.18: 12447213-12447213 on Build GRCh38
rs933591131 Chr.18: 12446962-12446962 on Build GRCh38
rs558211024 Chr.18: 12447132-12447132 on Build GRCh38
rs1052024047 Chr.18: 12447182-12447182 on Build GRCh38
rs925390610 Chr.18: 12447112-12447112 on Build GRCh38
rs769658939 Chr.18: 12447205-12447205 on Build GRCh38
rs1022840402 Chr.18: 12447302-12447302 on Build GRCh38
rs1037456642 Chr.18: 12447021-12447021 on Build GRCh38
rs539923149 Chr.18: 12447114-12447114 on Build GRCh38
rs372286706 Chr.18: 12447116-12447116 on Build GRCh38
rs496485 Chr.18: 12447304-12447304 on Build GRCh38
rs67360297 Chr.18: 12447284-12447285 on Build GRCh38
rs943534156 Chr.18: 12447196-12447196 on Build GRCh38
rs567405115 Chr.18: 12446967-12446967 on Build GRCh38
rs763563017 Chr.18: 12447259-12447259 on Build GRCh38
rs895507232 Chr.18: 12447254-12447254 on Build GRCh38
rs1007697078 Chr.18: 12447017-12447017 on Build GRCh38
rs969874408 Chr.18: 12447100-12447100 on Build GRCh38
rs745794564 Chr.18: 12447097-12447097 on Build GRCh38
rs1005847087 Chr.18: 12447297-12447297 on Build GRCh38
rs900821486 Chr.18: 12447027-12447027 on Build GRCh38
rs913461829 Chr.18: 12446945-12446945 on Build GRCh38
rs75307639 Chr.18: 12447283-12447283 on Build GRCh38
rs141041977 Chr.18: 12447151-12447151 on Build GRCh38
rs761477041 Chr.18: 12447306-12447306 on Build GRCh38
rs913929899 Chr.18: 12447137-12447137 on Build GRCh38
rs889321007 Chr.18: 12446989-12446989 on Build GRCh38
rs776440141 Chr.18: 12446954-12446954 on Build GRCh38
rs549704681 Chr.18: 12447063-12447063 on Build GRCh38
rs942888916 Chr.18: 12447203-12447204 on Build GRCh38
rs988835900 Chr.18: 12446893-12446893 on Build GRCh38
rs1045000149 Chr.18: 12447300-12447300 on Build GRCh38
rs910766816 Chr.18: 12447195-12447195 on Build GRCh38
rs551136246 Chr.18: 12446904-12446904 on Build GRCh38
rs878869257 Chr.18: 12447133-12447133 on Build GRCh38
rs58328517 Chr.18: 12447283-12447284 on Build GRCh38
rs183128089 Chr.18: 12447043-12447043 on Build GRCh38
rs770683183 Chr.18: 12446950-12446950 on Build GRCh38
rs993188348 Chr.18: 12447061-12447061 on Build GRCh38
rs1032836068 Chr.18: 12447325-12447325 on Build GRCh38
rs1005506514 Chr.18: 12447064-12447064 on Build GRCh38

View Full Product Details