Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22200984-22201494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

SNP Detail

SNP ID Chromosome Location
rs780463246 Chr.18: 22201223-22201223 on Build GRCh38
rs1049344206 Chr.18: 22201268-22201268 on Build GRCh38
rs1050276876 Chr.18: 22201411-22201411 on Build GRCh38
rs745903281 Chr.18: 22201046-22201046 on Build GRCh38
rs527490115 Chr.18: 22201253-22201253 on Build GRCh38
rs569560409 Chr.18: 22201471-22201471 on Build GRCh38
rs912526712 Chr.18: 22201201-22201201 on Build GRCh38
rs9965366 Chr.18: 22201149-22201149 on Build GRCh38
rs1011493645 Chr.18: 22201454-22201454 on Build GRCh38
rs766917077 Chr.18: 22201422-22201422 on Build GRCh38
rs985435709 Chr.18: 22201193-22201193 on Build GRCh38
rs749498961 Chr.18: 22201234-22201234 on Build GRCh38
rs756428094 Chr.18: 22201170-22201170 on Build GRCh38
rs1012637427 Chr.18: 22201305-22201305 on Build GRCh38
rs944667346 Chr.18: 22201209-22201209 on Build GRCh38
rs999025294 Chr.18: 22201113-22201113 on Build GRCh38
rs1011547307 Chr.18: 22201062-22201062 on Build GRCh38
rs576236856 Chr.18: 22201106-22201106 on Build GRCh38
rs372507351 Chr.18: 22201439-22201439 on Build GRCh38
rs1032296269 Chr.18: 22201065-22201065 on Build GRCh38
rs1036296999 Chr.18: 22201118-22201118 on Build GRCh38
rs140220180 Chr.18: 22201385-22201385 on Build GRCh38
rs889040700 Chr.18: 22201438-22201438 on Build GRCh38
rs564477642 Chr.18: 22201024-22201024 on Build GRCh38
rs560932418 Chr.18: 22201401-22201401 on Build GRCh38
rs967869225 Chr.18: 22201071-22201071 on Build GRCh38
rs908957782 Chr.18: 22201224-22201224 on Build GRCh38
rs529838347 Chr.18: 22201430-22201430 on Build GRCh38
rs954100957 Chr.18: 22201164-22201164 on Build GRCh38
rs145348592 Chr.18: 22201449-22201449 on Build GRCh38
rs1050369331 Chr.18: 22201042-22201042 on Build GRCh38
rs940471750 Chr.18: 22201243-22201243 on Build GRCh38
rs185025662 Chr.18: 22201148-22201148 on Build GRCh38
rs549751610 Chr.18: 22201461-22201461 on Build GRCh38
rs1003086938 Chr.18: 22201060-22201060 on Build GRCh38
rs1044086995 Chr.18: 22201314-22201314 on Build GRCh38
rs887546448 Chr.18: 22201284-22201284 on Build GRCh38
rs545102362 Chr.18: 22201168-22201170 on Build GRCh38
rs919111119 Chr.18: 22201231-22201231 on Build GRCh38
rs923122549 Chr.18: 22201353-22201353 on Build GRCh38
rs576673668 Chr.18: 22201306-22201307 on Build GRCh38
rs956466280 Chr.18: 22201463-22201463 on Build GRCh38
rs76974536 Chr.18: 22201052-22201052 on Build GRCh38
rs956219984 Chr.18: 22201133-22201133 on Build GRCh38
rs1021632376 Chr.18: 22201067-22201067 on Build GRCh38

View Full Product Details