Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46239883-46240387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf25
Entrez Gene ID
147339
Gene Name
chromosome 18 open reading frame 25
Gene Aliases
ARKL1, RNF111L1
UniGene
Hs.116486
Gene Chromosome Location
Chr.18: 46173553-46266992 on Build GRCh38
Transcripts
NM_001008239.2 NM_145055.3 XM_011525822.1 XM_017025555.1

SNP Detail

SNP ID Chromosome Location
rs371859115 Chr.18: 46240125-46240125 on Build GRCh38
rs746089804 Chr.18: 46240157-46240157 on Build GRCh38
rs755955544 Chr.18: 46240018-46240018 on Build GRCh38
rs758286941 Chr.18: 46240229-46240229 on Build GRCh38
rs750100615 Chr.18: 46240096-46240096 on Build GRCh38
rs369747226 Chr.18: 46240206-46240206 on Build GRCh38
rs182418826 Chr.18: 46239964-46239964 on Build GRCh38
rs775927184 Chr.18: 46240069-46240069 on Build GRCh38
rs750062741 Chr.18: 46240016-46240016 on Build GRCh38
rs770192767 Chr.18: 46240000-46240000 on Build GRCh38
rs199598870 Chr.18: 46240057-46240057 on Build GRCh38
rs370106063 Chr.18: 46240216-46240216 on Build GRCh38
rs757305523 Chr.18: 46240131-46240131 on Build GRCh38
rs778755413 Chr.18: 46240124-46240124 on Build GRCh38
rs760854338 Chr.18: 46240066-46240066 on Build GRCh38
rs1013765389 Chr.18: 46239909-46239909 on Build GRCh38
rs976587073 Chr.18: 46240081-46240081 on Build GRCh38
rs150379878 Chr.18: 46240113-46240113 on Build GRCh38
rs900921107 Chr.18: 46240156-46240156 on Build GRCh38
rs779779184 Chr.18: 46240019-46240019 on Build GRCh38
rs574910652 Chr.18: 46240136-46240136 on Build GRCh38
rs34315772 Chr.18: 46240196-46240197 on Build GRCh38
rs201943962 Chr.18: 46240098-46240098 on Build GRCh38
rs771516799 Chr.18: 46240186-46240186 on Build GRCh38
rs772643993 Chr.18: 46240187-46240187 on Build GRCh38
rs568126323 Chr.18: 46239941-46239941 on Build GRCh38
rs375021485 Chr.18: 46240104-46240104 on Build GRCh38
rs774585928 Chr.18: 46240230-46240230 on Build GRCh38
rs138098166 Chr.18: 46240228-46240228 on Build GRCh38
rs199579962 Chr.18: 46240044-46240044 on Build GRCh38
rs767146327 Chr.18: 46240011-46240011 on Build GRCh38
rs546548752 Chr.18: 46240358-46240358 on Build GRCh38
rs545893767 Chr.18: 46240290-46240290 on Build GRCh38
rs773480866 Chr.18: 46240019-46240020 on Build GRCh38
rs542049311 Chr.18: 46240166-46240166 on Build GRCh38
rs1027925928 Chr.18: 46240200-46240200 on Build GRCh38
rs770312036 Chr.18: 46240066-46240066 on Build GRCh38
rs774773206 Chr.18: 46240183-46240183 on Build GRCh38
rs972192268 Chr.18: 46239940-46239940 on Build GRCh38
rs534969207 Chr.18: 46240087-46240087 on Build GRCh38
rs749733137 Chr.18: 46240170-46240170 on Build GRCh38
rs765674095 Chr.18: 46239969-46239969 on Build GRCh38
rs756512764 Chr.18: 46240278-46240278 on Build GRCh38
rs375524406 Chr.18: 46240086-46240086 on Build GRCh38
rs767035336 Chr.18: 46240089-46240089 on Build GRCh38
rs762335890 Chr.18: 46240184-46240184 on Build GRCh38
rs556175036 Chr.18: 46240084-46240084 on Build GRCh38
rs539427927 Chr.18: 46240047-46240047 on Build GRCh38
rs773276454 Chr.18: 46240310-46240310 on Build GRCh38
rs569008106 Chr.18: 46239983-46239983 on Build GRCh38
rs115149790 Chr.18: 46240295-46240295 on Build GRCh38
rs375739138 Chr.18: 46240211-46240211 on Build GRCh38
rs765032474 Chr.18: 46240217-46240217 on Build GRCh38
rs762002661 Chr.18: 46240283-46240283 on Build GRCh38
rs749256900 Chr.18: 46240023-46240023 on Build GRCh38
rs367632018 Chr.18: 46240123-46240123 on Build GRCh38
rs763565276 Chr.18: 46240077-46240077 on Build GRCh38
rs972608834 Chr.18: 46240246-46240246 on Build GRCh38
rs776161944 Chr.18: 46239972-46239972 on Build GRCh38
rs34823539 Chr.18: 46240343-46240344 on Build GRCh38
rs919375326 Chr.18: 46240280-46240280 on Build GRCh38
rs769037405 Chr.18: 46240172-46240172 on Build GRCh38
rs766168336 Chr.18: 46240106-46240106 on Build GRCh38
rs528501151 Chr.18: 46240315-46240315 on Build GRCh38
rs942164484 Chr.18: 46239918-46239918 on Build GRCh38
rs200672013 Chr.18: 46240025-46240025 on Build GRCh38
rs777923788 Chr.18: 46240027-46240027 on Build GRCh38
rs923342591 Chr.18: 46240084-46240086 on Build GRCh38
rs747116361 Chr.18: 46240032-46240032 on Build GRCh38
rs201138073 Chr.18: 46240007-46240007 on Build GRCh38
rs1053953751 Chr.18: 46240304-46240304 on Build GRCh38
rs140369791 Chr.18: 46239973-46239973 on Build GRCh38

View Full Product Details