Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 61892704-61893210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF152
Entrez Gene ID
220441
Gene Name
ring finger protein 152
Gene Aliases
-
UniGene
Hs.165062
Gene Chromosome Location
Chr.18: 61808067-61894420 on Build GRCh38
Transcripts
NM_173557.2 XM_005266650.3 XM_005266652.3 XM_011525878.2 XM_011525879.2 XM_017025612.1 XM_017025613.1

SNP Detail

SNP ID Chromosome Location
rs901258221 Chr.18: 61893166-61893166 on Build GRCh38
rs756629226 Chr.18: 61892775-61892775 on Build GRCh38
rs916991479 Chr.18: 61893028-61893028 on Build GRCh38
rs940401683 Chr.18: 61893053-61893053 on Build GRCh38
rs549400573 Chr.18: 61892779-61892779 on Build GRCh38
rs1052761342 Chr.18: 61893065-61893065 on Build GRCh38
rs1052683944 Chr.18: 61892758-61892758 on Build GRCh38
rs891450929 Chr.18: 61893083-61893083 on Build GRCh38
rs373404623 Chr.18: 61893167-61893185 on Build GRCh38
rs555173390 Chr.18: 61893182-61893182 on Build GRCh38
rs1037281460 Chr.18: 61892809-61892809 on Build GRCh38
rs996122929 Chr.18: 61892831-61892831 on Build GRCh38
rs927166100 Chr.18: 61893004-61893004 on Build GRCh38
rs772858665 Chr.18: 61892770-61892770 on Build GRCh38
rs371019793 Chr.18: 61893181-61893181 on Build GRCh38
rs965404780 Chr.18: 61892923-61892923 on Build GRCh38
rs889687505 Chr.18: 61893153-61893153 on Build GRCh38
rs1017722329 Chr.18: 61892896-61892896 on Build GRCh38
rs761071149 Chr.18: 61893162-61893162 on Build GRCh38
rs550706892 Chr.18: 61893012-61893012 on Build GRCh38
rs1006997794 Chr.18: 61892884-61892884 on Build GRCh38
rs1049102938 Chr.18: 61893121-61893121 on Build GRCh38
rs566408109 Chr.18: 61892823-61892823 on Build GRCh38
rs973403356 Chr.18: 61892948-61892948 on Build GRCh38
rs552564747 Chr.18: 61893037-61893037 on Build GRCh38
rs771305576 Chr.18: 61893134-61893134 on Build GRCh38
rs566114821 Chr.18: 61893080-61893080 on Build GRCh38
rs567224203 Chr.18: 61893022-61893022 on Build GRCh38
rs994914888 Chr.18: 61893184-61893184 on Build GRCh38
rs538753069 Chr.18: 61893089-61893089 on Build GRCh38
rs1007461315 Chr.18: 61893160-61893160 on Build GRCh38
rs917446204 Chr.18: 61892996-61892996 on Build GRCh38
rs188499270 Chr.18: 61892861-61892861 on Build GRCh38
rs536113882 Chr.18: 61893025-61893025 on Build GRCh38
rs544482310 Chr.18: 61893170-61893170 on Build GRCh38
rs73004445 Chr.18: 61892794-61892794 on Build GRCh38
rs937118655 Chr.18: 61892738-61892738 on Build GRCh38
rs889987912 Chr.18: 61893140-61893140 on Build GRCh38
rs935714774 Chr.18: 61893063-61893063 on Build GRCh38
rs1020103547 Chr.18: 61892907-61892907 on Build GRCh38
rs995786632 Chr.18: 61893113-61893113 on Build GRCh38
rs895510967 Chr.18: 61892783-61892783 on Build GRCh38
rs144720420 Chr.18: 61892739-61892739 on Build GRCh38
rs913194842 Chr.18: 61892951-61892951 on Build GRCh38
rs138572818 Chr.18: 61893105-61893105 on Build GRCh38
rs193099914 Chr.18: 61893114-61893114 on Build GRCh38
rs530607570 Chr.18: 61893003-61893003 on Build GRCh38
rs1035989301 Chr.18: 61893097-61893097 on Build GRCh38
rs544963020 Chr.18: 61892785-61892785 on Build GRCh38
rs998509451 Chr.18: 61892785-61892787 on Build GRCh38
rs555702467 Chr.18: 61892926-61892926 on Build GRCh38
rs968628302 Chr.18: 61892979-61892979 on Build GRCh38
rs993474144 Chr.18: 61892958-61892958 on Build GRCh38
rs951030269 Chr.18: 61892957-61892957 on Build GRCh38
rs1027163842 Chr.18: 61892847-61892847 on Build GRCh38

View Full Product Details