Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41020775-41021275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFNL1
Entrez Gene ID
200172
Gene Name
schlafen like 1
Gene Aliases
-
UniGene
Hs.194609
Gene Chromosome Location
Chr.1: 41015597-41023444 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

Gene Symbol
SLFNL1-AS1
Entrez Gene ID
100507178
Gene Name
SLFNL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660056
Gene Chromosome Location
Chr.1: 41014590-41043890 on Build GRCh38
Transcripts
NM_001168247.2 NM_001300859.1 NM_144990.3 XM_005270597.2 XM_005270598.2 XM_005270599.2 XM_006710432.3 XM_011540943.1 XM_011540945.1 XM_011540947.1 XM_011540948.1 XM_011540949.1 XM_011540950.1 XM_011540951.1 XM_011540952.2 XM_011540953.2 XM_011540954.1 XM_017000573.1 XM_017000574.1 NR_037868.1

SNP Detail

SNP ID Chromosome Location
rs1026769772 Chr.1: 41020883-41020883 on Build GRCh38
rs1031574033 Chr.1: 41020974-41020974 on Build GRCh38
rs202184637 Chr.1: 41020928-41020928 on Build GRCh38
rs547173992 Chr.1: 41020826-41020826 on Build GRCh38
rs900546857 Chr.1: 41021215-41021215 on Build GRCh38
rs889377265 Chr.1: 41021003-41021003 on Build GRCh38
rs568345196 Chr.1: 41021043-41021043 on Build GRCh38
rs943609797 Chr.1: 41021004-41021004 on Build GRCh38
rs548042916 Chr.1: 41021041-41021041 on Build GRCh38
rs528289462 Chr.1: 41021040-41021040 on Build GRCh38
rs764441907 Chr.1: 41021237-41021237 on Build GRCh38
rs181950273 Chr.1: 41020969-41020969 on Build GRCh38
rs984758234 Chr.1: 41020894-41020894 on Build GRCh38
rs1007699177 Chr.1: 41020815-41020815 on Build GRCh38
rs570253446 Chr.1: 41020993-41020993 on Build GRCh38
rs991336121 Chr.1: 41020905-41020905 on Build GRCh38
rs869161704 Chr.1: 41020929-41020930 on Build GRCh38
rs572378316 Chr.1: 41021052-41021053 on Build GRCh38
rs996142866 Chr.1: 41020912-41020913 on Build GRCh38
rs930529032 Chr.1: 41021064-41021064 on Build GRCh38
rs34845599 Chr.1: 41020928-41020929 on Build GRCh38
rs1054026803 Chr.1: 41020998-41020998 on Build GRCh38
rs564314748 Chr.1: 41020930-41020930 on Build GRCh38
rs920478803 Chr.1: 41020872-41020872 on Build GRCh38
rs753142279 Chr.1: 41021007-41021007 on Build GRCh38
rs952069417 Chr.1: 41020884-41020884 on Build GRCh38
rs761027870 Chr.1: 41021032-41021032 on Build GRCh38
rs562419918 Chr.1: 41020859-41020859 on Build GRCh38
rs146960358 Chr.1: 41020927-41020928 on Build GRCh38
rs1019040846 Chr.1: 41020829-41020829 on Build GRCh38
rs575801816 Chr.1: 41020875-41020875 on Build GRCh38
rs1022305839 Chr.1: 41020923-41020923 on Build GRCh38
rs377102770 Chr.1: 41020964-41020964 on Build GRCh38
rs754120648 Chr.1: 41021250-41021250 on Build GRCh38
rs944587931 Chr.1: 41021153-41021153 on Build GRCh38
rs887822555 Chr.1: 41021081-41021081 on Build GRCh38
rs900493035 Chr.1: 41020911-41020911 on Build GRCh38
rs550589298 Chr.1: 41021208-41021208 on Build GRCh38
rs958589656 Chr.1: 41020903-41020903 on Build GRCh38
rs565788129 Chr.1: 41021213-41021213 on Build GRCh38
rs901951023 Chr.1: 41020845-41020845 on Build GRCh38
rs907684864 Chr.1: 41021092-41021092 on Build GRCh38
rs1016865395 Chr.1: 41020899-41020899 on Build GRCh38
rs370180361 Chr.1: 41021166-41021166 on Build GRCh38
rs775739576 Chr.1: 41021128-41021128 on Build GRCh38
rs767750045 Chr.1: 41021019-41021019 on Build GRCh38
rs185906605 Chr.1: 41021212-41021212 on Build GRCh38
rs951233655 Chr.1: 41021164-41021164 on Build GRCh38
rs924306532 Chr.1: 41020971-41020971 on Build GRCh38
rs1026404487 Chr.1: 41021080-41021080 on Build GRCh38
rs111436958 Chr.1: 41020929-41020929 on Build GRCh38
rs930581691 Chr.1: 41020882-41020882 on Build GRCh38
rs1024043542 Chr.1: 41021021-41021021 on Build GRCh38

View Full Product Details