Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79950665-79951201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQLC1
Entrez Gene ID
80148
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
UniGene
Hs.288284
Gene Chromosome Location
Chr.18: 79902420-79951665 on Build GRCh38
Transcripts
NM_001146343.1 NM_001146345.1 NM_025078.4 XM_005266770.2 XM_017025997.1 XM_017025998.1 XM_017025999.1 XM_017026000.1 XM_017026002.1 XM_017026003.1 XM_017026004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4170977 c.196C>T p.L66F Chr.18: 79950731-79950731 on Build GRCh38
708809 c.95C>T p.P32L Chr.18: 79950832-79950832 on Build GRCh38
4645312 c.25C>T p.L9F Chr.18: 79950902-79950902 on Build GRCh38
5462776 c.203+8G>A p.? Chr.18: 79950716-79950716 on Build GRCh38
7385361 c.124A>G p.R42G Chr.18: 79950803-79950803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012049296 Chr.18: 79951159-79951159 on Build GRCh38
rs776561436 Chr.18: 79950920-79950920 on Build GRCh38
rs772963556 Chr.18: 79950912-79950912 on Build GRCh38
rs746415827 Chr.18: 79950876-79950876 on Build GRCh38
rs914269492 Chr.18: 79951137-79951137 on Build GRCh38
rs200332963 Chr.18: 79950691-79950691 on Build GRCh38
rs752337487 Chr.18: 79950852-79950852 on Build GRCh38
rs766217119 Chr.18: 79950918-79950918 on Build GRCh38
rs370965811 Chr.18: 79950840-79950840 on Build GRCh38
rs578021350 Chr.18: 79950850-79950850 on Build GRCh38
rs562674735 Chr.18: 79950978-79950978 on Build GRCh38
rs769934770 Chr.18: 79950721-79950721 on Build GRCh38
rs906436142 Chr.18: 79951175-79951176 on Build GRCh38
rs764798435 Chr.18: 79950849-79950849 on Build GRCh38
rs1042772463 Chr.18: 79951136-79951136 on Build GRCh38
rs201497747 Chr.18: 79950831-79950831 on Build GRCh38
rs752427376 Chr.18: 79950925-79950925 on Build GRCh38
rs1012970957 Chr.18: 79951022-79951022 on Build GRCh38
rs907578847 Chr.18: 79950778-79950778 on Build GRCh38
rs541428874 Chr.18: 79950994-79950994 on Build GRCh38
rs747595521 Chr.18: 79950713-79950713 on Build GRCh38
rs202147190 Chr.18: 79950703-79950703 on Build GRCh38
rs759691294 Chr.18: 79950716-79950716 on Build GRCh38
rs751290398 Chr.18: 79950743-79950743 on Build GRCh38
rs1015758793 Chr.18: 79951071-79951071 on Build GRCh38
rs113759560 Chr.18: 79950715-79950715 on Build GRCh38
rs554178740 Chr.18: 79950789-79950789 on Build GRCh38
rs977693136 Chr.18: 79951146-79951146 on Build GRCh38
rs201364198 Chr.18: 79950711-79950711 on Build GRCh38
rs145964219 Chr.18: 79950821-79950821 on Build GRCh38
rs535499805 Chr.18: 79950758-79950758 on Build GRCh38
rs777963841 Chr.18: 79950712-79950712 on Build GRCh38
rs150915352 Chr.18: 79950772-79950772 on Build GRCh38
rs754559069 Chr.18: 79950777-79950777 on Build GRCh38
rs892944994 Chr.18: 79950753-79950753 on Build GRCh38
rs2012689 Chr.18: 79951157-79951157 on Build GRCh38
rs985908295 Chr.18: 79951131-79951131 on Build GRCh38
rs750294868 Chr.18: 79950767-79950767 on Build GRCh38
rs1041639785 Chr.18: 79950955-79950955 on Build GRCh38
rs1004748791 Chr.18: 79951063-79951063 on Build GRCh38
rs938625546 Chr.18: 79951177-79951177 on Build GRCh38
rs952998522 Chr.18: 79951130-79951130 on Build GRCh38
rs749922108 Chr.18: 79950692-79950692 on Build GRCh38
rs1046237976 Chr.18: 79951171-79951171 on Build GRCh38
rs903137986 Chr.18: 79950962-79950962 on Build GRCh38
rs931747024 Chr.18: 79950904-79950904 on Build GRCh38
rs539899520 Chr.18: 79950836-79950836 on Build GRCh38
rs757717280 Chr.18: 79950859-79950859 on Build GRCh38
rs200961253 Chr.18: 79950828-79950828 on Build GRCh38
rs757625428 Chr.18: 79950796-79950796 on Build GRCh38
rs867632983 Chr.18: 79950874-79950874 on Build GRCh38
rs1004923119 Chr.18: 79950975-79950975 on Build GRCh38
rs1051316766 Chr.18: 79951090-79951090 on Build GRCh38
rs150042611 Chr.18: 79950805-79950805 on Build GRCh38
rs2012687 Chr.18: 79951127-79951127 on Build GRCh38
rs200835703 Chr.18: 79950971-79950971 on Build GRCh38
rs2012676 Chr.18: 79950980-79950980 on Build GRCh38
rs914426241 Chr.18: 79950928-79950928 on Build GRCh38
rs895251755 Chr.18: 79951158-79951158 on Build GRCh38
rs376688558 Chr.18: 79950747-79950747 on Build GRCh38
rs754273428 Chr.18: 79950739-79950740 on Build GRCh38
rs976987756 Chr.18: 79951099-79951099 on Build GRCh38
rs111796089 Chr.18: 79950886-79950886 on Build GRCh38
rs374529705 Chr.18: 79950714-79950714 on Build GRCh38
rs747326697 Chr.18: 79950791-79950791 on Build GRCh38
rs775803098 Chr.18: 79950726-79950726 on Build GRCh38
rs767244950 Chr.18: 79950827-79950827 on Build GRCh38
rs936719152 Chr.18: 79951144-79951144 on Build GRCh38
rs1027454634 Chr.18: 79951035-79951035 on Build GRCh38
rs558272385 Chr.18: 79950900-79950900 on Build GRCh38
rs920080011 Chr.18: 79950997-79950997 on Build GRCh38
rs755636288 Chr.18: 79950768-79950768 on Build GRCh38
rs954182066 Chr.18: 79950999-79950999 on Build GRCh38
rs1004841516 Chr.18: 79951176-79951176 on Build GRCh38
rs754315074 Chr.18: 79950965-79950965 on Build GRCh38
rs973150680 Chr.18: 79950989-79950989 on Build GRCh38
rs985663805 Chr.18: 79951056-79951056 on Build GRCh38
rs770022926 Chr.18: 79950811-79950811 on Build GRCh38
rs371051372 Chr.18: 79950771-79950771 on Build GRCh38
rs963095989 Chr.18: 79950983-79950983 on Build GRCh38
rs965649922 Chr.18: 79951072-79951072 on Build GRCh38
rs755832572 Chr.18: 79950695-79950695 on Build GRCh38
rs373545819 Chr.18: 79950922-79950922 on Build GRCh38
rs750804122 Chr.18: 79950945-79950945 on Build GRCh38
rs895011599 Chr.18: 79950933-79950933 on Build GRCh38
rs763989959 Chr.18: 79950734-79950734 on Build GRCh38
rs888629408 Chr.18: 79951055-79951055 on Build GRCh38
rs768747575 Chr.18: 79950816-79950816 on Build GRCh38
rs894462777 Chr.18: 79950998-79950998 on Build GRCh38
rs764752437 Chr.18: 79950924-79950924 on Build GRCh38
rs932642077 Chr.18: 79951085-79951085 on Build GRCh38
rs924435700 Chr.18: 79951155-79951155 on Build GRCh38
rs373211142 Chr.18: 79950774-79950774 on Build GRCh38
rs761602267 Chr.18: 79950822-79950822 on Build GRCh38
rs983067581 Chr.18: 79950853-79950853 on Build GRCh38
rs746325682 Chr.18: 79950806-79950806 on Build GRCh38
rs902614369 Chr.18: 79950702-79950702 on Build GRCh38
rs778253180 Chr.18: 79950780-79950780 on Build GRCh38
rs947154938 Chr.18: 79951140-79951140 on Build GRCh38
rs139356651 Chr.18: 79950779-79950779 on Build GRCh38
rs749573631 Chr.18: 79950813-79950813 on Build GRCh38
rs749323876 Chr.18: 79950892-79950892 on Build GRCh38
rs763044428 Chr.18: 79950729-79950729 on Build GRCh38
rs756555931 Chr.18: 79950885-79950885 on Build GRCh38
rs781654591 Chr.18: 79950860-79950860 on Build GRCh38
rs891399290 Chr.18: 79951095-79951095 on Build GRCh38
rs771888362 Chr.18: 79950901-79950901 on Build GRCh38
rs768041385 Chr.18: 79950935-79950935 on Build GRCh38
rs114092865 Chr.18: 79950823-79950823 on Build GRCh38
rs765957443 Chr.18: 79950839-79950839 on Build GRCh38
rs756711323 Chr.18: 79950947-79950947 on Build GRCh38
rs762661813 Chr.18: 79950934-79950934 on Build GRCh38
rs775401070 Chr.18: 79950812-79950812 on Build GRCh38
rs767439174 Chr.18: 79950765-79950765 on Build GRCh38
rs922974052 Chr.18: 79950719-79950719 on Build GRCh38
rs760657947 Chr.18: 79950915-79950915 on Build GRCh38
rs754723342 Chr.18: 79950846-79950846 on Build GRCh38
rs577009928 Chr.18: 79950949-79950949 on Build GRCh38
rs779687354 Chr.18: 79950699-79950699 on Build GRCh38

View Full Product Details