Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 282762-283196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP2
Entrez Gene ID
8612
Gene Name
phospholipid phosphatase 2
Gene Aliases
LPP2, PAP-2c, PAP2-g, PPAP2C
UniGene
Hs.465506
Gene Chromosome Location
Chr.19: 281040-291435 on Build GRCh38
Transcripts
NM_003712.3 NM_177526.2 NM_177543.2 XM_011528396.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2749689 c.497C>T p.S166L Chr.19: 282795-282795 on Build GRCh38
5389269 c.516G>A p.G172G Chr.19: 282776-282776 on Build GRCh38
1711538 c.572C>T p.S191F Chr.19: 282783-282783 on Build GRCh38
1711539 c.509C>T p.S170F Chr.19: 282783-282783 on Build GRCh38
7303952 c.525C>T p.C175C Chr.19: 282767-282767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959107251 Chr.19: 282791-282791 on Build GRCh38
rs759505707 Chr.19: 282992-282992 on Build GRCh38
rs375095686 Chr.19: 282839-282839 on Build GRCh38
rs1041002314 Chr.19: 283057-283057 on Build GRCh38
rs529134081 Chr.19: 282874-282874 on Build GRCh38
rs1053500575 Chr.19: 282991-282991 on Build GRCh38
rs982303297 Chr.19: 283149-283149 on Build GRCh38
rs961577909 Chr.19: 283137-283137 on Build GRCh38
rs750008162 Chr.19: 282803-282803 on Build GRCh38
rs899639459 Chr.19: 283062-283062 on Build GRCh38
rs182065351 Chr.19: 282788-282788 on Build GRCh38
rs919716201 Chr.19: 282984-282984 on Build GRCh38
rs755773345 Chr.19: 282808-282808 on Build GRCh38
rs1023243822 Chr.19: 282821-282821 on Build GRCh38
rs570284749 Chr.19: 282924-282924 on Build GRCh38
rs958189074 Chr.19: 283086-283086 on Build GRCh38
rs747867943 Chr.19: 282808-282809 on Build GRCh38
rs766276434 Chr.19: 282901-282901 on Build GRCh38
rs201440917 Chr.19: 282795-282795 on Build GRCh38
rs375052903 Chr.19: 282845-282845 on Build GRCh38
rs187183516 Chr.19: 282899-282899 on Build GRCh38
rs369641940 Chr.19: 282842-282842 on Build GRCh38
rs189308198 Chr.19: 283037-283037 on Build GRCh38
rs1046765510 Chr.19: 283033-283033 on Build GRCh38
rs930955420 Chr.19: 283031-283031 on Build GRCh38
rs62103050 Chr.19: 282875-282875 on Build GRCh38
rs751510050 Chr.19: 282977-282977 on Build GRCh38
rs985457567 Chr.19: 283152-283152 on Build GRCh38
rs1000532815 Chr.19: 282948-282948 on Build GRCh38
rs555680331 Chr.19: 283139-283139 on Build GRCh38
rs868042069 Chr.19: 282966-282966 on Build GRCh38
rs777320276 Chr.19: 282937-282937 on Build GRCh38
rs145357715 Chr.19: 282797-282797 on Build GRCh38
rs981584057 Chr.19: 282827-282827 on Build GRCh38
rs200354416 Chr.19: 282844-282844 on Build GRCh38
rs996722907 Chr.19: 283069-283069 on Build GRCh38
rs779642827 Chr.19: 282834-282834 on Build GRCh38
rs1042176390 Chr.19: 282921-282921 on Build GRCh38
rs997315987 Chr.19: 282861-282861 on Build GRCh38
rs368979165 Chr.19: 282869-282869 on Build GRCh38
rs565662258 Chr.19: 282882-282882 on Build GRCh38
rs752480951 Chr.19: 283120-283120 on Build GRCh38
rs372787162 Chr.19: 282978-282978 on Build GRCh38
rs970703845 Chr.19: 283145-283145 on Build GRCh38
rs1032824867 Chr.19: 283085-283085 on Build GRCh38
rs1032443187 Chr.19: 282867-282867 on Build GRCh38
rs971952133 Chr.19: 282826-282826 on Build GRCh38
rs974147982 Chr.19: 283138-283138 on Build GRCh38
rs763007978 Chr.19: 282785-282785 on Build GRCh38
rs777150847 Chr.19: 282859-282859 on Build GRCh38
rs1008110511 Chr.19: 283041-283041 on Build GRCh38
rs1009977640 Chr.19: 283092-283092 on Build GRCh38
rs767441599 Chr.19: 283006-283006 on Build GRCh38
rs746294915 Chr.19: 282860-282860 on Build GRCh38
rs532909774 Chr.19: 282895-282895 on Build GRCh38
rs914154741 Chr.19: 282890-282890 on Build GRCh38
rs751500921 Chr.19: 282794-282794 on Build GRCh38
rs938197906 Chr.19: 282923-282923 on Build GRCh38
rs546905584 Chr.19: 282934-282934 on Build GRCh38

View Full Product Details