Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1111399-1111890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
UniGene
Hs.408708
Gene Chromosome Location
Chr.19: 1107634-1174283 on Build GRCh38
Transcripts
NM_001100122.1 NM_014963.2 XM_011527804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5444023 c.2742C>T p.H914H Chr.19: 1111594-1111594 on Build GRCh38
990742 c.2795A>C p.Q932P Chr.19: 1111541-1111541 on Build GRCh38
4484328 c.2799C>T p.T933T Chr.19: 1111516-1111516 on Build GRCh38
4832717 c.2805C>G p.F935L Chr.19: 1111510-1111510 on Build GRCh38
6410781 c.2744G>A p.C915Y Chr.19: 1111592-1111592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750621267 Chr.19: 1111509-1111509 on Build GRCh38
rs200893425 Chr.19: 1111635-1111635 on Build GRCh38
rs748673754 Chr.19: 1111548-1111548 on Build GRCh38
rs187413204 Chr.19: 1111828-1111828 on Build GRCh38
rs375526013 Chr.19: 1111657-1111657 on Build GRCh38
rs976691380 Chr.19: 1111684-1111684 on Build GRCh38
rs368071071 Chr.19: 1111532-1111532 on Build GRCh38
rs764115868 Chr.19: 1111471-1111471 on Build GRCh38
rs528244947 Chr.19: 1111779-1111779 on Build GRCh38
rs1030747384 Chr.19: 1111576-1111576 on Build GRCh38
rs765270388 Chr.19: 1111624-1111624 on Build GRCh38
rs971756296 Chr.19: 1111466-1111466 on Build GRCh38
rs778619478 Chr.19: 1111551-1111551 on Build GRCh38
rs781470534 Chr.19: 1111647-1111647 on Build GRCh38
rs956683326 Chr.19: 1111601-1111601 on Build GRCh38
rs947893190 Chr.19: 1111625-1111625 on Build GRCh38
rs967670100 Chr.19: 1111528-1111528 on Build GRCh38
rs1000514328 Chr.19: 1111827-1111827 on Build GRCh38
rs768400137 Chr.19: 1111605-1111605 on Build GRCh38
rs984544877 Chr.19: 1111644-1111644 on Build GRCh38
rs989034235 Chr.19: 1111734-1111734 on Build GRCh38
rs891310066 Chr.19: 1111488-1111488 on Build GRCh38
rs753791543 Chr.19: 1111474-1111474 on Build GRCh38
rs951736319 Chr.19: 1111516-1111516 on Build GRCh38
rs771668642 Chr.19: 1111462-1111463 on Build GRCh38
rs1038359498 Chr.19: 1111645-1111645 on Build GRCh38
rs1056661033 Chr.19: 1111857-1111857 on Build GRCh38
rs1011052453 Chr.19: 1111723-1111723 on Build GRCh38
rs78026836 Chr.19: 1111630-1111630 on Build GRCh38
rs745485718 Chr.19: 1111552-1111552 on Build GRCh38
rs905975325 Chr.19: 1111817-1111817 on Build GRCh38
rs891247141 Chr.19: 1111774-1111774 on Build GRCh38
rs757352145 Chr.19: 1111507-1111507 on Build GRCh38
rs757053531 Chr.19: 1111486-1111486 on Build GRCh38
rs1002485647 Chr.19: 1111613-1111613 on Build GRCh38
rs761705215 Chr.19: 1111621-1111621 on Build GRCh38
rs541122496 Chr.19: 1111446-1111446 on Build GRCh38
rs753229821 Chr.19: 1111649-1111649 on Build GRCh38
rs745395137 Chr.19: 1111653-1111653 on Build GRCh38
rs530533550 Chr.19: 1111702-1111702 on Build GRCh38
rs760491526 Chr.19: 1111465-1111465 on Build GRCh38
rs775377324 Chr.19: 1111597-1111597 on Build GRCh38
rs760533823 Chr.19: 1111599-1111599 on Build GRCh38
rs542602283 Chr.19: 1111705-1111705 on Build GRCh38
rs1031157533 Chr.19: 1111694-1111694 on Build GRCh38
rs771883775 Chr.19: 1111554-1111554 on Build GRCh38
rs142180762 Chr.19: 1111650-1111650 on Build GRCh38
rs765358327 Chr.19: 1111508-1111508 on Build GRCh38
rs781725914 Chr.19: 1111534-1111534 on Build GRCh38
rs778527388 Chr.19: 1111651-1111651 on Build GRCh38
rs896337883 Chr.19: 1111866-1111866 on Build GRCh38
rs750529249 Chr.19: 1111627-1111627 on Build GRCh38
rs753593129 Chr.19: 1111435-1111435 on Build GRCh38
rs956882718 Chr.19: 1111698-1111698 on Build GRCh38
rs560966999 Chr.19: 1111775-1111775 on Build GRCh38
rs1053016858 Chr.19: 1111550-1111550 on Build GRCh38
rs934876450 Chr.19: 1111716-1111716 on Build GRCh38
rs553330282 Chr.19: 1111462-1111462 on Build GRCh38
rs917713164 Chr.19: 1111608-1111608 on Build GRCh38
rs779750000 Chr.19: 1111659-1111659 on Build GRCh38
rs1044481757 Chr.19: 1111801-1111801 on Build GRCh38
rs923452046 Chr.19: 1111687-1111687 on Build GRCh38
rs941363493 Chr.19: 1111637-1111637 on Build GRCh38
rs758654791 Chr.19: 1111523-1111523 on Build GRCh38
rs1051208764 Chr.19: 1111773-1111773 on Build GRCh38
rs1044902892 Chr.19: 1111604-1111604 on Build GRCh38
rs746697108 Chr.19: 1111660-1111660 on Build GRCh38
rs371477857 Chr.19: 1111607-1111607 on Build GRCh38
rs780352421 Chr.19: 1111524-1111524 on Build GRCh38
rs371604439 Chr.19: 1111634-1111634 on Build GRCh38
rs747420226 Chr.19: 1111525-1111525 on Build GRCh38
rs930025138 Chr.19: 1111757-1111757 on Build GRCh38
rs763109163 Chr.19: 1111628-1111628 on Build GRCh38
rs1034934914 Chr.19: 1111622-1111622 on Build GRCh38
rs1039969044 Chr.19: 1111782-1111782 on Build GRCh38
rs904142854 Chr.19: 1111800-1111800 on Build GRCh38
rs893154734 Chr.19: 1111594-1111594 on Build GRCh38

View Full Product Details