Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1784729-1785222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B3
Entrez Gene ID
148229
Gene Name
ATPase phospholipid transporting 8B3
Gene Aliases
ATPIK
UniGene
Hs.306212
Gene Chromosome Location
Chr.19: 1782070-1812276 on Build GRCh38
Transcripts
NM_001178002.2 NM_138813.3 NR_047593.2 XM_006722654.2 XM_006722655.2 XM_006722656.2 XM_011527707.1 XM_011527708.1 XM_011527709.1 XM_011527710.2 XM_011527712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5702653 c.3386T>G p.L1129R Chr.19: 1785194-1785194 on Build GRCh38
3530536 c.3676G>A p.E1226K Chr.19: 1784833-1784833 on Build GRCh38
6305714 c.3673A>C p.K1225Q Chr.19: 1784836-1784836 on Build GRCh38
6851414 c.3564T>C p.Y1188Y Chr.19: 1784945-1784945 on Build GRCh38
3796779 c.3619G>A p.V1207M Chr.19: 1784890-1784890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888889726 Chr.19: 1785049-1785049 on Build GRCh38
rs567922973 Chr.19: 1784767-1784767 on Build GRCh38
rs369463834 Chr.19: 1785134-1785134 on Build GRCh38
rs372149389 Chr.19: 1785108-1785108 on Build GRCh38
rs754359377 Chr.19: 1784946-1784946 on Build GRCh38
rs757755479 Chr.19: 1784938-1784949 on Build GRCh38
rs774122132 Chr.19: 1785172-1785172 on Build GRCh38
rs748648165 Chr.19: 1784788-1784788 on Build GRCh38
rs761753808 Chr.19: 1784836-1784836 on Build GRCh38
rs776441117 Chr.19: 1784833-1784833 on Build GRCh38
rs781549083 Chr.19: 1784856-1784856 on Build GRCh38
rs3764605 Chr.19: 1784891-1784891 on Build GRCh38
rs1026185938 Chr.19: 1784764-1784764 on Build GRCh38
rs577438787 Chr.19: 1784944-1784944 on Build GRCh38
rs201211115 Chr.19: 1785184-1785184 on Build GRCh38
rs761156019 Chr.19: 1785126-1785126 on Build GRCh38
rs939619525 Chr.19: 1785016-1785016 on Build GRCh38
rs975260653 Chr.19: 1784834-1784834 on Build GRCh38
rs535423206 Chr.19: 1784772-1784772 on Build GRCh38
rs1047042983 Chr.19: 1784992-1784992 on Build GRCh38
rs763041235 Chr.19: 1784935-1784935 on Build GRCh38
rs376028226 Chr.19: 1785176-1785176 on Build GRCh38
rs957254092 Chr.19: 1785080-1785080 on Build GRCh38
rs953214342 Chr.19: 1784808-1784808 on Build GRCh38
rs759527013 Chr.19: 1785109-1785109 on Build GRCh38
rs915206733 Chr.19: 1785106-1785106 on Build GRCh38
rs750570289 Chr.19: 1784840-1784840 on Build GRCh38
rs200320156 Chr.19: 1785185-1785185 on Build GRCh38
rs757897405 Chr.19: 1785133-1785133 on Build GRCh38
rs745356280 Chr.19: 1784824-1784824 on Build GRCh38
rs765072869 Chr.19: 1784837-1784837 on Build GRCh38
rs199919557 Chr.19: 1784852-1784852 on Build GRCh38
rs564293418 Chr.19: 1785103-1785103 on Build GRCh38
rs769764948 Chr.19: 1784921-1784921 on Build GRCh38
rs114965447 Chr.19: 1785038-1785038 on Build GRCh38
rs767293026 Chr.19: 1784848-1784848 on Build GRCh38
rs570758383 Chr.19: 1785070-1785070 on Build GRCh38
rs749065705 Chr.19: 1785167-1785167 on Build GRCh38
rs373100122 Chr.19: 1785175-1785175 on Build GRCh38
rs755328810 Chr.19: 1784854-1784854 on Build GRCh38
rs764489514 Chr.19: 1785129-1785129 on Build GRCh38
rs766404738 Chr.19: 1784941-1784941 on Build GRCh38
rs758443161 Chr.19: 1784844-1784844 on Build GRCh38
rs754426217 Chr.19: 1785144-1785144 on Build GRCh38
rs545053309 Chr.19: 1785104-1785104 on Build GRCh38
rs763914274 Chr.19: 1784831-1784831 on Build GRCh38
rs746488649 Chr.19: 1784952-1784952 on Build GRCh38
rs547355137 Chr.19: 1785178-1785178 on Build GRCh38
rs757989595 Chr.19: 1784950-1784950 on Build GRCh38
rs148714490 Chr.19: 1784748-1784748 on Build GRCh38
rs913664921 Chr.19: 1784871-1784871 on Build GRCh38
rs770919440 Chr.19: 1785169-1785169 on Build GRCh38
rs370111495 Chr.19: 1784980-1784980 on Build GRCh38
rs565945224 Chr.19: 1784880-1784880 on Build GRCh38
rs777486565 Chr.19: 1785166-1785166 on Build GRCh38
rs1027705548 Chr.19: 1785045-1785045 on Build GRCh38
rs769527335 Chr.19: 1784979-1784979 on Build GRCh38
rs753155554 Chr.19: 1784857-1784857 on Build GRCh38
rs771007699 Chr.19: 1784983-1784983 on Build GRCh38
rs773935809 Chr.19: 1784810-1784810 on Build GRCh38
rs754414343 Chr.19: 1785131-1785131 on Build GRCh38
rs377166900 Chr.19: 1784756-1784756 on Build GRCh38
rs560654004 Chr.19: 1785074-1785074 on Build GRCh38
rs760412040 Chr.19: 1784827-1784827 on Build GRCh38
rs866688246 Chr.19: 1784937-1784937 on Build GRCh38
rs779888089 Chr.19: 1784890-1784890 on Build GRCh38
rs1008042558 Chr.19: 1784785-1784785 on Build GRCh38
rs1008241215 Chr.19: 1785028-1785028 on Build GRCh38
rs188876412 Chr.19: 1785069-1785069 on Build GRCh38
rs965847644 Chr.19: 1785071-1785071 on Build GRCh38
rs1036615342 Chr.19: 1785033-1785033 on Build GRCh38
rs527951485 Chr.19: 1785075-1785075 on Build GRCh38
rs898094058 Chr.19: 1785140-1785140 on Build GRCh38
rs761510958 Chr.19: 1784917-1784917 on Build GRCh38
rs146819777 Chr.19: 1785114-1785114 on Build GRCh38
rs948109449 Chr.19: 1784977-1784977 on Build GRCh38
rs767818521 Chr.19: 1785110-1785110 on Build GRCh38
rs376910813 Chr.19: 1785154-1785154 on Build GRCh38
rs35797178 Chr.19: 1784875-1784876 on Build GRCh38
rs142249158 Chr.19: 1784757-1784757 on Build GRCh38
rs773109057 Chr.19: 1784929-1784929 on Build GRCh38
rs202079167 Chr.19: 1784914-1784914 on Build GRCh38
rs901566769 Chr.19: 1785193-1785193 on Build GRCh38
rs768349090 Chr.19: 1784912-1784912 on Build GRCh38
rs367724631 Chr.19: 1784766-1784766 on Build GRCh38
rs774543871 Chr.19: 1784989-1784989 on Build GRCh38
rs751940032 Chr.19: 1784880-1784880 on Build GRCh38
rs1031441219 Chr.19: 1785198-1785198 on Build GRCh38
rs369167085 Chr.19: 1785156-1785156 on Build GRCh38
rs748026744 Chr.19: 1784966-1784966 on Build GRCh38
rs192519682 Chr.19: 1785090-1785090 on Build GRCh38
rs16994563 Chr.19: 1785186-1785186 on Build GRCh38
rs957976785 Chr.19: 1784862-1784862 on Build GRCh38
rs1019376435 Chr.19: 1784798-1784798 on Build GRCh38
rs553828577 Chr.19: 1784782-1784782 on Build GRCh38
rs778417302 Chr.19: 1784873-1784873 on Build GRCh38
rs755906541 Chr.19: 1785157-1785157 on Build GRCh38
rs994513603 Chr.19: 1785039-1785039 on Build GRCh38
rs928765762 Chr.19: 1784933-1784933 on Build GRCh38
rs998656940 Chr.19: 1785197-1785197 on Build GRCh38
rs996626057 Chr.19: 1785047-1785047 on Build GRCh38
rs756699166 Chr.19: 1784867-1784867 on Build GRCh38
rs751784716 Chr.19: 1784942-1784942 on Build GRCh38
rs374897856 Chr.19: 1784960-1784960 on Build GRCh38
rs770348097 Chr.19: 1784792-1784792 on Build GRCh38
rs779657321 Chr.19: 1784951-1784951 on Build GRCh38
rs531314437 Chr.19: 1785107-1785107 on Build GRCh38
rs751122501 Chr.19: 1785136-1785136 on Build GRCh38
rs371471335 Chr.19: 1784959-1784959 on Build GRCh38
rs745422063 Chr.19: 1784879-1784879 on Build GRCh38
rs765823919 Chr.19: 1785188-1785188 on Build GRCh38
rs565854681 Chr.19: 1785179-1785179 on Build GRCh38
rs3764606 Chr.19: 1784945-1784945 on Build GRCh38
rs991671424 Chr.19: 1785130-1785130 on Build GRCh38
rs183931951 Chr.19: 1784932-1784932 on Build GRCh38
rs899641567 Chr.19: 1785044-1785044 on Build GRCh38
rs771914558 Chr.19: 1784826-1784826 on Build GRCh38
rs987822017 Chr.19: 1784870-1784870 on Build GRCh38
rs1007707870 Chr.19: 1785057-1785057 on Build GRCh38
rs989512306 Chr.19: 1785091-1785091 on Build GRCh38
rs887201329 Chr.19: 1785010-1785010 on Build GRCh38

View Full Product Details