Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1817093-1817594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REXO1
Entrez Gene ID
57455
Gene Name
RNA exonuclease 1 homolog
Gene Aliases
ELOABP1, EloA-BP1, REX1, TCEB3BP1
UniGene
Hs.192477
Gene Chromosome Location
Chr.19: 1815246-1848684 on Build GRCh38
Transcripts
NM_020695.3 XM_011528144.1 XM_011528146.2 XM_017027028.1 XM_017027029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6288932 c.3136A>G p.K1046E Chr.19: 1817284-1817284 on Build GRCh38
7076165 c.3145G>A p.E1049K Chr.19: 1817275-1817275 on Build GRCh38
3989803 c.3167C>G p.T1056S Chr.19: 1817253-1817253 on Build GRCh38
5364819 c.3173C>T p.P1058L Chr.19: 1817247-1817247 on Build GRCh38
7182906 c.3169C>T p.H1057Y Chr.19: 1817251-1817251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918294971 Chr.19: 1817366-1817366 on Build GRCh38
rs369771918 Chr.19: 1817183-1817183 on Build GRCh38
rs527379571 Chr.19: 1817507-1817507 on Build GRCh38
rs191983967 Chr.19: 1817379-1817379 on Build GRCh38
rs992735866 Chr.19: 1817160-1817160 on Build GRCh38
rs571940026 Chr.19: 1817375-1817375 on Build GRCh38
rs201459041 Chr.19: 1817215-1817215 on Build GRCh38
rs377467891 Chr.19: 1817251-1817251 on Build GRCh38
rs575021554 Chr.19: 1817144-1817144 on Build GRCh38
rs535950702 Chr.19: 1817486-1817486 on Build GRCh38
rs367804217 Chr.19: 1817347-1817347 on Build GRCh38
rs929794753 Chr.19: 1817169-1817169 on Build GRCh38
rs773188755 Chr.19: 1817401-1817401 on Build GRCh38
rs942074146 Chr.19: 1817193-1817193 on Build GRCh38
rs542308115 Chr.19: 1817157-1817157 on Build GRCh38
rs371729745 Chr.19: 1817378-1817378 on Build GRCh38
rs749802516 Chr.19: 1817363-1817363 on Build GRCh38
rs748635031 Chr.19: 1817261-1817261 on Build GRCh38
rs183172046 Chr.19: 1817260-1817260 on Build GRCh38
rs200984356 Chr.19: 1817253-1817253 on Build GRCh38
rs779553137 Chr.19: 1817364-1817364 on Build GRCh38
rs377356877 Chr.19: 1817198-1817198 on Build GRCh38
rs762819611 Chr.19: 1817403-1817403 on Build GRCh38
rs913538467 Chr.19: 1817116-1817116 on Build GRCh38
rs775342592 Chr.19: 1817287-1817287 on Build GRCh38
rs984007831 Chr.19: 1817563-1817563 on Build GRCh38
rs200320632 Chr.19: 1817311-1817311 on Build GRCh38
rs12976128 Chr.19: 1817114-1817114 on Build GRCh38
rs779003850 Chr.19: 1817555-1817555 on Build GRCh38
rs530871935 Chr.19: 1817248-1817248 on Build GRCh38
rs775255921 Chr.19: 1817194-1817194 on Build GRCh38
rs1020899691 Chr.19: 1817539-1817539 on Build GRCh38
rs996671831 Chr.19: 1817276-1817276 on Build GRCh38
rs920177565 Chr.19: 1817163-1817163 on Build GRCh38
rs778228825 Chr.19: 1817437-1817437 on Build GRCh38
rs72399352 Chr.19: 1817146-1817147 on Build GRCh38
rs200953332 Chr.19: 1817319-1817319 on Build GRCh38
rs752859479 Chr.19: 1817420-1817420 on Build GRCh38
rs201975652 Chr.19: 1817362-1817362 on Build GRCh38
rs557176899 Chr.19: 1817412-1817412 on Build GRCh38
rs1052189546 Chr.19: 1817485-1817485 on Build GRCh38
rs557115544 Chr.19: 1817135-1817135 on Build GRCh38
rs146760890 Chr.19: 1817414-1817414 on Build GRCh38
rs747248686 Chr.19: 1817249-1817249 on Build GRCh38
rs769202518 Chr.19: 1817426-1817426 on Build GRCh38
rs957943652 Chr.19: 1817113-1817113 on Build GRCh38
rs141773521 Chr.19: 1817324-1817324 on Build GRCh38
rs970444367 Chr.19: 1817323-1817323 on Build GRCh38
rs3841563 Chr.19: 1817156-1817157 on Build GRCh38
rs766234108 Chr.19: 1817368-1817368 on Build GRCh38
rs756608081 Chr.19: 1817428-1817428 on Build GRCh38
rs983054210 Chr.19: 1817392-1817392 on Build GRCh38
rs150552735 Chr.19: 1817246-1817246 on Build GRCh38
rs554196333 Chr.19: 1817489-1817489 on Build GRCh38
rs371555346 Chr.19: 1817201-1817201 on Build GRCh38
rs756316821 Chr.19: 1817351-1817352 on Build GRCh38
rs979846322 Chr.19: 1817115-1817115 on Build GRCh38
rs766431160 Chr.19: 1817334-1817334 on Build GRCh38
rs760456894 Chr.19: 1817195-1817195 on Build GRCh38
rs200454205 Chr.19: 1817132-1817132 on Build GRCh38
rs1043304869 Chr.19: 1817495-1817496 on Build GRCh38
rs368062875 Chr.19: 1817301-1817301 on Build GRCh38
rs370311753 Chr.19: 1817310-1817310 on Build GRCh38
rs773959948 Chr.19: 1817182-1817182 on Build GRCh38
rs757803150 Chr.19: 1817461-1817461 on Build GRCh38
rs906434596 Chr.19: 1817541-1817541 on Build GRCh38
rs549492488 Chr.19: 1817302-1817302 on Build GRCh38
rs558253216 Chr.19: 1817553-1817553 on Build GRCh38
rs749655817 Chr.19: 1817424-1817425 on Build GRCh38
rs779456001 Chr.19: 1817478-1817478 on Build GRCh38
rs959165024 Chr.19: 1817556-1817556 on Build GRCh38
rs771706361 Chr.19: 1817282-1817282 on Build GRCh38
rs373237890 Chr.19: 1817196-1817196 on Build GRCh38
rs937553657 Chr.19: 1817149-1817149 on Build GRCh38
rs769571188 Chr.19: 1817400-1817400 on Build GRCh38
rs1010432063 Chr.19: 1817489-1817491 on Build GRCh38
rs370685976 Chr.19: 1817501-1817501 on Build GRCh38
rs751743055 Chr.19: 1817341-1817341 on Build GRCh38
rs139908971 Chr.19: 1817321-1817321 on Build GRCh38
rs763086043 Chr.19: 1817333-1817333 on Build GRCh38
rs918504787 Chr.19: 1817165-1817165 on Build GRCh38
rs745411810 Chr.19: 1817279-1817279 on Build GRCh38
rs778404656 Chr.19: 1817265-1817265 on Build GRCh38
rs770286989 Chr.19: 1817264-1817264 on Build GRCh38
rs1033729781 Chr.19: 1817554-1817554 on Build GRCh38
rs766449707 Chr.19: 1817408-1817408 on Build GRCh38
rs267605353 Chr.19: 1817288-1817288 on Build GRCh38
rs371332447 Chr.19: 1817135-1817136 on Build GRCh38
rs777070361 Chr.19: 1817173-1817173 on Build GRCh38
rs759512613 Chr.19: 1817413-1817413 on Build GRCh38
rs141798167 Chr.19: 1817225-1817225 on Build GRCh38
rs926348871 Chr.19: 1817124-1817124 on Build GRCh38
rs750451978 Chr.19: 1817240-1817240 on Build GRCh38
rs769589310 Chr.19: 1817508-1817508 on Build GRCh38
rs926966985 Chr.19: 1817145-1817146 on Build GRCh38
rs761704878 Chr.19: 1817380-1817380 on Build GRCh38
rs1034070964 Chr.19: 1817534-1817534 on Build GRCh38
rs754406008 Chr.19: 1817441-1817441 on Build GRCh38
rs746668606 Chr.19: 1817370-1817370 on Build GRCh38
rs554184166 Chr.19: 1817176-1817176 on Build GRCh38
rs944085086 Chr.19: 1817410-1817410 on Build GRCh38
rs970897820 Chr.19: 1817118-1817118 on Build GRCh38
rs909701959 Chr.19: 1817386-1817388 on Build GRCh38
rs770360172 Chr.19: 1817180-1817180 on Build GRCh38
rs960053756 Chr.19: 1817538-1817538 on Build GRCh38
rs951054029 Chr.19: 1817562-1817562 on Build GRCh38
rs981862623 Chr.19: 1817123-1817123 on Build GRCh38
rs373527155 Chr.19: 1817237-1817237 on Build GRCh38
rs753156376 Chr.19: 1817361-1817361 on Build GRCh38
rs113647181 Chr.19: 1817344-1817344 on Build GRCh38
rs780447812 Chr.19: 1817372-1817373 on Build GRCh38

View Full Product Details