Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3977137-3977640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2
Entrez Gene ID
1938
Gene Name
eukaryotic translation elongation factor 2
Gene Aliases
EEF-2, EF-2, EF2, SCA26
UniGene
Hs.515070
Gene Chromosome Location
Chr.19: 3976056-3985463 on Build GRCh38
Transcripts
NM_001961.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7412381 c.2366C>T p.P789L Chr.19: 3977232-3977232 on Build GRCh38
4077924 c.2368G>A p.V790I Chr.19: 3977230-3977230 on Build GRCh38
4077925 c.2101C>T p.R701C Chr.19: 3977577-3977577 on Build GRCh38
5519585 c.2307C>T p.H769H Chr.19: 3977291-3977291 on Build GRCh38
3970978 c.2362C>T p.L788L Chr.19: 3977236-3977236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772173797 Chr.19: 3977437-3977437 on Build GRCh38
rs777130257 Chr.19: 3977420-3977420 on Build GRCh38
rs773851698 Chr.19: 3977351-3977351 on Build GRCh38
rs2230564 Chr.19: 3977473-3977473 on Build GRCh38
rs376043516 Chr.19: 3977379-3977379 on Build GRCh38
rs200198271 Chr.19: 3977201-3977201 on Build GRCh38
rs181721180 Chr.19: 3977384-3977384 on Build GRCh38
rs777557385 Chr.19: 3977506-3977506 on Build GRCh38
rs766224831 Chr.19: 3977484-3977484 on Build GRCh38
rs547495456 Chr.19: 3977377-3977377 on Build GRCh38
rs757833144 Chr.19: 3977184-3977184 on Build GRCh38
rs771481480 Chr.19: 3977416-3977416 on Build GRCh38
rs2305112 Chr.19: 3977361-3977361 on Build GRCh38
rs757087170 Chr.19: 3977279-3977279 on Build GRCh38
rs764307072 Chr.19: 3977464-3977464 on Build GRCh38
rs770033999 Chr.19: 3977621-3977621 on Build GRCh38
rs777077931 Chr.19: 3977236-3977236 on Build GRCh38
rs147961884 Chr.19: 3977366-3977366 on Build GRCh38
rs749017577 Chr.19: 3977521-3977521 on Build GRCh38
rs367985360 Chr.19: 3977572-3977572 on Build GRCh38
rs755091330 Chr.19: 3977357-3977357 on Build GRCh38
rs908928973 Chr.19: 3977605-3977605 on Build GRCh38
rs773670601 Chr.19: 3977216-3977216 on Build GRCh38
rs113688813 Chr.19: 3977552-3977552 on Build GRCh38
rs774037219 Chr.19: 3977559-3977559 on Build GRCh38
rs775458404 Chr.19: 3977446-3977446 on Build GRCh38
rs751710484 Chr.19: 3977355-3977355 on Build GRCh38
rs751796646 Chr.19: 3977261-3977261 on Build GRCh38
rs748319866 Chr.19: 3977608-3977608 on Build GRCh38
rs529147290 Chr.19: 3977298-3977298 on Build GRCh38
rs767003023 Chr.19: 3977501-3977501 on Build GRCh38
rs746340768 Chr.19: 3977536-3977536 on Build GRCh38
rs1036212863 Chr.19: 3977408-3977408 on Build GRCh38
rs199566207 Chr.19: 3977575-3977575 on Build GRCh38
rs202184545 Chr.19: 3977415-3977415 on Build GRCh38
rs113568904 Chr.19: 3977427-3977427 on Build GRCh38
rs779501154 Chr.19: 3977371-3977371 on Build GRCh38
rs35805642 Chr.19: 3977291-3977291 on Build GRCh38
rs528278063 Chr.19: 3977368-3977368 on Build GRCh38
rs759052423 Chr.19: 3977407-3977407 on Build GRCh38
rs376426961 Chr.19: 3977276-3977276 on Build GRCh38
rs756996547 Chr.19: 3977576-3977576 on Build GRCh38
rs151205847 Chr.19: 3977219-3977219 on Build GRCh38
rs879723404 Chr.19: 3977616-3977616 on Build GRCh38
rs1027485386 Chr.19: 3977193-3977193 on Build GRCh38
rs769691602 Chr.19: 3977192-3977192 on Build GRCh38
rs747615893 Chr.19: 3977602-3977602 on Build GRCh38
rs140987549 Chr.19: 3977285-3977285 on Build GRCh38
rs750546057 Chr.19: 3977188-3977188 on Build GRCh38
rs767165183 Chr.19: 3977173-3977173 on Build GRCh38
rs775576430 Chr.19: 3977542-3977542 on Build GRCh38
rs746524810 Chr.19: 3977376-3977376 on Build GRCh38
rs758537899 Chr.19: 3977257-3977257 on Build GRCh38
rs759649184 Chr.19: 3977392-3977392 on Build GRCh38
rs36527 Chr.19: 3977488-3977488 on Build GRCh38
rs764509324 Chr.19: 3977183-3977183 on Build GRCh38
rs768779185 Chr.19: 3977547-3977547 on Build GRCh38
rs931740881 Chr.19: 3977378-3977378 on Build GRCh38
rs976336742 Chr.19: 3977566-3977566 on Build GRCh38
rs536497175 Chr.19: 3977414-3977414 on Build GRCh38
rs375866867 Chr.19: 3977606-3977606 on Build GRCh38
rs780436610 Chr.19: 3977200-3977200 on Build GRCh38
rs765777095 Chr.19: 3977240-3977240 on Build GRCh38
rs771632914 Chr.19: 3977225-3977225 on Build GRCh38
rs368935155 Chr.19: 3977189-3977189 on Build GRCh38
rs766140202 Chr.19: 3977554-3977554 on Build GRCh38
rs553260662 Chr.19: 3977164-3977164 on Build GRCh38
rs113350830 Chr.19: 3977612-3977612 on Build GRCh38
rs117017020 Chr.19: 3977503-3977503 on Build GRCh38
rs780410239 Chr.19: 3977589-3977589 on Build GRCh38
rs368428296 Chr.19: 3977418-3977418 on Build GRCh38
rs772490991 Chr.19: 3977541-3977541 on Build GRCh38
rs770164294 Chr.19: 3977381-3977381 on Build GRCh38
rs537182273 Chr.19: 3977391-3977391 on Build GRCh38
rs1024307622 Chr.19: 3977431-3977431 on Build GRCh38
rs779977590 Chr.19: 3977402-3977402 on Build GRCh38
rs373921290 Chr.19: 3977318-3977318 on Build GRCh38
rs766629202 Chr.19: 3977258-3977258 on Build GRCh38
rs753005587 Chr.19: 3977175-3977175 on Build GRCh38
rs752178810 Chr.19: 3977502-3977502 on Build GRCh38
rs778618200 Chr.19: 3977363-3977363 on Build GRCh38
rs200631073 Chr.19: 3977413-3977413 on Build GRCh38
rs763503957 Chr.19: 3977389-3977389 on Build GRCh38
rs138319587 Chr.19: 3977569-3977569 on Build GRCh38
rs758267034 Chr.19: 3977399-3977399 on Build GRCh38
rs764238318 Chr.19: 3977376-3977376 on Build GRCh38
rs767795812 Chr.19: 3977358-3977358 on Build GRCh38
rs770934899 Chr.19: 3977209-3977209 on Build GRCh38
rs747817117 Chr.19: 3977380-3977380 on Build GRCh38
rs541085914 Chr.19: 3977208-3977208 on Build GRCh38
rs774021948 Chr.19: 3977170-3977170 on Build GRCh38
rs201245104 Chr.19: 3977560-3977560 on Build GRCh38
rs762185492 Chr.19: 3977238-3977238 on Build GRCh38
rs1004696690 Chr.19: 3977396-3977399 on Build GRCh38
rs978729255 Chr.19: 3977186-3977186 on Build GRCh38
rs779300501 Chr.19: 3977579-3977579 on Build GRCh38
rs1036180093 Chr.19: 3977620-3977620 on Build GRCh38
rs200627243 Chr.19: 3977565-3977565 on Build GRCh38
rs746226898 Chr.19: 3977585-3977585 on Build GRCh38
rs779468968 Chr.19: 3977530-3977530 on Build GRCh38
rs776736515 Chr.19: 3977607-3977607 on Build GRCh38
rs757666325 Chr.19: 3977529-3977529 on Build GRCh38
rs111422588 Chr.19: 3977426-3977426 on Build GRCh38
rs768383239 Chr.19: 3977301-3977301 on Build GRCh38
rs758884682 Chr.19: 3977586-3977586 on Build GRCh38
rs770310895 Chr.19: 3977344-3977344 on Build GRCh38
rs776826199 Chr.19: 3977548-3977548 on Build GRCh38
rs745877479 Chr.19: 3977290-3977290 on Build GRCh38
rs760914805 Chr.19: 3977457-3977457 on Build GRCh38
rs141817922 Chr.19: 3977553-3977553 on Build GRCh38
rs374126801 Chr.19: 3977545-3977545 on Build GRCh38
rs74172613 Chr.19: 3977491-3977491 on Build GRCh38
rs533090086 Chr.19: 3977267-3977267 on Build GRCh38
rs776792190 Chr.19: 3977313-3977313 on Build GRCh38
rs775031599 Chr.19: 3977297-3977297 on Build GRCh38
rs200990204 Chr.19: 3977397-3977397 on Build GRCh38
rs761019324 Chr.19: 3977176-3977176 on Build GRCh38
rs113046366 Chr.19: 3977212-3977212 on Build GRCh38
rs763593671 Chr.19: 3977354-3977354 on Build GRCh38
rs760858843 Chr.19: 3977395-3977395 on Build GRCh38
rs879217358 Chr.19: 3977524-3977524 on Build GRCh38
rs760361343 Chr.19: 3977231-3977231 on Build GRCh38
rs112866535 Chr.19: 3977213-3977213 on Build GRCh38
rs777381071 Chr.19: 3977269-3977269 on Build GRCh38
rs563092089 Chr.19: 3977563-3977563 on Build GRCh38
rs369457690 Chr.19: 3977249-3977249 on Build GRCh38
rs556899179 Chr.19: 3977393-3977393 on Build GRCh38
rs747813721 Chr.19: 3977411-3977411 on Build GRCh38
rs775119611 Chr.19: 3977226-3977226 on Build GRCh38
rs377137866 Chr.19: 3977330-3977330 on Build GRCh38
rs778808287 Chr.19: 3977185-3977185 on Build GRCh38

View Full Product Details