Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4668494-4669041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYDGF
Entrez Gene ID
56005
Gene Name
myeloid derived growth factor
Gene Aliases
C19orf10, EUROIMAGE1875335, IL25, IL27, IL27w, R33729_1, SF20
UniGene
Hs.465645
Gene Chromosome Location
Chr.19: 4657545-4670403 on Build GRCh38
Transcripts
NM_019107.3 XM_017026987.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998384 c.211G>T p.G71* Chr.19: 4668609-4668609 on Build GRCh38
439821 c.218C>A p.T73N Chr.19: 4668602-4668602 on Build GRCh38
6283078 c.195C>A p.F65L Chr.19: 4668625-4668625 on Build GRCh38
3404399 c.188G>A p.C63Y Chr.19: 4668632-4668632 on Build GRCh38
4667127 c.223G>T p.E75* Chr.19: 4668597-4668597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779054337 Chr.19: 4668606-4668606 on Build GRCh38
rs748682317 Chr.19: 4668660-4668661 on Build GRCh38
rs764561257 Chr.19: 4668850-4668850 on Build GRCh38
rs765611823 Chr.19: 4668660-4668660 on Build GRCh38
rs548892883 Chr.19: 4668829-4668829 on Build GRCh38
rs1030938082 Chr.19: 4668826-4668826 on Build GRCh38
rs763892689 Chr.19: 4668690-4668690 on Build GRCh38
rs560896322 Chr.19: 4668683-4668683 on Build GRCh38
rs766763656 Chr.19: 4668914-4668914 on Build GRCh38
rs1025944020 Chr.19: 4668957-4668957 on Build GRCh38
rs753405533 Chr.19: 4668702-4668702 on Build GRCh38
rs778417303 Chr.19: 4668678-4668679 on Build GRCh38
rs781504597 Chr.19: 4668783-4668783 on Build GRCh38
rs919855835 Chr.19: 4668865-4668866 on Build GRCh38
rs968672709 Chr.19: 4668802-4668802 on Build GRCh38
rs773946741 Chr.19: 4668632-4668632 on Build GRCh38
rs111741122 Chr.19: 4668594-4668594 on Build GRCh38
rs941503251 Chr.19: 4668824-4668824 on Build GRCh38
rs544849312 Chr.19: 4668531-4668531 on Build GRCh38
rs545327396 Chr.19: 4668919-4668919 on Build GRCh38
rs780099603 Chr.19: 4668663-4668663 on Build GRCh38
rs188898864 Chr.19: 4669010-4669010 on Build GRCh38
rs745439454 Chr.19: 4668679-4668680 on Build GRCh38
rs192310282 Chr.19: 4668746-4668746 on Build GRCh38
rs565926071 Chr.19: 4668936-4668936 on Build GRCh38
rs575384736 Chr.19: 4668646-4668646 on Build GRCh38
rs554161909 Chr.19: 4668889-4668889 on Build GRCh38
rs758445978 Chr.19: 4668662-4668662 on Build GRCh38
rs934087623 Chr.19: 4668888-4668888 on Build GRCh38
rs758911230 Chr.19: 4668661-4668661 on Build GRCh38
rs967843207 Chr.19: 4668706-4668706 on Build GRCh38
rs778765793 Chr.19: 4668681-4668681 on Build GRCh38
rs569416320 Chr.19: 4668806-4668806 on Build GRCh38
rs200767767 Chr.19: 4668635-4668635 on Build GRCh38
rs754463444 Chr.19: 4668598-4668598 on Build GRCh38
rs780186597 Chr.19: 4668617-4668617 on Build GRCh38
rs59730499 Chr.19: 4668866-4668867 on Build GRCh38
rs750429027 Chr.19: 4668610-4668610 on Build GRCh38
rs1002477456 Chr.19: 4668939-4668939 on Build GRCh38
rs955469593 Chr.19: 4668828-4668828 on Build GRCh38
rs764522107 Chr.19: 4668586-4668586 on Build GRCh38
rs960780555 Chr.19: 4668947-4668947 on Build GRCh38
rs770709755 Chr.19: 4668623-4668623 on Build GRCh38
rs991886812 Chr.19: 4668956-4668956 on Build GRCh38
rs895338117 Chr.19: 4668748-4668748 on Build GRCh38
rs368804337 Chr.19: 4668554-4668554 on Build GRCh38
rs372418013 Chr.19: 4668667-4668667 on Build GRCh38
rs184739106 Chr.19: 4668976-4668976 on Build GRCh38
rs1033512063 Chr.19: 4668941-4668941 on Build GRCh38
rs749045200 Chr.19: 4668673-4668673 on Build GRCh38
rs76525069 Chr.19: 4668866-4668866 on Build GRCh38
rs765336649 Chr.19: 4668576-4668576 on Build GRCh38
rs568782759 Chr.19: 4668832-4668832 on Build GRCh38
rs930230922 Chr.19: 4668794-4668794 on Build GRCh38
rs539851858 Chr.19: 4668707-4668707 on Build GRCh38
rs112535007 Chr.19: 4668867-4668867 on Build GRCh38
rs531968646 Chr.19: 4668812-4668812 on Build GRCh38
rs570825136 Chr.19: 4668937-4668937 on Build GRCh38
rs201976051 Chr.19: 4668652-4668652 on Build GRCh38
rs929858134 Chr.19: 4668900-4668900 on Build GRCh38
rs917530101 Chr.19: 4668551-4668551 on Build GRCh38
rs1044587488 Chr.19: 4668840-4668840 on Build GRCh38
rs139888497 Chr.19: 4668599-4668599 on Build GRCh38
rs113129733 Chr.19: 4668647-4668647 on Build GRCh38
rs757243884 Chr.19: 4668678-4668678 on Build GRCh38
rs963945905 Chr.19: 4668839-4668839 on Build GRCh38
rs773680717 Chr.19: 4668550-4668550 on Build GRCh38
rs1040165060 Chr.19: 4668649-4668649 on Build GRCh38
rs938594044 Chr.19: 4668901-4668901 on Build GRCh38
rs753136647 Chr.19: 4668580-4668580 on Build GRCh38
rs928311561 Chr.19: 4668922-4668922 on Build GRCh38
rs927233376 Chr.19: 4668880-4668880 on Build GRCh38
rs867680552 Chr.19: 4668865-4668865 on Build GRCh38
rs111943446 Chr.19: 4668727-4668727 on Build GRCh38
rs542327063 Chr.19: 4668850-4668851 on Build GRCh38
rs552459764 Chr.19: 4668772-4668772 on Build GRCh38
rs529632347 Chr.19: 4668689-4668689 on Build GRCh38
rs771190588 Chr.19: 4668540-4668540 on Build GRCh38
rs371227444 Chr.19: 4668618-4668618 on Build GRCh38
rs745788733 Chr.19: 4668682-4668682 on Build GRCh38
rs367760869 Chr.19: 4668654-4668654 on Build GRCh38
rs767040183 Chr.19: 4668562-4668562 on Build GRCh38
rs974278317 Chr.19: 4668857-4668858 on Build GRCh38
rs1047261307 Chr.19: 4668581-4668581 on Build GRCh38
rs759462604 Chr.19: 4668634-4668634 on Build GRCh38
rs113802909 Chr.19: 4668836-4668836 on Build GRCh38
rs558733179 Chr.19: 4668619-4668619 on Build GRCh38
rs764934236 Chr.19: 4668738-4668738 on Build GRCh38
rs1022142345 Chr.19: 4668704-4668704 on Build GRCh38
rs999704707 Chr.19: 4668971-4668971 on Build GRCh38
rs774712409 Chr.19: 4668614-4668614 on Build GRCh38
rs756651783 Chr.19: 4668723-4668723 on Build GRCh38
rs761172370 Chr.19: 4668582-4668582 on Build GRCh38
rs755381211 Chr.19: 4668752-4668752 on Build GRCh38
rs750917384 Chr.19: 4668661-4668661 on Build GRCh38
rs755138171 Chr.19: 4668669-4668669 on Build GRCh38
rs758666045 Chr.19: 4668616-4668616 on Build GRCh38

View Full Product Details