Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12969511-12970000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAND5
Entrez Gene ID
199699
Gene Name
DAN domain BMP antagonist family member 5
Gene Aliases
CER2, CERL2, CKTSF1B3, COCO, CRL2, DANTE, GREM3, SP1
UniGene
Hs.331981
Gene Chromosome Location
Chr.19: 12969618-12974753 on Build GRCh38
Transcripts
NM_152654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750162 c.55A>G p.T19A Chr.19: 12969715-12969715 on Build GRCh38
3528940 c.6C>T p.L2L Chr.19: 12969666-12969666 on Build GRCh38
3528941 c.7C>T p.L3F Chr.19: 12969667-12969667 on Build GRCh38
6639987 c.210G>T p.R70S Chr.19: 12969870-12969870 on Build GRCh38
7164176 c.71C>A p.P24H Chr.19: 12969731-12969731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776364109 Chr.19: 12969918-12969918 on Build GRCh38
rs763900976 Chr.19: 12969944-12969944 on Build GRCh38
rs1010734332 Chr.19: 12969912-12969912 on Build GRCh38
rs147724853 Chr.19: 12969923-12969923 on Build GRCh38
rs762853421 Chr.19: 12969943-12969943 on Build GRCh38
rs773434053 Chr.19: 12969715-12969715 on Build GRCh38
rs750069440 Chr.19: 12969693-12969693 on Build GRCh38
rs200141574 Chr.19: 12969612-12969612 on Build GRCh38
rs769512017 Chr.19: 12969616-12969616 on Build GRCh38
rs545711323 Chr.19: 12969751-12969751 on Build GRCh38
rs751522800 Chr.19: 12969957-12969957 on Build GRCh38
rs753662561 Chr.19: 12969782-12969782 on Build GRCh38
rs772584983 Chr.19: 12969854-12969854 on Build GRCh38
rs982529803 Chr.19: 12969759-12969759 on Build GRCh38
rs751370645 Chr.19: 12969787-12969801 on Build GRCh38
rs775895802 Chr.19: 12969748-12969748 on Build GRCh38
rs758442482 Chr.19: 12969800-12969800 on Build GRCh38
rs971214541 Chr.19: 12969530-12969530 on Build GRCh38
rs751061099 Chr.19: 12969682-12969682 on Build GRCh38
rs776267420 Chr.19: 12969626-12969637 on Build GRCh38
rs868422472 Chr.19: 12969884-12969884 on Build GRCh38
rs767162205 Chr.19: 12969690-12969690 on Build GRCh38
rs574524356 Chr.19: 12969647-12969647 on Build GRCh38
rs778868509 Chr.19: 12969792-12969792 on Build GRCh38
rs770133755 Chr.19: 12969899-12969899 on Build GRCh38
rs748058455 Chr.19: 12969794-12969794 on Build GRCh38
rs763374408 Chr.19: 12969845-12969845 on Build GRCh38
rs199651835 Chr.19: 12969650-12969650 on Build GRCh38
rs752326128 Chr.19: 12969654-12969672 on Build GRCh38
rs376183812 Chr.19: 12969651-12969651 on Build GRCh38
rs752417980 Chr.19: 12969938-12969938 on Build GRCh38
rs761854671 Chr.19: 12969637-12969637 on Build GRCh38
rs751104409 Chr.19: 12969595-12969595 on Build GRCh38
rs749163835 Chr.19: 12969705-12969705 on Build GRCh38
rs750026070 Chr.19: 12969754-12969754 on Build GRCh38
rs140618957 Chr.19: 12969624-12969627 on Build GRCh38
rs142870531 Chr.19: 12969749-12969749 on Build GRCh38
rs142500515 Chr.19: 12969930-12969930 on Build GRCh38
rs948330262 Chr.19: 12969892-12969892 on Build GRCh38
rs552889359 Chr.19: 12969979-12969979 on Build GRCh38
rs772314725 Chr.19: 12969638-12969638 on Build GRCh38
rs140227871 Chr.19: 12969905-12969905 on Build GRCh38
rs760992114 Chr.19: 12969540-12969540 on Build GRCh38
rs748168861 Chr.19: 12969615-12969615 on Build GRCh38
rs746194097 Chr.19: 12969820-12969820 on Build GRCh38
rs185960974 Chr.19: 12969596-12969596 on Build GRCh38
rs926700324 Chr.19: 12969907-12969907 on Build GRCh38
rs771214242 Chr.19: 12969730-12969730 on Build GRCh38
rs756760109 Chr.19: 12969685-12969685 on Build GRCh38
rs900959629 Chr.19: 12969695-12969695 on Build GRCh38
rs973575617 Chr.19: 12969867-12969867 on Build GRCh38
rs759948696 Chr.19: 12969740-12969740 on Build GRCh38
rs760038338 Chr.19: 12969676-12969676 on Build GRCh38
rs773345547 Chr.19: 12969646-12969646 on Build GRCh38
rs770169770 Chr.19: 12969827-12969827 on Build GRCh38
rs749251252 Chr.19: 12969623-12969623 on Build GRCh38
rs747204394 Chr.19: 12969716-12969716 on Build GRCh38
rs754954986 Chr.19: 12969707-12969707 on Build GRCh38
rs1042525086 Chr.19: 12969683-12969683 on Build GRCh38
rs201782751 Chr.19: 12969781-12969781 on Build GRCh38
rs777274489 Chr.19: 12969581-12969581 on Build GRCh38
rs757851821 Chr.19: 12969578-12969578 on Build GRCh38
rs778763400 Chr.19: 12969710-12969710 on Build GRCh38
rs377332706 Chr.19: 12969628-12969628 on Build GRCh38
rs776011842 Chr.19: 12969837-12969837 on Build GRCh38
rs558107171 Chr.19: 12969856-12969856 on Build GRCh38
rs754373850 Chr.19: 12969564-12969564 on Build GRCh38
rs757360937 Chr.19: 12969895-12969895 on Build GRCh38
rs758986698 Chr.19: 12969625-12969626 on Build GRCh38
rs199796992 Chr.19: 12969858-12969858 on Build GRCh38
rs764742041 Chr.19: 12969634-12969637 on Build GRCh38
rs370847923 Chr.19: 12969636-12969636 on Build GRCh38
rs535257159 Chr.19: 12969633-12969633 on Build GRCh38
rs780978688 Chr.19: 12969966-12969966 on Build GRCh38
rs1005197294 Chr.19: 12969833-12969833 on Build GRCh38
rs191208589 Chr.19: 12969622-12969622 on Build GRCh38
rs528256480 Chr.19: 12969802-12969802 on Build GRCh38
rs149784597 Chr.19: 12969896-12969896 on Build GRCh38
rs780760051 Chr.19: 12969605-12969605 on Build GRCh38
rs757273419 Chr.19: 12969959-12969959 on Build GRCh38
rs776972145 Chr.19: 12969736-12969736 on Build GRCh38
rs140253089 Chr.19: 12969906-12969906 on Build GRCh38
rs760221389 Chr.19: 12969911-12969911 on Build GRCh38
rs747156125 Chr.19: 12969806-12969806 on Build GRCh38
rs772075043 Chr.19: 12969713-12969713 on Build GRCh38
rs780000656 Chr.19: 12969703-12969703 on Build GRCh38
rs765798296 Chr.19: 12969677-12969677 on Build GRCh38
rs777879219 Chr.19: 12969570-12969570 on Build GRCh38
rs751613321 Chr.19: 12969889-12969889 on Build GRCh38
rs765704497 Chr.19: 12969745-12969745 on Build GRCh38
rs768790938 Chr.19: 12969627-12969627 on Build GRCh38
rs753575680 Chr.19: 12969871-12969871 on Build GRCh38
rs899740574 Chr.19: 12969935-12969935 on Build GRCh38
rs368182043 Chr.19: 12969779-12969779 on Build GRCh38
rs754859803 Chr.19: 12969787-12969787 on Build GRCh38
rs757330887 Chr.19: 12969856-12969857 on Build GRCh38
rs181131869 Chr.19: 12969702-12969702 on Build GRCh38
rs922766361 Chr.19: 12969973-12969973 on Build GRCh38
rs200246387 Chr.19: 12969962-12969962 on Build GRCh38
rs915470966 Chr.19: 12969642-12969642 on Build GRCh38
rs765003751 Chr.19: 12969929-12969929 on Build GRCh38
rs758259852 Chr.19: 12969888-12969888 on Build GRCh38
rs145704405 Chr.19: 12969900-12969900 on Build GRCh38
rs762647763 Chr.19: 12969758-12969759 on Build GRCh38
rs200778930 Chr.19: 12969807-12969807 on Build GRCh38
rs763197681 Chr.19: 12969698-12969698 on Build GRCh38
rs752608720 Chr.19: 12969887-12969887 on Build GRCh38
rs559589192 Chr.19: 12969810-12969810 on Build GRCh38
rs575145470 Chr.19: 12969712-12969712 on Build GRCh38
rs764031444 Chr.19: 12969767-12969768 on Build GRCh38
rs998345885 Chr.19: 12969729-12969729 on Build GRCh38
rs764729850 Chr.19: 12969847-12969847 on Build GRCh38
rs199609805 Chr.19: 12969960-12969960 on Build GRCh38
rs369819134 Chr.19: 12969877-12969877 on Build GRCh38
rs201362267 Chr.19: 12969965-12969965 on Build GRCh38
rs570354102 Chr.19: 12969542-12969542 on Build GRCh38

View Full Product Details