Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12974413-12974954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAND5
Entrez Gene ID
199699
Gene Name
DAN domain BMP antagonist family member 5
Gene Aliases
CER2, CERL2, CKTSF1B3, COCO, CRL2, DANTE, GREM3, SP1
UniGene
Hs.331981
Gene Chromosome Location
Chr.19: 12969618-12974753 on Build GRCh38
Transcripts
NM_152654.2

SNP Detail

SNP ID Chromosome Location
rs780816046 Chr.19: 12974873-12974873 on Build GRCh38
rs535071527 Chr.19: 12974476-12974476 on Build GRCh38
rs542197619 Chr.19: 12974874-12974874 on Build GRCh38
rs1016794506 Chr.19: 12974679-12974679 on Build GRCh38
rs5827168 Chr.19: 12974530-12974531 on Build GRCh38
rs900802138 Chr.19: 12974467-12974467 on Build GRCh38
rs915016502 Chr.19: 12974811-12974811 on Build GRCh38
rs896946607 Chr.19: 12974903-12974903 on Build GRCh38
rs900855948 Chr.19: 12974870-12974870 on Build GRCh38
rs867196396 Chr.19: 12974662-12974662 on Build GRCh38
rs777097335 Chr.19: 12974643-12974643 on Build GRCh38
rs746477910 Chr.19: 12974447-12974447 on Build GRCh38
rs1009060148 Chr.19: 12974681-12974681 on Build GRCh38
rs149262640 Chr.19: 12974446-12974446 on Build GRCh38
rs183132777 Chr.19: 12974686-12974686 on Build GRCh38
rs563692879 Chr.19: 12974878-12974878 on Build GRCh38
rs74464467 Chr.19: 12974530-12974530 on Build GRCh38
rs546102161 Chr.19: 12974571-12974571 on Build GRCh38
rs970674785 Chr.19: 12974718-12974718 on Build GRCh38
rs556550672 Chr.19: 12974477-12974477 on Build GRCh38
rs989657546 Chr.19: 12974805-12974805 on Build GRCh38
rs1028218423 Chr.19: 12974629-12974629 on Build GRCh38
rs1010552506 Chr.19: 12974482-12974482 on Build GRCh38
rs1043811766 Chr.19: 12974485-12974485 on Build GRCh38
rs1038208272 Chr.19: 12974899-12974899 on Build GRCh38
rs998113858 Chr.19: 12974556-12974556 on Build GRCh38
rs187226360 Chr.19: 12974715-12974715 on Build GRCh38
rs996963151 Chr.19: 12974549-12974549 on Build GRCh38
rs962639553 Chr.19: 12974694-12974694 on Build GRCh38
rs397823684 Chr.19: 12974542-12974543 on Build GRCh38
rs531075784 Chr.19: 12974880-12974880 on Build GRCh38
rs573454193 Chr.19: 12974782-12974782 on Build GRCh38
rs73507306 Chr.19: 12974509-12974509 on Build GRCh38
rs73006810 Chr.19: 12974529-12974529 on Build GRCh38
rs889699224 Chr.19: 12974877-12974877 on Build GRCh38
rs191674347 Chr.19: 12974836-12974836 on Build GRCh38
rs982348621 Chr.19: 12974735-12974735 on Build GRCh38
rs969416861 Chr.19: 12974827-12974827 on Build GRCh38
rs73925239 Chr.19: 12974826-12974826 on Build GRCh38
rs1042055578 Chr.19: 12974445-12974445 on Build GRCh38
rs745575121 Chr.19: 12974898-12974898 on Build GRCh38
rs534670180 Chr.19: 12974529-12974530 on Build GRCh38
rs190938539 Chr.19: 12974684-12974684 on Build GRCh38
rs770421435 Chr.19: 12974737-12974737 on Build GRCh38
rs144454318 Chr.19: 12974884-12974884 on Build GRCh38
rs35443764 Chr.19: 12974520-12974520 on Build GRCh38
rs1043652976 Chr.19: 12974909-12974909 on Build GRCh38
rs972518978 Chr.19: 12974693-12974693 on Build GRCh38

View Full Product Details