Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15453093-15453601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASAL3
Entrez Gene ID
64926
Gene Name
RAS protein activator like 3
Gene Aliases
-
UniGene
Hs.136979
Gene Chromosome Location
Chr.19: 15451624-15464571 on Build GRCh38
Transcripts
NM_022904.1 XM_011528185.1 XM_011528186.1 XM_011528187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6501662 c.2592G>A p.P864P Chr.19: 15453185-15453185 on Build GRCh38
3283703 c.2628A>T p.Q876H Chr.19: 15453149-15453149 on Build GRCh38
6434611 c.2579T>C p.L860P Chr.19: 15453198-15453198 on Build GRCh38
5506525 c.2632C>A p.R878R Chr.19: 15453145-15453145 on Build GRCh38
709677 c.2559G>A p.W853* Chr.19: 15453218-15453218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980768598 Chr.19: 15453269-15453269 on Build GRCh38
rs776961594 Chr.19: 15453384-15453384 on Build GRCh38
rs770084092 Chr.19: 15453402-15453402 on Build GRCh38
rs749630434 Chr.19: 15453260-15453260 on Build GRCh38
rs771222303 Chr.19: 15453270-15453270 on Build GRCh38
rs751141154 Chr.19: 15453191-15453191 on Build GRCh38
rs769208217 Chr.19: 15453321-15453322 on Build GRCh38
rs777821670 Chr.19: 15453464-15453464 on Build GRCh38
rs772826484 Chr.19: 15453544-15453544 on Build GRCh38
rs760301052 Chr.19: 15453547-15453547 on Build GRCh38
rs752292289 Chr.19: 15453170-15453170 on Build GRCh38
rs374340432 Chr.19: 15453372-15453372 on Build GRCh38
rs537057707 Chr.19: 15453505-15453505 on Build GRCh38
rs371535313 Chr.19: 15453308-15453308 on Build GRCh38
rs773720098 Chr.19: 15453264-15453264 on Build GRCh38
rs369005462 Chr.19: 15453125-15453125 on Build GRCh38
rs750999746 Chr.19: 15453288-15453288 on Build GRCh38
rs774002840 Chr.19: 15453425-15453425 on Build GRCh38
rs1040815750 Chr.19: 15453310-15453310 on Build GRCh38
rs760675007 Chr.19: 15453440-15453440 on Build GRCh38
rs779157603 Chr.19: 15453370-15453370 on Build GRCh38
rs912101444 Chr.19: 15453549-15453549 on Build GRCh38
rs546889324 Chr.19: 15453376-15453376 on Build GRCh38
rs773419216 Chr.19: 15453157-15453157 on Build GRCh38
rs750324054 Chr.19: 15453316-15453316 on Build GRCh38
rs376252289 Chr.19: 15453230-15453230 on Build GRCh38
rs868344911 Chr.19: 15453429-15453429 on Build GRCh38
rs1002156484 Chr.19: 15453454-15453454 on Build GRCh38
rs866006271 Chr.19: 15453154-15453154 on Build GRCh38
rs988057493 Chr.19: 15453247-15453247 on Build GRCh38
rs745721368 Chr.19: 15453114-15453114 on Build GRCh38
rs777051699 Chr.19: 15453271-15453271 on Build GRCh38
rs758054481 Chr.19: 15453174-15453174 on Build GRCh38
rs530611301 Chr.19: 15453265-15453265 on Build GRCh38
rs570701843 Chr.19: 15453320-15453320 on Build GRCh38
rs753859387 Chr.19: 15453450-15453450 on Build GRCh38
rs865778477 Chr.19: 15453490-15453490 on Build GRCh38
rs765830799 Chr.19: 15453459-15453460 on Build GRCh38
rs953003950 Chr.19: 15453299-15453299 on Build GRCh38
rs566724491 Chr.19: 15453382-15453382 on Build GRCh38
rs762582951 Chr.19: 15453281-15453281 on Build GRCh38
rs763859307 Chr.19: 15453284-15453284 on Build GRCh38
rs940881324 Chr.19: 15453550-15453550 on Build GRCh38
rs781227149 Chr.19: 15453213-15453213 on Build GRCh38
rs750021078 Chr.19: 15453438-15453438 on Build GRCh38
rs766752674 Chr.19: 15453162-15453162 on Build GRCh38
rs770135771 Chr.19: 15453486-15453486 on Build GRCh38
rs746380278 Chr.19: 15453479-15453479 on Build GRCh38
rs528100081 Chr.19: 15453118-15453118 on Build GRCh38
rs1031507809 Chr.19: 15453142-15453142 on Build GRCh38
rs780248104 Chr.19: 15453566-15453566 on Build GRCh38
rs762325565 Chr.19: 15453414-15453415 on Build GRCh38
rs373662111 Chr.19: 15453510-15453510 on Build GRCh38
rs757733502 Chr.19: 15453477-15453477 on Build GRCh38
rs766466907 Chr.19: 15453447-15453447 on Build GRCh38
rs372540608 Chr.19: 15453205-15453205 on Build GRCh38
rs768987938 Chr.19: 15453244-15453244 on Build GRCh38
rs1014527793 Chr.19: 15453399-15453399 on Build GRCh38
rs1016344546 Chr.19: 15453179-15453179 on Build GRCh38
rs749539937 Chr.19: 15453126-15453126 on Build GRCh38
rs759981877 Chr.19: 15453394-15453394 on Build GRCh38
rs373762155 Chr.19: 15453258-15453258 on Build GRCh38
rs770107622 Chr.19: 15453123-15453123 on Build GRCh38
rs766984583 Chr.19: 15453434-15453434 on Build GRCh38
rs904467535 Chr.19: 15453121-15453121 on Build GRCh38
rs761344879 Chr.19: 15453298-15453298 on Build GRCh38
rs768797116 Chr.19: 15453136-15453136 on Build GRCh38
rs780006940 Chr.19: 15453237-15453237 on Build GRCh38
rs764955727 Chr.19: 15453460-15453460 on Build GRCh38
rs774715758 Chr.19: 15453328-15453329 on Build GRCh38
rs768119646 Chr.19: 15453153-15453153 on Build GRCh38
rs778843503 Chr.19: 15453245-15453245 on Build GRCh38
rs755126096 Chr.19: 15453351-15453351 on Build GRCh38
rs761382253 Chr.19: 15453431-15453431 on Build GRCh38
rs939735127 Chr.19: 15453309-15453309 on Build GRCh38
rs756125499 Chr.19: 15453330-15453330 on Build GRCh38
rs750413610 Chr.19: 15453222-15453222 on Build GRCh38
rs370506034 Chr.19: 15453462-15453462 on Build GRCh38
rs138850805 Chr.19: 15453249-15453249 on Build GRCh38
rs779990548 Chr.19: 15453335-15453335 on Build GRCh38
rs774492792 Chr.19: 15453146-15453146 on Build GRCh38
rs760988718 Chr.19: 15453159-15453159 on Build GRCh38
rs547835084 Chr.19: 15453148-15453148 on Build GRCh38
rs377706363 Chr.19: 15453459-15453459 on Build GRCh38
rs891645372 Chr.19: 15453377-15453377 on Build GRCh38
rs763511038 Chr.19: 15453182-15453182 on Build GRCh38
rs771906473 Chr.19: 15453513-15453513 on Build GRCh38
rs753742401 Chr.19: 15453348-15453348 on Build GRCh38
rs775719835 Chr.19: 15453497-15453497 on Build GRCh38
rs1025046819 Chr.19: 15453406-15453406 on Build GRCh38
rs539384460 Chr.19: 15453375-15453375 on Build GRCh38
rs898796004 Chr.19: 15453573-15453573 on Build GRCh38
rs747145692 Chr.19: 15453465-15453465 on Build GRCh38
rs1049464034 Chr.19: 15453338-15453338 on Build GRCh38
rs57208996 Chr.19: 15453304-15453304 on Build GRCh38

View Full Product Details