Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15949248-15949749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10H4
Entrez Gene ID
126541
Gene Name
olfactory receptor family 10 subfamily H member 4
Gene Aliases
OR19-28
UniGene
Hs.553579
Gene Chromosome Location
Chr.19: 15949008-15949958 on Build GRCh38
Transcripts
NM_001004465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362735 c.268C>A p.H90N Chr.19: 15949275-15949275 on Build GRCh38
3284333 c.445A>G p.T149A Chr.19: 15949452-15949452 on Build GRCh38
3822205 c.737A>C p.H246P Chr.19: 15949744-15949744 on Build GRCh38
6150232 c.428C>A p.A143D Chr.19: 15949435-15949435 on Build GRCh38
5039733 c.455G>A p.G152D Chr.19: 15949462-15949462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748990566 Chr.19: 15949653-15949653 on Build GRCh38
rs769411240 Chr.19: 15949482-15949482 on Build GRCh38
rs762794061 Chr.19: 15949425-15949425 on Build GRCh38
rs114480958 Chr.19: 15949681-15949681 on Build GRCh38
rs201781055 Chr.19: 15949493-15949493 on Build GRCh38
rs781294206 Chr.19: 15949359-15949359 on Build GRCh38
rs767737538 Chr.19: 15949507-15949507 on Build GRCh38
rs141039294 Chr.19: 15949375-15949375 on Build GRCh38
rs367648687 Chr.19: 15949634-15949634 on Build GRCh38
rs887957881 Chr.19: 15949679-15949679 on Build GRCh38
rs532538232 Chr.19: 15949288-15949288 on Build GRCh38
rs144850333 Chr.19: 15949426-15949426 on Build GRCh38
rs753357526 Chr.19: 15949678-15949678 on Build GRCh38
rs117610912 Chr.19: 15949457-15949457 on Build GRCh38
rs61732447 Chr.19: 15949713-15949713 on Build GRCh38
rs565552384 Chr.19: 15949598-15949598 on Build GRCh38
rs1012084319 Chr.19: 15949286-15949286 on Build GRCh38
rs774844646 Chr.19: 15949487-15949487 on Build GRCh38
rs764567314 Chr.19: 15949664-15949664 on Build GRCh38
rs771226041 Chr.19: 15949366-15949366 on Build GRCh38
rs577304997 Chr.19: 15949580-15949580 on Build GRCh38
rs760805537 Chr.19: 15949517-15949517 on Build GRCh38
rs751651699 Chr.19: 15949291-15949291 on Build GRCh38
rs199816165 Chr.19: 15949374-15949374 on Build GRCh38
rs546161132 Chr.19: 15949582-15949582 on Build GRCh38
rs191127043 Chr.19: 15949402-15949402 on Build GRCh38
rs777657234 Chr.19: 15949597-15949599 on Build GRCh38
rs754437892 Chr.19: 15949462-15949462 on Build GRCh38
rs776322230 Chr.19: 15949410-15949410 on Build GRCh38
rs183387692 Chr.19: 15949441-15949441 on Build GRCh38
rs868676405 Chr.19: 15949558-15949558 on Build GRCh38
rs767682500 Chr.19: 15949450-15949450 on Build GRCh38
rs771886757 Chr.19: 15949700-15949700 on Build GRCh38
rs751807168 Chr.19: 15949612-15949612 on Build GRCh38
rs752516537 Chr.19: 15949272-15949272 on Build GRCh38
rs768599148 Chr.19: 15949606-15949606 on Build GRCh38
rs115548355 Chr.19: 15949643-15949643 on Build GRCh38
rs759049418 Chr.19: 15949413-15949413 on Build GRCh38
rs746431060 Chr.19: 15949303-15949303 on Build GRCh38
rs747937693 Chr.19: 15949609-15949609 on Build GRCh38
rs771297714 Chr.19: 15949484-15949484 on Build GRCh38
rs897816385 Chr.19: 15949632-15949632 on Build GRCh38
rs765001177 Chr.19: 15949270-15949270 on Build GRCh38
rs147935606 Chr.19: 15949542-15949542 on Build GRCh38
rs759516199 Chr.19: 15949268-15949268 on Build GRCh38
rs749621084 Chr.19: 15949305-15949305 on Build GRCh38
rs375432649 Chr.19: 15949613-15949613 on Build GRCh38
rs763970552 Chr.19: 15949592-15949592 on Build GRCh38
rs748492620 Chr.19: 15949468-15949468 on Build GRCh38
rs751839001 Chr.19: 15949666-15949666 on Build GRCh38
rs1025495725 Chr.19: 15949339-15949339 on Build GRCh38
rs146994094 Chr.19: 15949535-15949535 on Build GRCh38
rs777994504 Chr.19: 15949584-15949584 on Build GRCh38
rs759640387 Chr.19: 15949651-15949651 on Build GRCh38
rs16980994 Chr.19: 15949307-15949307 on Build GRCh38
rs899490498 Chr.19: 15949644-15949644 on Build GRCh38
rs779472770 Chr.19: 15949466-15949466 on Build GRCh38
rs929570985 Chr.19: 15949636-15949636 on Build GRCh38
rs761010089 Chr.19: 15949633-15949633 on Build GRCh38
rs374738958 Chr.19: 15949347-15949347 on Build GRCh38
rs138110639 Chr.19: 15949604-15949604 on Build GRCh38
rs780558033 Chr.19: 15949371-15949371 on Build GRCh38
rs554308775 Chr.19: 15949520-15949520 on Build GRCh38
rs765214351 Chr.19: 15949297-15949298 on Build GRCh38
rs766803949 Chr.19: 15949461-15949461 on Build GRCh38
rs777932646 Chr.19: 15949289-15949289 on Build GRCh38
rs200645846 Chr.19: 15949333-15949333 on Build GRCh38
rs1038401737 Chr.19: 15949486-15949486 on Build GRCh38
rs769553270 Chr.19: 15949519-15949519 on Build GRCh38
rs755700465 Chr.19: 15949603-15949603 on Build GRCh38
rs770635041 Chr.19: 15949308-15949308 on Build GRCh38
rs757000730 Chr.19: 15949469-15949469 on Build GRCh38
rs370476895 Chr.19: 15949612-15949612 on Build GRCh38
rs756605298 Chr.19: 15949367-15949367 on Build GRCh38
rs755532471 Chr.19: 15949465-15949465 on Build GRCh38
rs148878741 Chr.19: 15949451-15949451 on Build GRCh38
rs150695702 Chr.19: 15949696-15949696 on Build GRCh38
rs1046981188 Chr.19: 15949499-15949499 on Build GRCh38
rs141816381 Chr.19: 15949683-15949683 on Build GRCh38
rs150172076 Chr.19: 15949418-15949418 on Build GRCh38
rs780850429 Chr.19: 15949477-15949477 on Build GRCh38
rs752508065 Chr.19: 15949302-15949303 on Build GRCh38
rs753136890 Chr.19: 15949561-15949561 on Build GRCh38
rs765192021 Chr.19: 15949312-15949312 on Build GRCh38
rs146373804 Chr.19: 15949714-15949714 on Build GRCh38
rs979770571 Chr.19: 15949720-15949720 on Build GRCh38
rs140667599 Chr.19: 15949401-15949401 on Build GRCh38
rs753190028 Chr.19: 15949655-15949655 on Build GRCh38
rs78394835 Chr.19: 15949300-15949300 on Build GRCh38
rs370516180 Chr.19: 15949491-15949491 on Build GRCh38
rs778393198 Chr.19: 15949694-15949694 on Build GRCh38
rs758779788 Chr.19: 15949566-15949566 on Build GRCh38
rs779417665 Chr.19: 15949384-15949384 on Build GRCh38
rs898324289 Chr.19: 15949330-15949330 on Build GRCh38
rs761544886 Chr.19: 15949504-15949504 on Build GRCh38
rs147053389 Chr.19: 15949675-15949675 on Build GRCh38
rs745384244 Chr.19: 15949478-15949478 on Build GRCh38
rs751846122 Chr.19: 15949342-15949342 on Build GRCh38
rs146434797 Chr.19: 15949337-15949337 on Build GRCh38
rs187591554 Chr.19: 15949495-15949495 on Build GRCh38
rs377654715 Chr.19: 15949538-15949538 on Build GRCh38
rs758516001 Chr.19: 15949336-15949339 on Build GRCh38
rs752466845 Chr.19: 15949319-15949319 on Build GRCh38
rs78203625 Chr.19: 15949302-15949302 on Build GRCh38
rs745590458 Chr.19: 15949596-15949596 on Build GRCh38
rs563798121 Chr.19: 15949579-15949579 on Build GRCh38
rs775429459 Chr.19: 15949420-15949420 on Build GRCh38
rs989912765 Chr.19: 15949407-15949407 on Build GRCh38
rs370722157 Chr.19: 15949706-15949706 on Build GRCh38
rs779519011 Chr.19: 15949677-15949677 on Build GRCh38
rs776270952 Chr.19: 15949310-15949310 on Build GRCh38
rs765854391 Chr.19: 15949654-15949654 on Build GRCh38
rs200715633 Chr.19: 15949271-15949271 on Build GRCh38
rs749272961 Chr.19: 15949492-15949492 on Build GRCh38
rs759442965 Chr.19: 15949311-15949311 on Build GRCh38
rs140031931 Chr.19: 15949707-15949707 on Build GRCh38
rs16980822 Chr.19: 15949438-15949438 on Build GRCh38

View Full Product Details