Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16325133-16325629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF2
Entrez Gene ID
10365
Gene Name
Kruppel like factor 2
Gene Aliases
LKLF
UniGene
Hs.685136
Gene Chromosome Location
Chr.19: 16324826-16328662 on Build GRCh38
Transcripts
NM_016270.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7256094 c.175G>A p.E59K Chr.19: 16325315-16325315 on Build GRCh38
5791601 c.151A>G p.M51V Chr.19: 16325291-16325291 on Build GRCh38
4131353 c.311T>C p.L104P Chr.19: 16325451-16325451 on Build GRCh38
7219396 c.87C>G p.R29R Chr.19: 16325227-16325227 on Build GRCh38
3357096 c.145C>G p.L49V Chr.19: 16325285-16325285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774003652 Chr.19: 16325459-16325459 on Build GRCh38
rs746535872 Chr.19: 16325209-16325209 on Build GRCh38
rs925532181 Chr.19: 16325576-16325576 on Build GRCh38
rs757297362 Chr.19: 16325304-16325304 on Build GRCh38
rs545243384 Chr.19: 16325455-16325455 on Build GRCh38
rs762685036 Chr.19: 16325293-16325293 on Build GRCh38
rs535621242 Chr.19: 16325255-16325255 on Build GRCh38
rs914484327 Chr.19: 16325156-16325156 on Build GRCh38
rs922229917 Chr.19: 16325340-16325340 on Build GRCh38
rs982550376 Chr.19: 16325305-16325305 on Build GRCh38
rs3181011 Chr.19: 16325564-16325564 on Build GRCh38
rs45595636 Chr.19: 16325164-16325164 on Build GRCh38
rs745345261 Chr.19: 16325430-16325430 on Build GRCh38
rs995855849 Chr.19: 16325503-16325503 on Build GRCh38
rs777576062 Chr.19: 16325404-16325404 on Build GRCh38
rs969671856 Chr.19: 16325251-16325251 on Build GRCh38
rs1063905 Chr.19: 16325389-16325389 on Build GRCh38
rs943079736 Chr.19: 16325175-16325175 on Build GRCh38
rs1023559821 Chr.19: 16325201-16325201 on Build GRCh38
rs775365299 Chr.19: 16325452-16325452 on Build GRCh38
rs555916400 Chr.19: 16325278-16325278 on Build GRCh38
rs767604411 Chr.19: 16325472-16325472 on Build GRCh38
rs969344834 Chr.19: 16325484-16325484 on Build GRCh38
rs774636435 Chr.19: 16325333-16325344 on Build GRCh38
rs904054692 Chr.19: 16325399-16325399 on Build GRCh38
rs750566660 Chr.19: 16325483-16325483 on Build GRCh38
rs982602898 Chr.19: 16325531-16325531 on Build GRCh38
rs865852677 Chr.19: 16325313-16325313 on Build GRCh38
rs947055171 Chr.19: 16325571-16325571 on Build GRCh38
rs901623443 Chr.19: 16325366-16325366 on Build GRCh38
rs772953981 Chr.19: 16325197-16325197 on Build GRCh38
rs760789877 Chr.19: 16325525-16325525 on Build GRCh38
rs1013590216 Chr.19: 16325469-16325469 on Build GRCh38
rs770411886 Chr.19: 16325212-16325212 on Build GRCh38
rs767693885 Chr.19: 16325311-16325311 on Build GRCh38
rs780791292 Chr.19: 16325418-16325418 on Build GRCh38
rs377405135 Chr.19: 16325173-16325173 on Build GRCh38
rs575772626 Chr.19: 16325414-16325414 on Build GRCh38
rs780634579 Chr.19: 16325169-16325169 on Build GRCh38
rs867893660 Chr.19: 16325295-16325295 on Build GRCh38
rs935510326 Chr.19: 16325590-16325590 on Build GRCh38
rs756287258 Chr.19: 16325211-16325212 on Build GRCh38
rs896592284 Chr.19: 16325608-16325608 on Build GRCh38
rs771902374 Chr.19: 16325187-16325187 on Build GRCh38
rs932198908 Chr.19: 16325347-16325347 on Build GRCh38
rs768902221 Chr.19: 16325328-16325329 on Build GRCh38
rs751736338 Chr.19: 16325297-16325297 on Build GRCh38
rs750465917 Chr.19: 16325321-16325321 on Build GRCh38
rs868428153 Chr.19: 16325228-16325228 on Build GRCh38
rs775987539 Chr.19: 16325258-16325258 on Build GRCh38
rs780246337 Chr.19: 16325329-16325331 on Build GRCh38
rs1026883554 Chr.19: 16325523-16325523 on Build GRCh38
rs768382004 Chr.19: 16325456-16325456 on Build GRCh38
rs3745319 Chr.19: 16325565-16325565 on Build GRCh38
rs951180057 Chr.19: 16325291-16325291 on Build GRCh38
rs746618681 Chr.19: 16325412-16325412 on Build GRCh38
rs45586032 Chr.19: 16325574-16325574 on Build GRCh38
rs749516918 Chr.19: 16325329-16325337 on Build GRCh38
rs1031028435 Chr.19: 16325165-16325165 on Build GRCh38
rs780686081 Chr.19: 16325342-16325342 on Build GRCh38
rs951231105 Chr.19: 16325528-16325528 on Build GRCh38
rs762162306 Chr.19: 16325468-16325468 on Build GRCh38
rs1052513234 Chr.19: 16325596-16325596 on Build GRCh38
rs912318503 Chr.19: 16325539-16325539 on Build GRCh38
rs754279560 Chr.19: 16325393-16325393 on Build GRCh38
rs765125372 Chr.19: 16325273-16325273 on Build GRCh38
rs756727179 Chr.19: 16325327-16325327 on Build GRCh38
rs865931278 Chr.19: 16325281-16325281 on Build GRCh38
rs764329434 Chr.19: 16325296-16325296 on Build GRCh38
rs949469924 Chr.19: 16325610-16325610 on Build GRCh38
rs3181012 Chr.19: 16325384-16325384 on Build GRCh38
rs748543995 Chr.19: 16325336-16325344 on Build GRCh38
rs965102587 Chr.19: 16325551-16325551 on Build GRCh38
rs919725556 Chr.19: 16325284-16325284 on Build GRCh38
rs759581494 Chr.19: 16325268-16325268 on Build GRCh38
rs755357629 Chr.19: 16325403-16325403 on Build GRCh38
rs772362936 Chr.19: 16325527-16325529 on Build GRCh38

View Full Product Details