Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16483957-16484499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALR3
Entrez Gene ID
125972
Gene Name
calreticulin 3
Gene Aliases
CMH19, CRT2, CT93
UniGene
Hs.304020
Gene Chromosome Location
Chr.19: 16479057-16496192 on Build GRCh38
Transcripts
NM_145046.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992462 c.588A>G p.I196M Chr.19: 16484020-16484020 on Build GRCh38
992461 c.591G>T p.E197D Chr.19: 16484017-16484017 on Build GRCh38
4279129 c.641C>T p.S214L Chr.19: 16483967-16483967 on Build GRCh38
474301 c.578C>A p.S193Y Chr.19: 16484030-16484030 on Build GRCh38
3530025 c.557T>C p.I186T Chr.19: 16484051-16484051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566199743 Chr.19: 16484121-16484121 on Build GRCh38
rs776906320 Chr.19: 16483999-16483999 on Build GRCh38
rs1036345230 Chr.19: 16484270-16484270 on Build GRCh38
rs765076974 Chr.19: 16484161-16484161 on Build GRCh38
rs773013367 Chr.19: 16484139-16484140 on Build GRCh38
rs1035181108 Chr.19: 16484063-16484063 on Build GRCh38
rs953217374 Chr.19: 16484173-16484173 on Build GRCh38
rs780912191 Chr.19: 16484115-16484115 on Build GRCh38
rs111870908 Chr.19: 16483989-16483989 on Build GRCh38
rs912660935 Chr.19: 16484179-16484179 on Build GRCh38
rs1021889409 Chr.19: 16484459-16484459 on Build GRCh38
rs527766637 Chr.19: 16484183-16484183 on Build GRCh38
rs557839212 Chr.19: 16484211-16484211 on Build GRCh38
rs765033019 Chr.19: 16484143-16484143 on Build GRCh38
rs574421825 Chr.19: 16484333-16484333 on Build GRCh38
rs201047203 Chr.19: 16484138-16484138 on Build GRCh38
rs557232322 Chr.19: 16484174-16484174 on Build GRCh38
rs779172220 Chr.19: 16484247-16484247 on Build GRCh38
rs760562255 Chr.19: 16484179-16484180 on Build GRCh38
rs758005878 Chr.19: 16484168-16484178 on Build GRCh38
rs986780164 Chr.19: 16484406-16484406 on Build GRCh38
rs1023463085 Chr.19: 16484355-16484355 on Build GRCh38
rs376733366 Chr.19: 16484100-16484100 on Build GRCh38
rs940781478 Chr.19: 16484255-16484255 on Build GRCh38
rs746871999 Chr.19: 16483982-16483982 on Build GRCh38
rs759282607 Chr.19: 16484139-16484139 on Build GRCh38
rs145046810 Chr.19: 16484014-16484014 on Build GRCh38
rs374187284 Chr.19: 16484091-16484091 on Build GRCh38
rs775336557 Chr.19: 16484161-16484162 on Build GRCh38
rs762301226 Chr.19: 16484047-16484047 on Build GRCh38
rs749422682 Chr.19: 16484131-16484131 on Build GRCh38
rs764707663 Chr.19: 16484028-16484028 on Build GRCh38
rs1009292171 Chr.19: 16484454-16484454 on Build GRCh38
rs71334622 Chr.19: 16484179-16484179 on Build GRCh38
rs763498941 Chr.19: 16484161-16484161 on Build GRCh38
rs149108772 Chr.19: 16484029-16484029 on Build GRCh38
rs552364605 Chr.19: 16484092-16484092 on Build GRCh38
rs776266496 Chr.19: 16484157-16484158 on Build GRCh38
rs973943547 Chr.19: 16484368-16484368 on Build GRCh38
rs902159738 Chr.19: 16484300-16484300 on Build GRCh38
rs763030941 Chr.19: 16484162-16484179 on Build GRCh38
rs746613806 Chr.19: 16484155-16484156 on Build GRCh38
rs756711263 Chr.19: 16484163-16484164 on Build GRCh38
rs1039874788 Chr.19: 16484223-16484223 on Build GRCh38
rs577084803 Chr.19: 16484178-16484178 on Build GRCh38
rs999218527 Chr.19: 16484309-16484309 on Build GRCh38
rs912597954 Chr.19: 16484422-16484422 on Build GRCh38
rs1003780395 Chr.19: 16484345-16484345 on Build GRCh38
rs780712291 Chr.19: 16484167-16484168 on Build GRCh38
rs183655549 Chr.19: 16484281-16484281 on Build GRCh38
rs752204583 Chr.19: 16484097-16484097 on Build GRCh38
rs535147112 Chr.19: 16484127-16484127 on Build GRCh38
rs774977077 Chr.19: 16484133-16484133 on Build GRCh38
rs918533406 Chr.19: 16484397-16484397 on Build GRCh38
rs539662798 Chr.19: 16484230-16484230 on Build GRCh38
rs7252242 Chr.19: 16484168-16484168 on Build GRCh38
rs922091764 Chr.19: 16484231-16484231 on Build GRCh38
rs772004043 Chr.19: 16483976-16483976 on Build GRCh38
rs746917889 Chr.19: 16484108-16484108 on Build GRCh38
rs778627769 Chr.19: 16484038-16484041 on Build GRCh38
rs150143294 Chr.19: 16484232-16484232 on Build GRCh38
rs768610670 Chr.19: 16484132-16484132 on Build GRCh38
rs942806764 Chr.19: 16484442-16484442 on Build GRCh38
rs962902610 Chr.19: 16484366-16484366 on Build GRCh38
rs775484097 Chr.19: 16484007-16484007 on Build GRCh38
rs769879360 Chr.19: 16484004-16484004 on Build GRCh38
rs532420030 Chr.19: 16484469-16484469 on Build GRCh38
rs1034788641 Chr.19: 16484017-16484017 on Build GRCh38
rs892199678 Chr.19: 16484396-16484396 on Build GRCh38
rs960492198 Chr.19: 16484324-16484324 on Build GRCh38
rs143671909 Chr.19: 16484088-16484088 on Build GRCh38
rs747656642 Chr.19: 16484044-16484044 on Build GRCh38
rs368704371 Chr.19: 16484253-16484253 on Build GRCh38
rs10413787 Chr.19: 16484156-16484156 on Build GRCh38
rs762165230 Chr.19: 16484137-16484137 on Build GRCh38
rs930518209 Chr.19: 16484314-16484314 on Build GRCh38
rs369476772 Chr.19: 16484062-16484062 on Build GRCh38
rs767978157 Chr.19: 16484163-16484178 on Build GRCh38
rs150376706 Chr.19: 16484000-16484000 on Build GRCh38
rs752368241 Chr.19: 16484153-16484153 on Build GRCh38
rs369414539 Chr.19: 16484168-16484168 on Build GRCh38
rs777454147 Chr.19: 16483981-16483981 on Build GRCh38
rs951355699 Chr.19: 16484403-16484403 on Build GRCh38
rs769966759 Chr.19: 16484128-16484128 on Build GRCh38
rs775571848 Chr.19: 16484130-16484130 on Build GRCh38
rs1034737026 Chr.19: 16484317-16484317 on Build GRCh38
rs770798077 Chr.19: 16484158-16484174 on Build GRCh38
rs968920652 Chr.19: 16484216-16484216 on Build GRCh38
rs543401822 Chr.19: 16484344-16484344 on Build GRCh38
rs138628911 Chr.19: 16484468-16484468 on Build GRCh38
rs750744314 Chr.19: 16484164-16484178 on Build GRCh38
rs771642699 Chr.19: 16484127-16484128 on Build GRCh38
rs560811635 Chr.19: 16484318-16484318 on Build GRCh38
rs1056329098 Chr.19: 16484384-16484384 on Build GRCh38

View Full Product Details