Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17603274-17603801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC13A
Entrez Gene ID
23025
Gene Name
unc-13 homolog A
Gene Aliases
Munc13-1
UniGene
Hs.164502
Gene Chromosome Location
Chr.19: 17601328-17688344 on Build GRCh38
Transcripts
NM_001080421.2 XM_011527810.2 XM_011527811.2 XM_017026502.1

SNP Detail

SNP ID Chromosome Location
rs8100532 Chr.19: 17603777-17603777 on Build GRCh38
rs537903203 Chr.19: 17603698-17603698 on Build GRCh38
rs900689350 Chr.19: 17603305-17603305 on Build GRCh38
rs547415692 Chr.19: 17603557-17603557 on Build GRCh38
rs145398602 Chr.19: 17603341-17603341 on Build GRCh38
rs535710097 Chr.19: 17603332-17603332 on Build GRCh38
rs867528130 Chr.19: 17603349-17603349 on Build GRCh38
rs146603005 Chr.19: 17603744-17603744 on Build GRCh38
rs778246882 Chr.19: 17603390-17603390 on Build GRCh38
rs1005290990 Chr.19: 17603385-17603385 on Build GRCh38
rs565242714 Chr.19: 17603522-17603522 on Build GRCh38
rs183852948 Chr.19: 17603420-17603420 on Build GRCh38
rs974503045 Chr.19: 17603519-17603519 on Build GRCh38
rs867547518 Chr.19: 17603658-17603658 on Build GRCh38
rs755634077 Chr.19: 17603304-17603304 on Build GRCh38
rs866998558 Chr.19: 17603423-17603423 on Build GRCh38
rs1036021526 Chr.19: 17603450-17603450 on Build GRCh38
rs1007430713 Chr.19: 17603439-17603439 on Build GRCh38
rs1021198730 Chr.19: 17603734-17603734 on Build GRCh38
rs529435457 Chr.19: 17603483-17603483 on Build GRCh38
rs527707358 Chr.19: 17603544-17603544 on Build GRCh38
rs182382903 Chr.19: 17603583-17603583 on Build GRCh38
rs933486791 Chr.19: 17603528-17603528 on Build GRCh38
rs867404103 Chr.19: 17603594-17603594 on Build GRCh38
rs988868421 Chr.19: 17603486-17603486 on Build GRCh38
rs968380911 Chr.19: 17603684-17603684 on Build GRCh38
rs563117369 Chr.19: 17603394-17603394 on Build GRCh38
rs922826274 Chr.19: 17603591-17603591 on Build GRCh38
rs557580271 Chr.19: 17603701-17603701 on Build GRCh38
rs746323593 Chr.19: 17603573-17603573 on Build GRCh38
rs1032224544 Chr.19: 17603449-17603449 on Build GRCh38
rs560750798 Chr.19: 17603337-17603337 on Build GRCh38
rs1009465765 Chr.19: 17603733-17603733 on Build GRCh38
rs866531535 Chr.19: 17603624-17603624 on Build GRCh38
rs145762295 Chr.19: 17603645-17603645 on Build GRCh38
rs899190302 Chr.19: 17603566-17603566 on Build GRCh38
rs568971797 Chr.19: 17603722-17603722 on Build GRCh38
rs893704212 Chr.19: 17603710-17603710 on Build GRCh38
rs970869949 Chr.19: 17603535-17603535 on Build GRCh38
rs867333008 Chr.19: 17603347-17603347 on Build GRCh38
rs1049879207 Chr.19: 17603625-17603625 on Build GRCh38
rs988761175 Chr.19: 17603655-17603655 on Build GRCh38
rs1012309756 Chr.19: 17603414-17603414 on Build GRCh38
rs568958631 Chr.19: 17603671-17603671 on Build GRCh38
rs753516958 Chr.19: 17603336-17603336 on Build GRCh38
rs529189580 Chr.19: 17603616-17603616 on Build GRCh38
rs768741070 Chr.19: 17603740-17603740 on Build GRCh38
rs781762077 Chr.19: 17603532-17603532 on Build GRCh38
rs914261929 Chr.19: 17603488-17603488 on Build GRCh38
rs969880859 Chr.19: 17603440-17603440 on Build GRCh38
rs1003664208 Chr.19: 17603771-17603771 on Build GRCh38
rs779472316 Chr.19: 17603321-17603321 on Build GRCh38
rs754534606 Chr.19: 17603348-17603348 on Build GRCh38
rs557200021 Chr.19: 17603540-17603540 on Build GRCh38
rs936015564 Chr.19: 17603664-17603664 on Build GRCh38
rs1009370871 Chr.19: 17603462-17603462 on Build GRCh38
rs981348634 Chr.19: 17603444-17603444 on Build GRCh38
rs940982986 Chr.19: 17603550-17603550 on Build GRCh38
rs186715514 Chr.19: 17603457-17603457 on Build GRCh38
rs1012259206 Chr.19: 17603569-17603569 on Build GRCh38
rs889830559 Chr.19: 17603437-17603437 on Build GRCh38
rs910149910 Chr.19: 17603577-17603577 on Build GRCh38
rs1037091456 Chr.19: 17603312-17603312 on Build GRCh38
rs947093714 Chr.19: 17603513-17603513 on Build GRCh38
rs934064048 Chr.19: 17603599-17603599 on Build GRCh38
rs769998467 Chr.19: 17603580-17603580 on Build GRCh38
rs191459813 Chr.19: 17603559-17603559 on Build GRCh38
rs771537981 Chr.19: 17603477-17603477 on Build GRCh38
rs865938925 Chr.19: 17603402-17603402 on Build GRCh38

View Full Product Details