Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18067998-18068540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
UniGene
Hs.567294
Gene Chromosome Location
Chr.19: 18058994-18099027 on Build GRCh38
Transcripts
NM_001290023.1 NM_001290024.1 NM_005535.2 XM_006722741.3 XM_011527966.2 XM_011527967.2 XM_011527968.2 XM_011527969.2 XM_011527970.2 XM_011527971.2 XM_011527972.2 XM_011527973.2 XM_011527974.2 XM_011527975.2 XM_011527976.2 XM_017026762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4075560 c.1217G>T p.G406V Chr.19: 18068499-18068499 on Build GRCh38
4075558 c.1260C>G p.A420A Chr.19: 18068456-18068456 on Build GRCh38
992990 c.1295C>T p.T432M Chr.19: 18068421-18068421 on Build GRCh38
5909717 c.1275G>T p.E425D Chr.19: 18068441-18068441 on Build GRCh38
6684849 c.1268A>C p.H423P Chr.19: 18068448-18068448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550155096 Chr.19: 18068336-18068336 on Build GRCh38
rs760969127 Chr.19: 18068346-18068346 on Build GRCh38
rs996544775 Chr.19: 18068337-18068337 on Build GRCh38
rs775923325 Chr.19: 18068443-18068443 on Build GRCh38
rs779947911 Chr.19: 18068395-18068395 on Build GRCh38
rs913507246 Chr.19: 18068455-18068455 on Build GRCh38
rs754764047 Chr.19: 18068466-18068466 on Build GRCh38
rs752642420 Chr.19: 18068475-18068475 on Build GRCh38
rs758241253 Chr.19: 18068486-18068486 on Build GRCh38
rs759893194 Chr.19: 18068431-18068431 on Build GRCh38
rs567872726 Chr.19: 18068302-18068302 on Build GRCh38
rs938987168 Chr.19: 18068223-18068223 on Build GRCh38
rs774555779 Chr.19: 18068516-18068516 on Build GRCh38
rs986591115 Chr.19: 18068112-18068112 on Build GRCh38
rs774271493 Chr.19: 18068450-18068450 on Build GRCh38
rs367647802 Chr.19: 18068447-18068447 on Build GRCh38
rs780762892 Chr.19: 18068494-18068494 on Build GRCh38
rs17882076 Chr.19: 18068153-18068153 on Build GRCh38
rs757004204 Chr.19: 18068493-18068493 on Build GRCh38
rs772864032 Chr.19: 18068339-18068339 on Build GRCh38
rs376271 Chr.19: 18068474-18068474 on Build GRCh38
rs771562593 Chr.19: 18068416-18068416 on Build GRCh38
rs757390309 Chr.19: 18068393-18068393 on Build GRCh38
rs181757087 Chr.19: 18068117-18068117 on Build GRCh38
rs767846981 Chr.19: 18068451-18068451 on Build GRCh38
rs777395626 Chr.19: 18068487-18068487 on Build GRCh38
rs1043211799 Chr.19: 18068492-18068492 on Build GRCh38
rs866509098 Chr.19: 18068044-18068044 on Build GRCh38
rs930950631 Chr.19: 18068175-18068175 on Build GRCh38
rs1008207269 Chr.19: 18068279-18068279 on Build GRCh38
rs190491500 Chr.19: 18068390-18068390 on Build GRCh38
rs1037897928 Chr.19: 18068179-18068179 on Build GRCh38
rs375654921 Chr.19: 18068404-18068404 on Build GRCh38
rs747676268 Chr.19: 18068410-18068410 on Build GRCh38
rs899879403 Chr.19: 18068292-18068292 on Build GRCh38
rs745398503 Chr.19: 18068277-18068277 on Build GRCh38
rs867396174 Chr.19: 18068433-18068433 on Build GRCh38
rs547323080 Chr.19: 18068236-18068236 on Build GRCh38
rs559106838 Chr.19: 18068420-18068420 on Build GRCh38
rs779406776 Chr.19: 18068111-18068111 on Build GRCh38
rs1044076780 Chr.19: 18068182-18068182 on Build GRCh38
rs972579837 Chr.19: 18068432-18068432 on Build GRCh38
rs777322101 Chr.19: 18068417-18068417 on Build GRCh38
rs751760867 Chr.19: 18068391-18068391 on Build GRCh38
rs527312275 Chr.19: 18068186-18068186 on Build GRCh38
rs924310151 Chr.19: 18068118-18068118 on Build GRCh38
rs201548803 Chr.19: 18068454-18068454 on Build GRCh38
rs748901032 Chr.19: 18068271-18068271 on Build GRCh38
rs770296365 Chr.19: 18068437-18068437 on Build GRCh38
rs777041534 Chr.19: 18068423-18068423 on Build GRCh38
rs1047626022 Chr.19: 18068275-18068275 on Build GRCh38
rs185883069 Chr.19: 18068380-18068380 on Build GRCh38
rs753670992 Chr.19: 18068397-18068397 on Build GRCh38
rs148549363 Chr.19: 18068129-18068129 on Build GRCh38
rs1019723661 Chr.19: 18068291-18068291 on Build GRCh38
rs1043101659 Chr.19: 18068306-18068306 on Build GRCh38
rs972505046 Chr.19: 18068140-18068140 on Build GRCh38
rs775796804 Chr.19: 18068506-18068506 on Build GRCh38
rs531336116 Chr.19: 18068124-18068124 on Build GRCh38
rs845379 Chr.19: 18068305-18068305 on Build GRCh38
rs199504990 Chr.19: 18068499-18068499 on Build GRCh38
rs746490724 Chr.19: 18068489-18068489 on Build GRCh38
rs199755161 Chr.19: 18068444-18068444 on Build GRCh38
rs202048215 Chr.19: 18068508-18068508 on Build GRCh38
rs969415632 Chr.19: 18068089-18068089 on Build GRCh38
rs771290769 Chr.19: 18068351-18068351 on Build GRCh38
rs776953409 Chr.19: 18068361-18068361 on Build GRCh38
rs539168120 Chr.19: 18068407-18068407 on Build GRCh38
rs367944399 Chr.19: 18068389-18068389 on Build GRCh38
rs927498004 Chr.19: 18068187-18068187 on Build GRCh38
rs747639226 Chr.19: 18068375-18068376 on Build GRCh38
rs200875188 Chr.19: 18068421-18068421 on Build GRCh38

View Full Product Details