Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18612730-18613223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM59L
Entrez Gene ID
25789
Gene Name
transmembrane protein 59 like
Gene Aliases
BSMAP, C19orf4
UniGene
Hs.329850
Gene Chromosome Location
Chr.19: 18612872-18621039 on Build GRCh38
Transcripts
NM_012109.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5567950 c.81C>T p.S27S Chr.19: 18613039-18613039 on Build GRCh38
7332530 c.46C>T p.L16L Chr.19: 18613004-18613004 on Build GRCh38
3388739 c.109G>A p.G37R Chr.19: 18613067-18613067 on Build GRCh38
6036402 c.98delC p.Q35fs*39 Chr.19: 18613056-18613056 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032122252 Chr.19: 18613032-18613032 on Build GRCh38
rs908691767 Chr.19: 18612952-18612952 on Build GRCh38
rs964024303 Chr.19: 18613116-18613116 on Build GRCh38
rs998659338 Chr.19: 18612808-18612808 on Build GRCh38
rs1011945767 Chr.19: 18613111-18613111 on Build GRCh38
rs553999558 Chr.19: 18613121-18613121 on Build GRCh38
rs887652259 Chr.19: 18612819-18612819 on Build GRCh38
rs1045765171 Chr.19: 18612807-18612807 on Build GRCh38
rs772823631 Chr.19: 18612963-18612963 on Build GRCh38
rs756039506 Chr.19: 18612978-18612979 on Build GRCh38
rs1005239239 Chr.19: 18612753-18612753 on Build GRCh38
rs892961368 Chr.19: 18613061-18613061 on Build GRCh38
rs888829119 Chr.19: 18613002-18613002 on Build GRCh38
rs760956959 Chr.19: 18613144-18613144 on Build GRCh38
rs906927809 Chr.19: 18613020-18613020 on Build GRCh38
rs113891862 Chr.19: 18613122-18613122 on Build GRCh38
rs997143603 Chr.19: 18612987-18612988 on Build GRCh38
rs761504820 Chr.19: 18612939-18612939 on Build GRCh38
rs913218859 Chr.19: 18612781-18612781 on Build GRCh38
rs1017480396 Chr.19: 18613115-18613115 on Build GRCh38
rs1018858090 Chr.19: 18612884-18612884 on Build GRCh38
rs773717742 Chr.19: 18612775-18612775 on Build GRCh38
rs768483704 Chr.19: 18612908-18612908 on Build GRCh38
rs548732316 Chr.19: 18612818-18612818 on Build GRCh38
rs1014872966 Chr.19: 18612829-18612829 on Build GRCh38
rs1048332661 Chr.19: 18612977-18612978 on Build GRCh38
rs957586645 Chr.19: 18612885-18612885 on Build GRCh38
rs897762213 Chr.19: 18612987-18612987 on Build GRCh38
rs971225919 Chr.19: 18612792-18612792 on Build GRCh38
rs897481274 Chr.19: 18612835-18612835 on Build GRCh38
rs777923741 Chr.19: 18613149-18613149 on Build GRCh38
rs748071337 Chr.19: 18612988-18612990 on Build GRCh38
rs957743093 Chr.19: 18613035-18613035 on Build GRCh38
rs928622806 Chr.19: 18613187-18613187 on Build GRCh38
rs934175632 Chr.19: 18612929-18612929 on Build GRCh38
rs11538357 Chr.19: 18613106-18613106 on Build GRCh38
rs534023153 Chr.19: 18612988-18612988 on Build GRCh38
rs953249255 Chr.19: 18612841-18612841 on Build GRCh38
rs946557559 Chr.19: 18612883-18612883 on Build GRCh38
rs537557252 Chr.19: 18612826-18612826 on Build GRCh38
rs1030059711 Chr.19: 18612840-18612840 on Build GRCh38
rs922655881 Chr.19: 18612922-18612922 on Build GRCh38
rs1012706939 Chr.19: 18612776-18612776 on Build GRCh38
rs985934608 Chr.19: 18612875-18612875 on Build GRCh38
rs911446948 Chr.19: 18612751-18612751 on Build GRCh38
rs967612950 Chr.19: 18613173-18613173 on Build GRCh38
rs957540488 Chr.19: 18612816-18612816 on Build GRCh38
rs946080792 Chr.19: 18612791-18612791 on Build GRCh38
rs958757641 Chr.19: 18613195-18613195 on Build GRCh38
rs997196164 Chr.19: 18612836-18612836 on Build GRCh38
rs1056041741 Chr.19: 18612815-18612815 on Build GRCh38
rs535735218 Chr.19: 18612777-18612777 on Build GRCh38
rs112432990 Chr.19: 18612825-18612825 on Build GRCh38
rs367939047 Chr.19: 18612954-18612954 on Build GRCh38
rs577762498 Chr.19: 18612920-18612920 on Build GRCh38
rs982892736 Chr.19: 18612938-18612938 on Build GRCh38
rs990350661 Chr.19: 18612914-18612914 on Build GRCh38
rs975357260 Chr.19: 18613135-18613135 on Build GRCh38
rs772594729 Chr.19: 18612771-18612771 on Build GRCh38
rs998587247 Chr.19: 18613031-18613031 on Build GRCh38
rs753347572 Chr.19: 18613096-18613096 on Build GRCh38
rs557790917 Chr.19: 18612859-18612859 on Build GRCh38
rs894127805 Chr.19: 18612764-18612764 on Build GRCh38
rs568809726 Chr.19: 18612823-18612823 on Build GRCh38
rs765831156 Chr.19: 18613036-18613036 on Build GRCh38
rs915160079 Chr.19: 18612765-18612765 on Build GRCh38
rs1038610920 Chr.19: 18612968-18612968 on Build GRCh38
rs1031606018 Chr.19: 18612814-18612814 on Build GRCh38
rs894172356 Chr.19: 18612970-18612970 on Build GRCh38
rs574018363 Chr.19: 18613158-18613158 on Build GRCh38
rs373764609 Chr.19: 18612755-18612755 on Build GRCh38
rs11538358 Chr.19: 18613005-18613005 on Build GRCh38
rs11538360 Chr.19: 18613103-18613103 on Build GRCh38
rs964074004 Chr.19: 18612837-18612837 on Build GRCh38
rs760338894 Chr.19: 18612986-18612986 on Build GRCh38
rs1045520475 Chr.19: 18613015-18613015 on Build GRCh38

View Full Product Details