Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19105479-19105974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A42
Entrez Gene ID
284439
Gene Name
solute carrier family 25 member 42
Gene Aliases
-
UniGene
Hs.303669
Gene Chromosome Location
Chr.19: 19063961-19113030 on Build GRCh38
Transcripts
NM_001321544.1 NM_178526.4 XM_005259861.3 XM_011527937.2 XM_017026670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993237 c.309C>T p.Y103Y Chr.19: 19105656-19105656 on Build GRCh38
6253415 c.250G>T p.E84* Chr.19: 19105597-19105597 on Build GRCh38
5748023 c.227T>C p.V76A Chr.19: 19105574-19105574 on Build GRCh38
4849626 c.231C>G p.L77L Chr.19: 19105578-19105578 on Build GRCh38
1304217 c.246C>T p.L82L Chr.19: 19105593-19105593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773263941 Chr.19: 19105554-19105554 on Build GRCh38
rs750048068 Chr.19: 19105692-19105692 on Build GRCh38
rs538380471 Chr.19: 19105854-19105854 on Build GRCh38
rs200500398 Chr.19: 19105593-19105593 on Build GRCh38
rs916048583 Chr.19: 19105675-19105675 on Build GRCh38
rs779734850 Chr.19: 19105629-19105629 on Build GRCh38
rs960278892 Chr.19: 19105663-19105663 on Build GRCh38
rs764158605 Chr.19: 19105683-19105683 on Build GRCh38
rs758371554 Chr.19: 19105872-19105872 on Build GRCh38
rs571742122 Chr.19: 19105780-19105780 on Build GRCh38
rs141712568 Chr.19: 19105703-19105703 on Build GRCh38
rs531748523 Chr.19: 19105660-19105660 on Build GRCh38
rs771181259 Chr.19: 19105721-19105721 on Build GRCh38
rs923609157 Chr.19: 19105759-19105759 on Build GRCh38
rs767453952 Chr.19: 19105691-19105691 on Build GRCh38
rs1051477020 Chr.19: 19105768-19105768 on Build GRCh38
rs967768416 Chr.19: 19105738-19105738 on Build GRCh38
rs758777237 Chr.19: 19105914-19105914 on Build GRCh38
rs145552056 Chr.19: 19105641-19105641 on Build GRCh38
rs977771341 Chr.19: 19105743-19105743 on Build GRCh38
rs766495888 Chr.19: 19105724-19105724 on Build GRCh38
rs758381313 Chr.19: 19105571-19105571 on Build GRCh38
rs530254742 Chr.19: 19105826-19105826 on Build GRCh38
rs774305075 Chr.19: 19105771-19105771 on Build GRCh38
rs772332647 Chr.19: 19105658-19105658 on Build GRCh38
rs769768551 Chr.19: 19105762-19105762 on Build GRCh38
rs997763925 Chr.19: 19105841-19105841 on Build GRCh38
rs138036287 Chr.19: 19105645-19105645 on Build GRCh38
rs760915278 Chr.19: 19105668-19105668 on Build GRCh38
rs1037729944 Chr.19: 19105716-19105716 on Build GRCh38
rs753469566 Chr.19: 19105701-19105701 on Build GRCh38
rs1030620937 Chr.19: 19105866-19105866 on Build GRCh38
rs779681889 Chr.19: 19105696-19105696 on Build GRCh38
rs776509686 Chr.19: 19105725-19105725 on Build GRCh38
rs941856943 Chr.19: 19105515-19105515 on Build GRCh38
rs1034656205 Chr.19: 19105888-19105888 on Build GRCh38
rs902298888 Chr.19: 19105865-19105865 on Build GRCh38
rs776687491 Chr.19: 19105736-19105736 on Build GRCh38
rs755883383 Chr.19: 19105693-19105693 on Build GRCh38
rs10420684 Chr.19: 19105790-19105790 on Build GRCh38
rs755647825 Chr.19: 19105611-19105611 on Build GRCh38
rs552143875 Chr.19: 19105883-19105883 on Build GRCh38
rs759718694 Chr.19: 19105761-19105761 on Build GRCh38
rs762767863 Chr.19: 19105681-19105681 on Build GRCh38
rs763772024 Chr.19: 19105581-19105581 on Build GRCh38
rs748604140 Chr.19: 19105705-19105705 on Build GRCh38
rs569714578 Chr.19: 19105838-19105838 on Build GRCh38
rs933649216 Chr.19: 19105764-19105764 on Build GRCh38
rs1038984189 Chr.19: 19105797-19105797 on Build GRCh38
rs374467322 Chr.19: 19105522-19105522 on Build GRCh38
rs761913865 Chr.19: 19105548-19105548 on Build GRCh38
rs12610978 Chr.19: 19105840-19105840 on Build GRCh38
rs371330383 Chr.19: 19105546-19105546 on Build GRCh38
rs767851398 Chr.19: 19105549-19105549 on Build GRCh38
rs375907273 Chr.19: 19105555-19105555 on Build GRCh38
rs751466309 Chr.19: 19105684-19105684 on Build GRCh38
rs765114657 Chr.19: 19105732-19105733 on Build GRCh38
rs747209186 Chr.19: 19105713-19105713 on Build GRCh38
rs900712359 Chr.19: 19105837-19105837 on Build GRCh38
rs983475762 Chr.19: 19105907-19105907 on Build GRCh38
rs749794223 Chr.19: 19105632-19105632 on Build GRCh38
rs534873604 Chr.19: 19105944-19105944 on Build GRCh38
rs4808920 Chr.19: 19105804-19105804 on Build GRCh38
rs770948242 Chr.19: 19105674-19105674 on Build GRCh38
rs549575771 Chr.19: 19105835-19105835 on Build GRCh38
rs75496133 Chr.19: 19105521-19105521 on Build GRCh38
rs373671442 Chr.19: 19105802-19105802 on Build GRCh38
rs367596755 Chr.19: 19105540-19105540 on Build GRCh38
rs760686291 Chr.19: 19105556-19105556 on Build GRCh38
rs745521216 Chr.19: 19105610-19105610 on Build GRCh38
rs990635680 Chr.19: 19105942-19105942 on Build GRCh38
rs776738600 Chr.19: 19105676-19105676 on Build GRCh38
rs564985191 Chr.19: 19105786-19105786 on Build GRCh38
rs752588368 Chr.19: 19105560-19105560 on Build GRCh38
rs542707849 Chr.19: 19105558-19105558 on Build GRCh38
rs761680761 Chr.19: 19105778-19105778 on Build GRCh38
rs916404648 Chr.19: 19105952-19105952 on Build GRCh38
rs1023195173 Chr.19: 19105949-19105949 on Build GRCh38
rs963809326 Chr.19: 19105939-19105939 on Build GRCh38
rs772439531 Chr.19: 19105706-19105706 on Build GRCh38
rs890457542 Chr.19: 19105783-19105786 on Build GRCh38
rs754874088 Chr.19: 19105904-19105904 on Build GRCh38
rs778129613 Chr.19: 19105712-19105712 on Build GRCh38
rs752702761 Chr.19: 19105596-19105596 on Build GRCh38
rs12977069 Chr.19: 19105919-19105919 on Build GRCh38
rs372578398 Chr.19: 19105583-19105583 on Build GRCh38
rs78016659 Chr.19: 19105810-19105810 on Build GRCh38

View Full Product Details