Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19146182-19146671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS8-MEF2B
Entrez Gene ID
4207
Gene Name
BORCS8-MEF2B readthrough
Gene Aliases
LOC729991-MEF2B, MEF2B, MEF2BNB-MEF2B
UniGene
Hs.153629
Gene Chromosome Location
Chr.19: 19145567-19192591 on Build GRCh38
Transcripts
NM_005919.3 NR_027307.1 NR_027308.1 NM_001145785.1

Gene Symbol
MEF2B
Entrez Gene ID
100271849
Gene Name
myocyte enhancer factor 2B
Gene Aliases
RSRFR2
UniGene
Hs.153629
Gene Chromosome Location
Chr.19: 19145567-19170289 on Build GRCh38
Transcripts
NM_005919.3 NR_027307.1 NR_027308.1 NM_001145785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6036946 c.676-10C>G p.? Chr.19: 19146658-19146658 on Build GRCh38
6955673 c.698A>T p.Q233L Chr.19: 19146626-19146626 on Build GRCh38
5677750 c.686A>G p.Y229C Chr.19: 19146638-19146638 on Build GRCh38
7192222 c.880A>T p.S294C Chr.19: 19146274-19146274 on Build GRCh38
6711064 c.747C>T p.P249P Chr.19: 19146577-19146577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756499405 Chr.19: 19146568-19146568 on Build GRCh38
rs528706676 Chr.19: 19146298-19146298 on Build GRCh38
rs754979220 Chr.19: 19146578-19146578 on Build GRCh38
rs539745270 Chr.19: 19146587-19146587 on Build GRCh38
rs767967351 Chr.19: 19146646-19146646 on Build GRCh38
rs773973419 Chr.19: 19146640-19146640 on Build GRCh38
rs999306943 Chr.19: 19146332-19146332 on Build GRCh38
rs886664033 Chr.19: 19146430-19146430 on Build GRCh38
rs750716565 Chr.19: 19146567-19146567 on Build GRCh38
rs1023222877 Chr.19: 19146484-19146484 on Build GRCh38
rs752557132 Chr.19: 19146261-19146261 on Build GRCh38
rs759478868 Chr.19: 19146227-19146227 on Build GRCh38
rs989384199 Chr.19: 19146292-19146292 on Build GRCh38
rs899163211 Chr.19: 19146482-19146482 on Build GRCh38
rs1027802105 Chr.19: 19146539-19146539 on Build GRCh38
rs761230964 Chr.19: 19146643-19146643 on Build GRCh38
rs143597015 Chr.19: 19146574-19146574 on Build GRCh38
rs530420169 Chr.19: 19146428-19146428 on Build GRCh38
rs1051958286 Chr.19: 19146359-19146359 on Build GRCh38
rs867151984 Chr.19: 19146212-19146212 on Build GRCh38
rs865989794 Chr.19: 19146476-19146476 on Build GRCh38
rs770627751 Chr.19: 19146257-19146257 on Build GRCh38
rs923830438 Chr.19: 19146259-19146259 on Build GRCh38
rs747771599 Chr.19: 19146289-19146289 on Build GRCh38
rs746905972 Chr.19: 19146591-19146591 on Build GRCh38
rs1043619674 Chr.19: 19146305-19146305 on Build GRCh38
rs1047785382 Chr.19: 19146235-19146235 on Build GRCh38
rs921954205 Chr.19: 19146634-19146634 on Build GRCh38
rs1016550700 Chr.19: 19146459-19146459 on Build GRCh38
rs914889969 Chr.19: 19146295-19146295 on Build GRCh38
rs762452595 Chr.19: 19146550-19146550 on Build GRCh38
rs775263101 Chr.19: 19146621-19146621 on Build GRCh38
rs768050646 Chr.19: 19146639-19146639 on Build GRCh38
rs900384526 Chr.19: 19146282-19146282 on Build GRCh38
rs371467297 Chr.19: 19146584-19146584 on Build GRCh38
rs1005565654 Chr.19: 19146299-19146299 on Build GRCh38
rs1005125486 Chr.19: 19146436-19146436 on Build GRCh38
rs900926644 Chr.19: 19146323-19146323 on Build GRCh38
rs547864988 Chr.19: 19146346-19146346 on Build GRCh38
rs192059129 Chr.19: 19146230-19146230 on Build GRCh38
rs546050202 Chr.19: 19146647-19146647 on Build GRCh38
rs774693538 Chr.19: 19146517-19146517 on Build GRCh38
rs1039419408 Chr.19: 19146306-19146306 on Build GRCh38
rs1043654252 Chr.19: 19146645-19146645 on Build GRCh38
rs778269452 Chr.19: 19146529-19146529 on Build GRCh38
rs546609857 Chr.19: 19146600-19146600 on Build GRCh38
rs1015160156 Chr.19: 19146334-19146334 on Build GRCh38
rs111253893 Chr.19: 19146355-19146355 on Build GRCh38
rs745619777 Chr.19: 19146615-19146615 on Build GRCh38
rs763712837 Chr.19: 19146511-19146511 on Build GRCh38
rs970747427 Chr.19: 19146490-19146490 on Build GRCh38
rs1023691199 Chr.19: 19146508-19146508 on Build GRCh38
rs769734787 Chr.19: 19146618-19146618 on Build GRCh38
rs1012288050 Chr.19: 19146463-19146463 on Build GRCh38
rs970854940 Chr.19: 19146518-19146518 on Build GRCh38
rs950872499 Chr.19: 19146560-19146560 on Build GRCh38
rs929429332 Chr.19: 19146234-19146234 on Build GRCh38
rs768022242 Chr.19: 19146562-19146562 on Build GRCh38
rs142032500 Chr.19: 19146601-19146601 on Build GRCh38
rs550161775 Chr.19: 19146500-19146500 on Build GRCh38
rs749012108 Chr.19: 19146630-19146630 on Build GRCh38
rs747005371 Chr.19: 19146253-19146253 on Build GRCh38
rs754106437 Chr.19: 19146577-19146577 on Build GRCh38

View Full Product Details