Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19201659-19202186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2C2AP
Entrez Gene ID
126382
Gene Name
nuclear receptor 2C2 associated protein
Gene Aliases
TRA16
UniGene
Hs.708153
Gene Chromosome Location
Chr.19: 19201409-19203453 on Build GRCh38
Transcripts
NM_001300945.1 NM_176880.5 XM_011527688.2 NM_001278727.1 NM_001278728.1 NM_003721.3 NM_134440.2 XM_005260134.4 XM_005260135.3 XM_005260136.4 XM_005260137.4 XM_006722930.3 XM_017027415.1 XM_017027416.1

Gene Symbol
RFXANK
Entrez Gene ID
8625
Gene Name
regulatory factor X associated ankyrin containing protein
Gene Aliases
ANKRA1, BLS, F14150_1, RFX-B
UniGene
Hs.153629
Gene Chromosome Location
Chr.19: 19192199-19201869 on Build GRCh38
Transcripts
NM_001300945.1 NM_176880.5 XM_011527688.2 NM_001278727.1 NM_001278728.1 NM_003721.3 NM_134440.2 XM_005260134.4 XM_005260135.3 XM_005260136.4 XM_005260137.4 XM_006722930.3 XM_017027415.1 XM_017027416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5883065 c.347C>T p.T116M Chr.19: 19201998-19201998 on Build GRCh38
3959747 c.409G>T p.E137* Chr.19: 19201936-19201936 on Build GRCh38
7270109 c.734A>G p.H245R Chr.19: 19201670-19201670 on Build GRCh38
6203892 c.772G>A p.D258N Chr.19: 19201708-19201708 on Build GRCh38
6582340 c.313A>G p.I105V Chr.19: 19202032-19202032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772784245 Chr.19: 19201734-19201734 on Build GRCh38
rs911228565 Chr.19: 19201923-19201923 on Build GRCh38
rs780619900 Chr.19: 19202083-19202083 on Build GRCh38
rs567107133 Chr.19: 19202099-19202099 on Build GRCh38
rs762105891 Chr.19: 19202067-19202068 on Build GRCh38
rs918643759 Chr.19: 19202142-19202142 on Build GRCh38
rs866844773 Chr.19: 19201946-19201946 on Build GRCh38
rs763024000 Chr.19: 19201956-19201956 on Build GRCh38
rs369432703 Chr.19: 19201969-19201969 on Build GRCh38
rs776943497 Chr.19: 19201742-19201742 on Build GRCh38
rs150729244 Chr.19: 19201714-19201714 on Build GRCh38
rs139450000 Chr.19: 19201988-19201988 on Build GRCh38
rs1050474 Chr.19: 19201839-19201839 on Build GRCh38
rs541991580 Chr.19: 19201853-19201853 on Build GRCh38
rs775833369 Chr.19: 19201955-19201955 on Build GRCh38
rs757949180 Chr.19: 19201995-19201995 on Build GRCh38
rs764895981 Chr.19: 19201937-19201937 on Build GRCh38
rs763080458 Chr.19: 19201881-19201881 on Build GRCh38
rs376935004 Chr.19: 19201759-19201759 on Build GRCh38
rs746304124 Chr.19: 19202016-19202016 on Build GRCh38
rs778694926 Chr.19: 19201913-19201913 on Build GRCh38
rs761097826 Chr.19: 19201690-19201690 on Build GRCh38
rs372322966 Chr.19: 19201761-19201761 on Build GRCh38
rs753884049 Chr.19: 19201970-19201970 on Build GRCh38
rs766369394 Chr.19: 19201902-19201902 on Build GRCh38
rs761991140 Chr.19: 19201968-19201968 on Build GRCh38
rs184084971 Chr.19: 19202047-19202047 on Build GRCh38
rs774252780 Chr.19: 19201963-19201963 on Build GRCh38
rs752747766 Chr.19: 19201704-19201704 on Build GRCh38
rs771314602 Chr.19: 19201930-19201930 on Build GRCh38
rs200263189 Chr.19: 19201732-19201732 on Build GRCh38
rs371480701 Chr.19: 19201982-19201982 on Build GRCh38
rs761785108 Chr.19: 19202046-19202046 on Build GRCh38
rs375809386 Chr.19: 19202044-19202044 on Build GRCh38
rs377064458 Chr.19: 19201681-19201681 on Build GRCh38
rs771493023 Chr.19: 19201740-19201740 on Build GRCh38
rs535513828 Chr.19: 19202130-19202130 on Build GRCh38
rs749954713 Chr.19: 19202089-19202089 on Build GRCh38
rs777522504 Chr.19: 19201997-19201997 on Build GRCh38
rs770214426 Chr.19: 19201954-19201954 on Build GRCh38
rs1006396261 Chr.19: 19201836-19201836 on Build GRCh38
rs113406972 Chr.19: 19201701-19201701 on Build GRCh38
rs1802498 Chr.19: 19201687-19201687 on Build GRCh38
rs543674488 Chr.19: 19202161-19202161 on Build GRCh38
rs751670866 Chr.19: 19201890-19201890 on Build GRCh38
rs765477759 Chr.19: 19201703-19201703 on Build GRCh38
rs751014086 Chr.19: 19201998-19201998 on Build GRCh38
rs759254274 Chr.19: 19201880-19201880 on Build GRCh38
rs984329680 Chr.19: 19202153-19202153 on Build GRCh38
rs781664408 Chr.19: 19202014-19202014 on Build GRCh38
rs148661146 Chr.19: 19202048-19202048 on Build GRCh38
rs377318066 Chr.19: 19201987-19201987 on Build GRCh38
rs749691196 Chr.19: 19202037-19202037 on Build GRCh38
rs753652611 Chr.19: 19201909-19201909 on Build GRCh38
rs888025153 Chr.19: 19201790-19201790 on Build GRCh38
rs768942949 Chr.19: 19201959-19201959 on Build GRCh38
rs112578985 Chr.19: 19202068-19202068 on Build GRCh38
rs762857778 Chr.19: 19202054-19202054 on Build GRCh38
rs764009534 Chr.19: 19201889-19201889 on Build GRCh38
rs925500111 Chr.19: 19201868-19201868 on Build GRCh38
rs73922832 Chr.19: 19201728-19201728 on Build GRCh38
rs371438200 Chr.19: 19201724-19201724 on Build GRCh38
rs774754199 Chr.19: 19201923-19201924 on Build GRCh38
rs749806448 Chr.19: 19201739-19201742 on Build GRCh38
rs888337184 Chr.19: 19202032-19202032 on Build GRCh38
rs10413280 Chr.19: 19201731-19201731 on Build GRCh38
rs1050467 Chr.19: 19201807-19201807 on Build GRCh38
rs756854678 Chr.19: 19202010-19202010 on Build GRCh38
rs373829001 Chr.19: 19201771-19201771 on Build GRCh38
rs1040844756 Chr.19: 19201713-19201713 on Build GRCh38
rs1013963321 Chr.19: 19201766-19201766 on Build GRCh38
rs766587567 Chr.19: 19201698-19201698 on Build GRCh38
rs199668878 Chr.19: 19202081-19202081 on Build GRCh38
rs754869961 Chr.19: 19201910-19201910 on Build GRCh38
rs761438820 Chr.19: 19201683-19201683 on Build GRCh38
rs372081950 Chr.19: 19201758-19201758 on Build GRCh38
rs777174194 Chr.19: 19201947-19201947 on Build GRCh38
rs577525380 Chr.19: 19201725-19201725 on Build GRCh38
rs370507391 Chr.19: 19201735-19201735 on Build GRCh38
rs199868077 Chr.19: 19201711-19201711 on Build GRCh38
rs747737597 Chr.19: 19201914-19201914 on Build GRCh38
rs930089492 Chr.19: 19202147-19202147 on Build GRCh38
rs571110961 Chr.19: 19202055-19202055 on Build GRCh38
rs746222485 Chr.19: 19201948-19201948 on Build GRCh38
rs965400041 Chr.19: 19202113-19202113 on Build GRCh38
rs756570933 Chr.19: 19202090-19202090 on Build GRCh38
rs769324444 Chr.19: 19201749-19201750 on Build GRCh38
rs967307869 Chr.19: 19201817-19201817 on Build GRCh38
rs746561492 Chr.19: 19201928-19201928 on Build GRCh38
rs976845763 Chr.19: 19202126-19202126 on Build GRCh38
rs770294319 Chr.19: 19202025-19202025 on Build GRCh38
rs752403849 Chr.19: 19202052-19202052 on Build GRCh38
rs796152075 Chr.19: 19202009-19202009 on Build GRCh38
rs985424456 Chr.19: 19201903-19201903 on Build GRCh38
rs1018279266 Chr.19: 19202107-19202107 on Build GRCh38
rs1048243561 Chr.19: 19202031-19202031 on Build GRCh38
rs780345865 Chr.19: 19202029-19202029 on Build GRCh38
rs958236971 Chr.19: 19202098-19202098 on Build GRCh38
rs879174733 Chr.19: 19201894-19201894 on Build GRCh38
rs755954287 Chr.19: 19201895-19201895 on Build GRCh38
rs759707164 Chr.19: 19201702-19201702 on Build GRCh38
rs144070913 Chr.19: 19201916-19201916 on Build GRCh38
rs756078268 Chr.19: 19201715-19201715 on Build GRCh38
rs749592649 Chr.19: 19202092-19202092 on Build GRCh38
rs1048062774 Chr.19: 19201827-19201827 on Build GRCh38

View Full Product Details