Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19793506-19794010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF506
Entrez Gene ID
440515
Gene Name
zinc finger protein 506
Gene Aliases
-
UniGene
Hs.351906
Gene Chromosome Location
Chr.19: 19792711-19821751 on Build GRCh38
Transcripts
NM_001099269.2 NM_001145404.1

SNP Detail

SNP ID Chromosome Location
rs763186164 Chr.19: 19793607-19793607 on Build GRCh38
rs151158157 Chr.19: 19793878-19793878 on Build GRCh38
rs564344368 Chr.19: 19793551-19793551 on Build GRCh38
rs370302196 Chr.19: 19793810-19793810 on Build GRCh38
rs1056015353 Chr.19: 19793581-19793581 on Build GRCh38
rs945673106 Chr.19: 19793549-19793549 on Build GRCh38
rs764795944 Chr.19: 19793929-19793929 on Build GRCh38
rs768122848 Chr.19: 19793863-19793863 on Build GRCh38
rs943584849 Chr.19: 19793639-19793639 on Build GRCh38
rs773197516 Chr.19: 19793556-19793556 on Build GRCh38
rs952394134 Chr.19: 19793700-19793700 on Build GRCh38
rs966171870 Chr.19: 19793725-19793725 on Build GRCh38
rs528917990 Chr.19: 19793716-19793716 on Build GRCh38
rs1049417746 Chr.19: 19793621-19793621 on Build GRCh38
rs976268969 Chr.19: 19793726-19793726 on Build GRCh38
rs910847219 Chr.19: 19793629-19793629 on Build GRCh38
rs56318245 Chr.19: 19793865-19793865 on Build GRCh38
rs897905357 Chr.19: 19793985-19793985 on Build GRCh38
rs935969370 Chr.19: 19793576-19793576 on Build GRCh38
rs7246983 Chr.19: 19793701-19793701 on Build GRCh38
rs761372668 Chr.19: 19793926-19793926 on Build GRCh38
rs1041388083 Chr.19: 19793555-19793555 on Build GRCh38
rs749918720 Chr.19: 19793930-19793930 on Build GRCh38
rs187636719 Chr.19: 19793686-19793686 on Build GRCh38
rs925663838 Chr.19: 19793804-19793804 on Build GRCh38
rs1014053399 Chr.19: 19793633-19793633 on Build GRCh38
rs539960884 Chr.19: 19793691-19793691 on Build GRCh38
rs142166700 Chr.19: 19793841-19793841 on Build GRCh38
rs113926510 Chr.19: 19793816-19793816 on Build GRCh38
rs1040592016 Chr.19: 19793657-19793657 on Build GRCh38
rs1045549541 Chr.19: 19793956-19793956 on Build GRCh38
rs977485593 Chr.19: 19793798-19793798 on Build GRCh38
rs993872370 Chr.19: 19793688-19793688 on Build GRCh38
rs924986490 Chr.19: 19793548-19793548 on Build GRCh38
rs770357836 Chr.19: 19793757-19793760 on Build GRCh38
rs774709290 Chr.19: 19793762-19793762 on Build GRCh38

View Full Product Details