Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 22634152-22634642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF492
Entrez Gene ID
57615
Gene Name
zinc finger protein 492
Gene Aliases
ZNF115
UniGene
Hs.232108
Gene Chromosome Location
Chr.19: 22634324-22667670 on Build GRCh38
Transcripts
NM_020855.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
86255 c.? p.? Chr.19: 22634476-22634476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574290464 Chr.19: 22634561-22634561 on Build GRCh38
rs905739280 Chr.19: 22634204-22634204 on Build GRCh38
rs565487949 Chr.19: 22634434-22634434 on Build GRCh38
rs1001855594 Chr.19: 22634213-22634213 on Build GRCh38
rs1015664710 Chr.19: 22634277-22634277 on Build GRCh38
rs998676280 Chr.19: 22634580-22634580 on Build GRCh38
rs1005663341 Chr.19: 22634258-22634258 on Build GRCh38
rs77432778 Chr.19: 22634221-22634221 on Build GRCh38
rs1029699441 Chr.19: 22634589-22634589 on Build GRCh38
rs912948844 Chr.19: 22634404-22634404 on Build GRCh38
rs891478256 Chr.19: 22634189-22634189 on Build GRCh38
rs562039976 Chr.19: 22634364-22634364 on Build GRCh38
rs78396732 Chr.19: 22634215-22634215 on Build GRCh38
rs971795326 Chr.19: 22634609-22634609 on Build GRCh38
rs1007483568 Chr.19: 22634524-22634524 on Build GRCh38
rs539589307 Chr.19: 22634454-22634454 on Build GRCh38
rs887628693 Chr.19: 22634564-22634564 on Build GRCh38
rs563376676 Chr.19: 22634216-22634216 on Build GRCh38
rs374683733 Chr.19: 22634473-22634473 on Build GRCh38
rs370410008 Chr.19: 22634476-22634476 on Build GRCh38
rs148713444 Chr.19: 22634475-22634476 on Build GRCh38
rs369506884 Chr.19: 22634341-22634341 on Build GRCh38
rs913911394 Chr.19: 22634210-22634210 on Build GRCh38
rs574179391 Chr.19: 22634249-22634249 on Build GRCh38
rs888943414 Chr.19: 22634305-22634305 on Build GRCh38
rs968042242 Chr.19: 22634407-22634407 on Build GRCh38
rs926846907 Chr.19: 22634451-22634451 on Build GRCh38
rs935788563 Chr.19: 22634187-22634187 on Build GRCh38
rs564111445 Chr.19: 22634318-22634318 on Build GRCh38
rs555447534 Chr.19: 22634499-22634499 on Build GRCh38
rs374454452 Chr.19: 22634578-22634578 on Build GRCh38
rs77957147 Chr.19: 22634315-22634315 on Build GRCh38
rs930325307 Chr.19: 22634485-22634485 on Build GRCh38
rs866666503 Chr.19: 22634263-22634263 on Build GRCh38
rs1027522643 Chr.19: 22634216-22634216 on Build GRCh38
rs551448919 Chr.19: 22634474-22634474 on Build GRCh38
rs796135727 Chr.19: 22634476-22634476 on Build GRCh38
rs3060761 Chr.19: 22634229-22634230 on Build GRCh38
rs537382407 Chr.19: 22634226-22634226 on Build GRCh38
rs377740459 Chr.19: 22634281-22634281 on Build GRCh38
rs986111578 Chr.19: 22634333-22634333 on Build GRCh38
rs534965519 Chr.19: 22634591-22634591 on Build GRCh38
rs1004690263 Chr.19: 22634572-22634572 on Build GRCh38
rs1048773296 Chr.19: 22634494-22634494 on Build GRCh38
rs376564253 Chr.19: 22634476-22634477 on Build GRCh38
rs547582278 Chr.19: 22634433-22634433 on Build GRCh38
rs901715103 Chr.19: 22634554-22634554 on Build GRCh38
rs961591617 Chr.19: 22634606-22634606 on Build GRCh38
rs1052814531 Chr.19: 22634398-22634398 on Build GRCh38
rs796721383 Chr.19: 22634228-22634229 on Build GRCh38
rs1021022551 Chr.19: 22634567-22634567 on Build GRCh38
rs868702541 Chr.19: 22634280-22634280 on Build GRCh38
rs370383978 Chr.19: 22634321-22634321 on Build GRCh38
rs951845864 Chr.19: 22634252-22634252 on Build GRCh38
rs201523211 Chr.19: 22634325-22634325 on Build GRCh38
rs553650119 Chr.19: 22634593-22634593 on Build GRCh38
rs986214392 Chr.19: 22634177-22634177 on Build GRCh38
rs529144819 Chr.19: 22634373-22634373 on Build GRCh38
rs1027146939 Chr.19: 22634504-22634504 on Build GRCh38
rs1013892060 Chr.19: 22634198-22634198 on Build GRCh38
rs895533528 Chr.19: 22634227-22634227 on Build GRCh38
rs58851429 Chr.19: 22634228-22634228 on Build GRCh38
rs936146880 Chr.19: 22634378-22634378 on Build GRCh38
rs201110647 Chr.19: 22634220-22634221 on Build GRCh38
rs569748392 Chr.19: 22634481-22634481 on Build GRCh38
rs61006338 Chr.19: 22634293-22634293 on Build GRCh38
rs1040601344 Chr.19: 22634542-22634542 on Build GRCh38
rs527939735 Chr.19: 22634605-22634605 on Build GRCh38
rs879133174 Chr.19: 22634491-22634491 on Build GRCh38
rs557730809 Chr.19: 22634202-22634202 on Build GRCh38
rs1033690179 Chr.19: 22634586-22634586 on Build GRCh38
rs1048990782 Chr.19: 22634544-22634544 on Build GRCh38
rs56049933 Chr.19: 22634314-22634314 on Build GRCh38
rs374239183 Chr.19: 22634385-22634385 on Build GRCh38
rs539509767 Chr.19: 22634190-22634190 on Build GRCh38
rs951647370 Chr.19: 22634596-22634596 on Build GRCh38
rs946961601 Chr.19: 22634214-22634214 on Build GRCh38
rs537430786 Chr.19: 22634487-22634487 on Build GRCh38
rs1052782387 Chr.19: 22634188-22634188 on Build GRCh38
rs779244351 Chr.19: 22634474-22634475 on Build GRCh38
rs386807978 Chr.19: 22634473-22634476 on Build GRCh38
rs879058088 Chr.19: 22634347-22634347 on Build GRCh38
rs1015695911 Chr.19: 22634576-22634576 on Build GRCh38
rs559655956 Chr.19: 22634295-22634295 on Build GRCh38
rs1020908304 Chr.19: 22634392-22634392 on Build GRCh38
rs1007776620 Chr.19: 22634312-22634312 on Build GRCh38
rs148291606 Chr.19: 22634473-22634474 on Build GRCh38
rs926773091 Chr.19: 22634216-22634217 on Build GRCh38

View Full Product Details