Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 23358645-23359180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF91
Entrez Gene ID
7644
Gene Name
zinc finger protein 91
Gene Aliases
HPF7, HTF10
UniGene
Hs.531632
Gene Chromosome Location
Chr.19: 23305004-23395560 on Build GRCh38
Transcripts
NM_001300951.1 NM_003430.3 XM_006722880.3 XM_011528265.2 XM_011528266.2 XM_017027235.1 XM_017027236.1

SNP Detail

SNP ID Chromosome Location
rs558817294 Chr.19: 23358734-23358734 on Build GRCh38
rs925118221 Chr.19: 23359105-23359105 on Build GRCh38
rs1050671860 Chr.19: 23358846-23358846 on Build GRCh38
rs3745110 Chr.19: 23358869-23358869 on Build GRCh38
rs558906615 Chr.19: 23359051-23359051 on Build GRCh38
rs543301935 Chr.19: 23358984-23358985 on Build GRCh38
rs544849374 Chr.19: 23358771-23358771 on Build GRCh38
rs985451024 Chr.19: 23358764-23358764 on Build GRCh38
rs544757982 Chr.19: 23358682-23358682 on Build GRCh38
rs1013828607 Chr.19: 23358898-23358898 on Build GRCh38
rs1027169995 Chr.19: 23358981-23358981 on Build GRCh38
rs533234475 Chr.19: 23359146-23359146 on Build GRCh38
rs576915509 Chr.19: 23358742-23358742 on Build GRCh38
rs972809031 Chr.19: 23358724-23358724 on Build GRCh38
rs986018248 Chr.19: 23359066-23359066 on Build GRCh38
rs984849062 Chr.19: 23358801-23358801 on Build GRCh38
rs570354845 Chr.19: 23359034-23359034 on Build GRCh38
rs972095289 Chr.19: 23358819-23358819 on Build GRCh38
rs923384988 Chr.19: 23359036-23359036 on Build GRCh38
rs1024706141 Chr.19: 23358905-23358905 on Build GRCh38
rs940789230 Chr.19: 23358810-23358810 on Build GRCh38
rs542519554 Chr.19: 23358818-23358818 on Build GRCh38
rs554604466 Chr.19: 23359147-23359147 on Build GRCh38
rs945307268 Chr.19: 23359086-23359086 on Build GRCh38
rs191208041 Chr.19: 23358882-23358882 on Build GRCh38
rs570613243 Chr.19: 23359082-23359082 on Build GRCh38
rs1034928791 Chr.19: 23358924-23358924 on Build GRCh38
rs295368 Chr.19: 23358670-23358670 on Build GRCh38
rs563093507 Chr.19: 23358777-23358777 on Build GRCh38
rs184008634 Chr.19: 23358995-23358995 on Build GRCh38
rs543826846 Chr.19: 23358820-23358820 on Build GRCh38
rs534175239 Chr.19: 23358725-23358725 on Build GRCh38
rs923128143 Chr.19: 23358841-23358841 on Build GRCh38
rs920030614 Chr.19: 23358733-23358733 on Build GRCh38
rs1016234901 Chr.19: 23359016-23359016 on Build GRCh38
rs901679644 Chr.19: 23358829-23358829 on Build GRCh38
rs1025106964 Chr.19: 23358715-23358715 on Build GRCh38
rs936397837 Chr.19: 23359136-23359136 on Build GRCh38
rs373754459 Chr.19: 23358689-23358689 on Build GRCh38
rs1053451739 Chr.19: 23358870-23358870 on Build GRCh38
rs542787413 Chr.19: 23359132-23359132 on Build GRCh38
rs185902520 Chr.19: 23358871-23358871 on Build GRCh38
rs994366092 Chr.19: 23358973-23358973 on Build GRCh38
rs530445093 Chr.19: 23358998-23358998 on Build GRCh38
rs575182677 Chr.19: 23358786-23358786 on Build GRCh38
rs961518231 Chr.19: 23358960-23358960 on Build GRCh38
rs763563169 Chr.19: 23358808-23358808 on Build GRCh38
rs894808136 Chr.19: 23359152-23359152 on Build GRCh38
rs945251913 Chr.19: 23358843-23358843 on Build GRCh38
rs145988188 Chr.19: 23358842-23358842 on Build GRCh38
rs759732126 Chr.19: 23359002-23359002 on Build GRCh38
rs6511433 Chr.19: 23358866-23358866 on Build GRCh38
rs971525734 Chr.19: 23358704-23358704 on Build GRCh38
rs982000365 Chr.19: 23358718-23358718 on Build GRCh38
rs959205566 Chr.19: 23358758-23358758 on Build GRCh38
rs1040703539 Chr.19: 23358827-23358827 on Build GRCh38
rs958990760 Chr.19: 23358978-23358978 on Build GRCh38
rs537905687 Chr.19: 23359102-23359102 on Build GRCh38
rs556749990 Chr.19: 23359115-23359115 on Build GRCh38
rs567137385 Chr.19: 23359026-23359026 on Build GRCh38
rs3745111 Chr.19: 23358683-23358683 on Build GRCh38
rs296094 Chr.19: 23358921-23358921 on Build GRCh38
rs763287990 Chr.19: 23358679-23358679 on Build GRCh38
rs575071114 Chr.19: 23359128-23359128 on Build GRCh38
rs961996055 Chr.19: 23359023-23359023 on Build GRCh38
rs752984386 Chr.19: 23359106-23359106 on Build GRCh38
rs548628925 Chr.19: 23359024-23359024 on Build GRCh38
rs906262806 Chr.19: 23358913-23358913 on Build GRCh38
rs767931822 Chr.19: 23358883-23358883 on Build GRCh38
rs533978001 Chr.19: 23359037-23359037 on Build GRCh38
rs774700760 Chr.19: 23358765-23358765 on Build GRCh38

View Full Product Details