Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40073984-40074500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF780A
Entrez Gene ID
284323
Gene Name
zinc finger protein 780A
Gene Aliases
ZNF780
UniGene
Hs.115140
Gene Chromosome Location
Chr.19: 40069152-40091706 on Build GRCh38
Transcripts
NM_001010880.2 NM_001142577.1 NM_001142578.1 NM_001142579.1 XM_005258773.2 XM_006723150.3 XM_011526771.2 XM_011526772.2 XM_017026611.1 XM_017026612.1 XM_017026613.1 XM_017026614.1 XM_017026615.1 XM_017026616.1 XM_017026617.1 XM_017026618.1 XM_017026619.1

SNP Detail

SNP ID Chromosome Location
rs141846160 Chr.19: 40074195-40074195 on Build GRCh38
rs909272702 Chr.19: 40074140-40074140 on Build GRCh38
rs1009689268 Chr.19: 40074141-40074141 on Build GRCh38
rs752335520 Chr.19: 40074391-40074391 on Build GRCh38
rs549698362 Chr.19: 40074278-40074278 on Build GRCh38
rs960969258 Chr.19: 40074026-40074026 on Build GRCh38
rs967799974 Chr.19: 40074330-40074330 on Build GRCh38
rs186542321 Chr.19: 40074032-40074032 on Build GRCh38
rs868598287 Chr.19: 40074407-40074407 on Build GRCh38
rs948072660 Chr.19: 40074048-40074048 on Build GRCh38
rs948392947 Chr.19: 40074020-40074020 on Build GRCh38
rs530286563 Chr.19: 40074405-40074405 on Build GRCh38
rs892543187 Chr.19: 40074132-40074132 on Build GRCh38
rs905463239 Chr.19: 40074057-40074057 on Build GRCh38
rs368846514 Chr.19: 40074272-40074273 on Build GRCh38
rs796075023 Chr.19: 40074033-40074033 on Build GRCh38
rs181631142 Chr.19: 40074016-40074016 on Build GRCh38
rs1000973881 Chr.19: 40074072-40074072 on Build GRCh38
rs537171872 Chr.19: 40074468-40074468 on Build GRCh38
rs73040569 Chr.19: 40074329-40074329 on Build GRCh38
rs992438447 Chr.19: 40074027-40074027 on Build GRCh38
rs527297826 Chr.19: 40074152-40074152 on Build GRCh38
rs992548682 Chr.19: 40074404-40074404 on Build GRCh38
rs754468316 Chr.19: 40074304-40074304 on Build GRCh38
rs750276787 Chr.19: 40074073-40074074 on Build GRCh38
rs940792077 Chr.19: 40074194-40074194 on Build GRCh38
rs368794924 Chr.19: 40074268-40074268 on Build GRCh38
rs533244447 Chr.19: 40074453-40074453 on Build GRCh38
rs758122436 Chr.19: 40074392-40074392 on Build GRCh38
rs1044069340 Chr.19: 40074462-40074462 on Build GRCh38
rs146284802 Chr.19: 40074209-40074209 on Build GRCh38
rs998050999 Chr.19: 40074253-40074253 on Build GRCh38
rs919124866 Chr.19: 40074340-40074340 on Build GRCh38
rs191350184 Chr.19: 40074254-40074254 on Build GRCh38
rs954695019 Chr.19: 40074199-40074199 on Build GRCh38
rs948384988 Chr.19: 40074454-40074454 on Build GRCh38
rs1053903428 Chr.19: 40074094-40074094 on Build GRCh38

View Full Product Details