Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40742354-40742851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf54
Entrez Gene ID
284325
Gene Name
chromosome 19 open reading frame 54
Gene Aliases
-
UniGene
Hs.585105
Gene Chromosome Location
Chr.19: 40740856-40750713 on Build GRCh38
Transcripts
NM_198476.3 XM_005258776.4 XM_005258778.4 XM_006723152.2 XM_006723153.2 XM_011526774.2 XM_011526775.1 XM_011526776.2 XM_011526777.2 XM_011526778.2 XM_011526779.2 XM_011526782.2 XM_011526783.2 XM_011526784.2 XM_011526785.2 XM_011526786.2 XM_017026620.1 XM_017026621.1 XM_017026622.1 XM_017026623.1 XM_017026624.1 XM_017026625.1 XM_017026626.1 XM_017026627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1134087 c.763G>A p.E255K Chr.19: 40742726-40742726 on Build GRCh38
7329284 c.930G>A p.L310L Chr.19: 40742559-40742559 on Build GRCh38
1148189 c.910C>T p.Q304* Chr.19: 40742579-40742579 on Build GRCh38
5452065 c.833A>G p.E278G Chr.19: 40742656-40742656 on Build GRCh38
5504548 c.901G>T p.D301Y Chr.19: 40742588-40742588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747017273 Chr.19: 40742782-40742782 on Build GRCh38
rs775590457 Chr.19: 40742577-40742577 on Build GRCh38
rs375338329 Chr.19: 40742493-40742493 on Build GRCh38
rs149384660 Chr.19: 40742644-40742644 on Build GRCh38
rs896071154 Chr.19: 40742466-40742466 on Build GRCh38
rs750946358 Chr.19: 40742520-40742520 on Build GRCh38
rs202130851 Chr.19: 40742405-40742405 on Build GRCh38
rs776416834 Chr.19: 40742651-40742653 on Build GRCh38
rs147072628 Chr.19: 40742422-40742422 on Build GRCh38
rs773664679 Chr.19: 40742710-40742710 on Build GRCh38
rs199637969 Chr.19: 40742541-40742541 on Build GRCh38
rs139556174 Chr.19: 40742678-40742678 on Build GRCh38
rs759330829 Chr.19: 40742494-40742494 on Build GRCh38
rs745634354 Chr.19: 40742675-40742675 on Build GRCh38
rs768785932 Chr.19: 40742685-40742685 on Build GRCh38
rs555576611 Chr.19: 40742423-40742423 on Build GRCh38
rs1029461754 Chr.19: 40742760-40742762 on Build GRCh38
rs748144436 Chr.19: 40742798-40742798 on Build GRCh38
rs190187490 Chr.19: 40742550-40742550 on Build GRCh38
rs549826596 Chr.19: 40742573-40742573 on Build GRCh38
rs201256321 Chr.19: 40742445-40742445 on Build GRCh38
rs760939718 Chr.19: 40742510-40742510 on Build GRCh38
rs34255454 Chr.19: 40742768-40742768 on Build GRCh38
rs776571337 Chr.19: 40742687-40742687 on Build GRCh38
rs759689710 Chr.19: 40742400-40742400 on Build GRCh38
rs754212936 Chr.19: 40742428-40742428 on Build GRCh38
rs953909412 Chr.19: 40742523-40742523 on Build GRCh38
rs924591701 Chr.19: 40742780-40742780 on Build GRCh38
rs769056847 Chr.19: 40742779-40742779 on Build GRCh38
rs768049311 Chr.19: 40742633-40742636 on Build GRCh38
rs746312792 Chr.19: 40742450-40742450 on Build GRCh38
rs765701310 Chr.19: 40742614-40742614 on Build GRCh38
rs116144364 Chr.19: 40742378-40742378 on Build GRCh38
rs944252059 Chr.19: 40742459-40742459 on Build GRCh38
rs758203416 Chr.19: 40742766-40742766 on Build GRCh38
rs199734797 Chr.19: 40742522-40742522 on Build GRCh38
rs574071969 Chr.19: 40742788-40742788 on Build GRCh38
rs766060214 Chr.19: 40742392-40742392 on Build GRCh38
rs745867235 Chr.19: 40742568-40742568 on Build GRCh38
rs372869249 Chr.19: 40742511-40742511 on Build GRCh38
rs766800802 Chr.19: 40742711-40742711 on Build GRCh38
rs921532588 Chr.19: 40742817-40742817 on Build GRCh38
rs200386941 Chr.19: 40742489-40742489 on Build GRCh38
rs778465187 Chr.19: 40742752-40742752 on Build GRCh38
rs201880720 Chr.19: 40742572-40742572 on Build GRCh38
rs779758935 Chr.19: 40742773-40742773 on Build GRCh38
rs948937137 Chr.19: 40742654-40742654 on Build GRCh38
rs775537114 Chr.19: 40742502-40742502 on Build GRCh38
rs780783995 Chr.19: 40742462-40742462 on Build GRCh38
rs1006784172 Chr.19: 40742791-40742791 on Build GRCh38
rs376109925 Chr.19: 40742488-40742488 on Build GRCh38
rs560353156 Chr.19: 40742525-40742525 on Build GRCh38
rs752927658 Chr.19: 40742408-40742408 on Build GRCh38
rs748138876 Chr.19: 40742508-40742508 on Build GRCh38
rs914664741 Chr.19: 40742386-40742386 on Build GRCh38
rs770588958 Chr.19: 40742490-40742490 on Build GRCh38
rs763601736 Chr.19: 40742626-40742626 on Build GRCh38
rs554015852 Chr.19: 40742379-40742379 on Build GRCh38
rs538945746 Chr.19: 40742637-40742637 on Build GRCh38
rs760778499 Chr.19: 40742582-40742582 on Build GRCh38
rs750032725 Chr.19: 40742753-40742753 on Build GRCh38
rs144724170 Chr.19: 40742646-40742646 on Build GRCh38
rs768674765 Chr.19: 40742583-40742583 on Build GRCh38
rs756852073 Chr.19: 40742652-40742652 on Build GRCh38
rs769555649 Chr.19: 40742487-40742487 on Build GRCh38
rs774334515 Chr.19: 40742399-40742399 on Build GRCh38
rs760049738 Chr.19: 40742735-40742735 on Build GRCh38
rs375465491 Chr.19: 40742731-40742731 on Build GRCh38
rs769878045 Chr.19: 40742800-40742800 on Build GRCh38
rs749565924 Chr.19: 40742736-40742736 on Build GRCh38
rs778502074 Chr.19: 40742562-40742562 on Build GRCh38
rs761876850 Chr.19: 40742603-40742603 on Build GRCh38
rs747299663 Chr.19: 40742797-40742797 on Build GRCh38
rs36071297 Chr.19: 40742627-40742627 on Build GRCh38
rs187557989 Chr.19: 40742501-40742501 on Build GRCh38
rs754551303 Chr.19: 40742457-40742457 on Build GRCh38
rs749460046 Chr.19: 40742816-40742816 on Build GRCh38
rs756977618 Chr.19: 40742739-40742739 on Build GRCh38
rs752071957 Chr.19: 40742635-40742635 on Build GRCh38
rs144199366 Chr.19: 40742709-40742709 on Build GRCh38
rs374032939 Chr.19: 40742478-40742478 on Build GRCh38
rs947413886 Chr.19: 40742413-40742413 on Build GRCh38
rs759157818 Chr.19: 40742654-40742654 on Build GRCh38
rs201574115 Chr.19: 40742683-40742683 on Build GRCh38
rs200543922 Chr.19: 40742519-40742519 on Build GRCh38
rs192098956 Chr.19: 40742533-40742533 on Build GRCh38
rs551943526 Chr.19: 40742784-40742784 on Build GRCh38
rs757663508 Chr.19: 40742518-40742518 on Build GRCh38
rs781379486 Chr.19: 40742785-40742785 on Build GRCh38
rs565913964 Chr.19: 40742682-40742682 on Build GRCh38
rs774806806 Chr.19: 40742801-40742801 on Build GRCh38
rs184492176 Chr.19: 40742534-40742534 on Build GRCh38
rs773590318 Chr.19: 40742805-40742805 on Build GRCh38
rs750738397 Chr.19: 40742618-40742618 on Build GRCh38
rs778821125 Chr.19: 40742668-40742668 on Build GRCh38
rs776926282 Chr.19: 40742593-40742593 on Build GRCh38
rs762249443 Chr.19: 40742581-40742581 on Build GRCh38

View Full Product Details