Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41275772-41276316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357139 c.689G>A p.S230N Chr.19: 41276201-41276201 on Build GRCh38
3534377 c.569C>T p.P190L Chr.19: 41276210-41276210 on Build GRCh38
6244034 c.764G>T p.G255V Chr.19: 41276276-41276276 on Build GRCh38
7202180 c.786G>A p.K262K Chr.19: 41276298-41276298 on Build GRCh38
7112937 c.575C>G p.T192R Chr.19: 41276216-41276216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753693640 Chr.19: 41276211-41276211 on Build GRCh38
rs751074290 Chr.19: 41276139-41276139 on Build GRCh38
rs914269655 Chr.19: 41276043-41276043 on Build GRCh38
rs889679048 Chr.19: 41275975-41275975 on Build GRCh38
rs144718578 Chr.19: 41276289-41276289 on Build GRCh38
rs918224549 Chr.19: 41275928-41275928 on Build GRCh38
rs774986953 Chr.19: 41276122-41276122 on Build GRCh38
rs1024497371 Chr.19: 41275814-41275814 on Build GRCh38
rs777092056 Chr.19: 41276136-41276136 on Build GRCh38
rs113431619 Chr.19: 41275836-41275836 on Build GRCh38
rs140064732 Chr.19: 41276166-41276166 on Build GRCh38
rs559177664 Chr.19: 41276019-41276019 on Build GRCh38
rs899665621 Chr.19: 41276102-41276102 on Build GRCh38
rs980253258 Chr.19: 41275858-41275858 on Build GRCh38
rs1014653615 Chr.19: 41276093-41276093 on Build GRCh38
rs1015865116 Chr.19: 41275974-41275974 on Build GRCh38
rs943924979 Chr.19: 41276085-41276085 on Build GRCh38
rs538638125 Chr.19: 41276128-41276128 on Build GRCh38
rs777410605 Chr.19: 41276203-41276203 on Build GRCh38
rs747445224 Chr.19: 41276270-41276270 on Build GRCh38
rs751419841 Chr.19: 41275869-41275869 on Build GRCh38
rs903062436 Chr.19: 41275829-41275829 on Build GRCh38
rs773856073 Chr.19: 41275985-41275985 on Build GRCh38
rs1040925136 Chr.19: 41276098-41276098 on Build GRCh38
rs111925003 Chr.19: 41275940-41275941 on Build GRCh38
rs867875500 Chr.19: 41275992-41275992 on Build GRCh38
rs779116610 Chr.19: 41276220-41276220 on Build GRCh38
rs549294227 Chr.19: 41276044-41276044 on Build GRCh38
rs769253732 Chr.19: 41276273-41276273 on Build GRCh38
rs149059713 Chr.19: 41276258-41276258 on Build GRCh38
rs781101057 Chr.19: 41276058-41276058 on Build GRCh38
rs543459404 Chr.19: 41275852-41275852 on Build GRCh38
rs1017074113 Chr.19: 41276169-41276169 on Build GRCh38
rs937129179 Chr.19: 41276032-41276032 on Build GRCh38
rs147982601 Chr.19: 41275808-41275808 on Build GRCh38
rs531856213 Chr.19: 41276068-41276068 on Build GRCh38
rs762268558 Chr.19: 41276279-41276279 on Build GRCh38
rs867333943 Chr.19: 41276099-41276099 on Build GRCh38
rs527912083 Chr.19: 41276034-41276034 on Build GRCh38
rs775990709 Chr.19: 41276110-41276110 on Build GRCh38
rs759118022 Chr.19: 41276143-41276143 on Build GRCh38
rs577019475 Chr.19: 41275900-41275900 on Build GRCh38
rs541165696 Chr.19: 41276013-41276013 on Build GRCh38
rs1008084585 Chr.19: 41275840-41275840 on Build GRCh38
rs182124729 Chr.19: 41275958-41275958 on Build GRCh38
rs564849184 Chr.19: 41275892-41275892 on Build GRCh38
rs549766552 Chr.19: 41276095-41276095 on Build GRCh38
rs942563244 Chr.19: 41276000-41276000 on Build GRCh38
rs933913472 Chr.19: 41275932-41275932 on Build GRCh38
rs757384508 Chr.19: 41276218-41276218 on Build GRCh38
rs903117055 Chr.19: 41276071-41276071 on Build GRCh38
rs972479305 Chr.19: 41276226-41276226 on Build GRCh38
rs766076652 Chr.19: 41276137-41276137 on Build GRCh38
rs770641111 Chr.19: 41276281-41276281 on Build GRCh38
rs759317274 Chr.19: 41276293-41276293 on Build GRCh38
rs150301101 Chr.19: 41276180-41276180 on Build GRCh38
rs952735945 Chr.19: 41275893-41275893 on Build GRCh38
rs867953637 Chr.19: 41276285-41276285 on Build GRCh38
rs1041641258 Chr.19: 41275828-41275828 on Build GRCh38
rs772125858 Chr.19: 41276252-41276252 on Build GRCh38
rs926774650 Chr.19: 41276027-41276027 on Build GRCh38
rs144925593 Chr.19: 41276247-41276247 on Build GRCh38
rs772644970 Chr.19: 41275904-41275905 on Build GRCh38
rs10401902 Chr.19: 41276103-41276103 on Build GRCh38
rs756140695 Chr.19: 41276187-41276187 on Build GRCh38
rs745841210 Chr.19: 41276061-41276061 on Build GRCh38
rs1019432033 Chr.19: 41275841-41275841 on Build GRCh38
rs1050943661 Chr.19: 41275940-41275940 on Build GRCh38
rs777506441 Chr.19: 41275868-41275868 on Build GRCh38
rs774004388 Chr.19: 41276057-41276058 on Build GRCh38
rs200747059 Chr.19: 41276122-41276122 on Build GRCh38
rs778927016 Chr.19: 41276152-41276152 on Build GRCh38
rs561641394 Chr.19: 41276057-41276057 on Build GRCh38
rs891773663 Chr.19: 41276092-41276092 on Build GRCh38
rs1048481368 Chr.19: 41276132-41276132 on Build GRCh38

View Full Product Details