Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41859814-41860303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS19
Entrez Gene ID
6223
Gene Name
ribosomal protein S19
Gene Aliases
DBA, DBA1, S19
UniGene
Hs.438429
Gene Chromosome Location
Chr.19: 41859918-41871416 on Build GRCh38
Transcripts
NM_001022.3 NM_001321484.1 NM_001321485.1 XM_017027113.1

SNP Detail

SNP ID Chromosome Location
rs782570955 Chr.19: 41859857-41859857 on Build GRCh38
rs566801304 Chr.19: 41859941-41859941 on Build GRCh38
rs1053550637 Chr.19: 41860100-41860100 on Build GRCh38
rs949340086 Chr.19: 41859840-41859840 on Build GRCh38
rs539033597 Chr.19: 41860125-41860125 on Build GRCh38
rs554238392 Chr.19: 41860184-41860184 on Build GRCh38
rs187442098 Chr.19: 41859894-41859894 on Build GRCh38
rs374817141 Chr.19: 41859842-41859847 on Build GRCh38
rs944451689 Chr.19: 41860286-41860286 on Build GRCh38
rs911560031 Chr.19: 41860216-41860216 on Build GRCh38
rs908421671 Chr.19: 41859837-41859837 on Build GRCh38
rs1042565260 Chr.19: 41860214-41860214 on Build GRCh38
rs886054468 Chr.19: 41860216-41860216 on Build GRCh38
rs538435296 Chr.19: 41860034-41860034 on Build GRCh38
rs981989445 Chr.19: 41860176-41860176 on Build GRCh38
rs1024294746 Chr.19: 41860162-41860162 on Build GRCh38
rs900758092 Chr.19: 41860128-41860128 on Build GRCh38
rs1046190572 Chr.19: 41859841-41859841 on Build GRCh38
rs905057157 Chr.19: 41859869-41859869 on Build GRCh38
rs977953928 Chr.19: 41860279-41860279 on Build GRCh38
rs971454169 Chr.19: 41860249-41860249 on Build GRCh38
rs893457876 Chr.19: 41859895-41859895 on Build GRCh38
rs77406992 Chr.19: 41860016-41860016 on Build GRCh38
rs1009178904 Chr.19: 41859946-41859946 on Build GRCh38
rs547036103 Chr.19: 41859908-41859908 on Build GRCh38
rs926449187 Chr.19: 41860081-41860081 on Build GRCh38
rs542523484 Chr.19: 41860243-41860243 on Build GRCh38
rs937747590 Chr.19: 41859889-41859889 on Build GRCh38
rs890691334 Chr.19: 41859933-41859933 on Build GRCh38
rs564643055 Chr.19: 41859890-41859890 on Build GRCh38
rs915073701 Chr.19: 41860275-41860275 on Build GRCh38
rs148756833 Chr.19: 41859968-41859968 on Build GRCh38
rs937883307 Chr.19: 41860099-41860099 on Build GRCh38
rs782338175 Chr.19: 41860109-41860109 on Build GRCh38
rs989252118 Chr.19: 41860196-41860196 on Build GRCh38
rs371456510 Chr.19: 41860023-41860023 on Build GRCh38
rs192556926 Chr.19: 41860002-41860002 on Build GRCh38
rs1038727599 Chr.19: 41860228-41860228 on Build GRCh38
rs761367207 Chr.19: 41859996-41859996 on Build GRCh38
rs893529938 Chr.19: 41860101-41860101 on Build GRCh38
rs762623833 Chr.19: 41859839-41859839 on Build GRCh38
rs989833722 Chr.19: 41860274-41860274 on Build GRCh38
rs1049210416 Chr.19: 41860238-41860238 on Build GRCh38
rs900726698 Chr.19: 41860229-41860229 on Build GRCh38
rs995105344 Chr.19: 41860236-41860236 on Build GRCh38
rs561259640 Chr.19: 41860257-41860257 on Build GRCh38
rs563430961 Chr.19: 41860121-41860121 on Build GRCh38
rs926500087 Chr.19: 41860181-41860181 on Build GRCh38
rs886054465 Chr.19: 41859951-41859951 on Build GRCh38
rs767047664 Chr.19: 41860006-41860006 on Build GRCh38
rs750930212 Chr.19: 41860130-41860130 on Build GRCh38
rs564634801 Chr.19: 41860280-41860280 on Build GRCh38
rs1021056176 Chr.19: 41859926-41859926 on Build GRCh38
rs959297056 Chr.19: 41860190-41860190 on Build GRCh38
rs142324259 Chr.19: 41860056-41860056 on Build GRCh38
rs983964819 Chr.19: 41860156-41860156 on Build GRCh38
rs543224166 Chr.19: 41860262-41860262 on Build GRCh38
rs578174704 Chr.19: 41860123-41860123 on Build GRCh38
rs549584014 Chr.19: 41859850-41859850 on Build GRCh38
rs576110254 Chr.19: 41860261-41860261 on Build GRCh38
rs368032603 Chr.19: 41859847-41859847 on Build GRCh38
rs782560175 Chr.19: 41859972-41859972 on Build GRCh38
rs151257482 Chr.19: 41860285-41860285 on Build GRCh38
rs945134757 Chr.19: 41860201-41860201 on Build GRCh38
rs1024847820 Chr.19: 41860245-41860245 on Build GRCh38
rs886054467 Chr.19: 41860151-41860151 on Build GRCh38
rs915084037 Chr.19: 41860200-41860200 on Build GRCh38
rs782540676 Chr.19: 41860278-41860278 on Build GRCh38
rs886634767 Chr.19: 41860240-41860240 on Build GRCh38
rs751081173 Chr.19: 41859921-41859921 on Build GRCh38
rs34020014 Chr.19: 41860144-41860145 on Build GRCh38
rs370562318 Chr.19: 41860199-41860199 on Build GRCh38
rs782624335 Chr.19: 41860001-41860001 on Build GRCh38
rs971403329 Chr.19: 41860173-41860173 on Build GRCh38
rs983443283 Chr.19: 41860011-41860011 on Build GRCh38
rs1028250648 Chr.19: 41860138-41860138 on Build GRCh38
rs982022047 Chr.19: 41860077-41860077 on Build GRCh38
rs782323756 Chr.19: 41860142-41860142 on Build GRCh38
rs781965764 Chr.19: 41860127-41860127 on Build GRCh38
rs782271430 Chr.19: 41859887-41859887 on Build GRCh38
rs886054466 Chr.19: 41860139-41860139 on Build GRCh38
rs1017773434 Chr.19: 41859957-41859957 on Build GRCh38
rs973481960 Chr.19: 41859977-41859977 on Build GRCh38
rs950806244 Chr.19: 41860151-41860151 on Build GRCh38
rs1042039194 Chr.19: 41860117-41860117 on Build GRCh38

View Full Product Details