Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43799538-43800035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPD5
Entrez Gene ID
284348
Gene Name
LY6/PLAUR domain containing 5
Gene Aliases
PRO4356
UniGene
Hs.44289
Gene Chromosome Location
Chr.19: 43794835-43827199 on Build GRCh38
Transcripts
NM_001031749.2 NM_001288763.1 NM_182573.2 XM_011526794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3289357 c.171G>T p.E57D Chr.19: 43799728-43799728 on Build GRCh38
3289355 c.193G>A p.G65R Chr.19: 43799706-43799706 on Build GRCh38
997712 c.121C>A p.L41I Chr.19: 43799778-43799778 on Build GRCh38
439725 c.1A>C p.M1L Chr.19: 43799769-43799769 on Build GRCh38
3960182 c.108T>C p.F36F Chr.19: 43799791-43799791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751506948 Chr.19: 43799690-43799690 on Build GRCh38
rs775261237 Chr.19: 43799858-43799858 on Build GRCh38
rs113152404 Chr.19: 43799936-43799936 on Build GRCh38
rs574556844 Chr.19: 43799974-43799974 on Build GRCh38
rs73556877 Chr.19: 43799588-43799588 on Build GRCh38
rs1048039511 Chr.19: 43799880-43799880 on Build GRCh38
rs373388413 Chr.19: 43799706-43799706 on Build GRCh38
rs556310410 Chr.19: 43799914-43799914 on Build GRCh38
rs200873630 Chr.19: 43799820-43799820 on Build GRCh38
rs986597726 Chr.19: 43799771-43799771 on Build GRCh38
rs1024400400 Chr.19: 43799923-43799923 on Build GRCh38
rs768715780 Chr.19: 43799830-43799830 on Build GRCh38
rs779489365 Chr.19: 43799725-43799725 on Build GRCh38
rs895733655 Chr.19: 43799676-43799676 on Build GRCh38
rs760239981 Chr.19: 43799799-43799799 on Build GRCh38
rs763687376 Chr.19: 43799866-43799866 on Build GRCh38
rs748686828 Chr.19: 43799678-43799678 on Build GRCh38
rs943464173 Chr.19: 43799591-43799591 on Build GRCh38
rs752893556 Chr.19: 43799696-43799696 on Build GRCh38
rs766199998 Chr.19: 43799686-43799686 on Build GRCh38
rs763738811 Chr.19: 43799802-43799802 on Build GRCh38
rs773956808 Chr.19: 43799786-43799786 on Build GRCh38
rs753538810 Chr.19: 43799805-43799805 on Build GRCh38
rs1052212452 Chr.19: 43799570-43799570 on Build GRCh38
rs780619337 Chr.19: 43799754-43799754 on Build GRCh38
rs558813718 Chr.19: 43799689-43799689 on Build GRCh38
rs188086193 Chr.19: 43799745-43799745 on Build GRCh38
rs754927813 Chr.19: 43799691-43799691 on Build GRCh38
rs764237516 Chr.19: 43799680-43799680 on Build GRCh38
rs767661783 Chr.19: 43799685-43799685 on Build GRCh38
rs1000626487 Chr.19: 43799630-43799630 on Build GRCh38
rs780400120 Chr.19: 43799826-43799826 on Build GRCh38
rs773513759 Chr.19: 43799849-43799849 on Build GRCh38
rs772405562 Chr.19: 43799753-43799753 on Build GRCh38
rs771950996 Chr.19: 43799857-43799857 on Build GRCh38
rs753344728 Chr.19: 43799867-43799867 on Build GRCh38
rs987702535 Chr.19: 43799800-43799800 on Build GRCh38
rs369824236 Chr.19: 43799697-43799697 on Build GRCh38
rs760762946 Chr.19: 43799862-43799862 on Build GRCh38
rs376766482 Chr.19: 43799815-43799815 on Build GRCh38
rs193146372 Chr.19: 43799755-43799755 on Build GRCh38
rs376561592 Chr.19: 43799660-43799660 on Build GRCh38
rs116523509 Chr.19: 43799759-43799759 on Build GRCh38
rs369809260 Chr.19: 43799672-43799672 on Build GRCh38
rs781304233 Chr.19: 43799835-43799835 on Build GRCh38
rs747587509 Chr.19: 43799829-43799829 on Build GRCh38
rs769935788 Chr.19: 43799848-43799848 on Build GRCh38
rs775493076 Chr.19: 43799671-43799671 on Build GRCh38
rs1033249476 Chr.19: 43799648-43799648 on Build GRCh38
rs1044356549 Chr.19: 43799942-43799942 on Build GRCh38
rs895609268 Chr.19: 43799894-43799894 on Build GRCh38
rs571088457 Chr.19: 43799635-43799635 on Build GRCh38
rs146241885 Chr.19: 43799847-43799847 on Build GRCh38
rs772191892 Chr.19: 43799657-43799657 on Build GRCh38
rs777147473 Chr.19: 43799593-43799593 on Build GRCh38
rs527869545 Chr.19: 43799807-43799807 on Build GRCh38
rs753596171 Chr.19: 43799714-43799714 on Build GRCh38
rs112412953 Chr.19: 43799804-43799804 on Build GRCh38
rs770698693 Chr.19: 43799768-43799768 on Build GRCh38
rs971088418 Chr.19: 43799687-43799687 on Build GRCh38
rs929362353 Chr.19: 43799871-43799871 on Build GRCh38
rs376531706 Chr.19: 43799655-43799655 on Build GRCh38
rs377339530 Chr.19: 43799788-43799788 on Build GRCh38
rs541650037 Chr.19: 43799816-43799816 on Build GRCh38
rs184820958 Chr.19: 43799685-43799685 on Build GRCh38
rs750691245 Chr.19: 43799819-43799819 on Build GRCh38
rs905778073 Chr.19: 43799955-43799955 on Build GRCh38
rs950014182 Chr.19: 43799938-43799938 on Build GRCh38
rs549874734 Chr.19: 43799922-43799922 on Build GRCh38
rs936006997 Chr.19: 43799834-43799834 on Build GRCh38
rs542210890 Chr.19: 43799720-43799720 on Build GRCh38
rs752704339 Chr.19: 43799795-43799795 on Build GRCh38
rs751359952 Chr.19: 43799656-43799657 on Build GRCh38
rs962388847 Chr.19: 43799707-43799707 on Build GRCh38
rs185328564 Chr.19: 43799852-43799852 on Build GRCh38
rs114779742 Chr.19: 43799760-43799760 on Build GRCh38
rs767316236 Chr.19: 43799794-43799794 on Build GRCh38
rs761637505 Chr.19: 43799684-43799685 on Build GRCh38

View Full Product Details