Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45141937-45142441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R37
Entrez Gene ID
284352
Gene Name
protein phosphatase 1 regulatory subunit 37
Gene Aliases
LRRC68
UniGene
Hs.285363
Gene Chromosome Location
Chr.19: 45093173-45147286 on Build GRCh38
Transcripts
NM_019121.1 XM_006723159.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7287186 c.699G>A p.G233G Chr.19: 45142192-45142192 on Build GRCh38
4853111 c.729G>A p.L243L Chr.19: 45142313-45142313 on Build GRCh38
6409717 c.573C>A p.S191R Chr.19: 45142066-45142066 on Build GRCh38
6405265 c.602C>A p.T201K Chr.19: 45142095-45142095 on Build GRCh38
5502114 c.739A>G p.M247V Chr.19: 45142323-45142323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559590448 Chr.19: 45142262-45142262 on Build GRCh38
rs565386331 Chr.19: 45142070-45142070 on Build GRCh38
rs1028406824 Chr.19: 45142179-45142179 on Build GRCh38
rs35904300 Chr.19: 45142177-45142177 on Build GRCh38
rs757346574 Chr.19: 45142062-45142062 on Build GRCh38
rs531551526 Chr.19: 45142379-45142379 on Build GRCh38
rs757543143 Chr.19: 45142341-45142341 on Build GRCh38
rs977174923 Chr.19: 45142038-45142038 on Build GRCh38
rs984722522 Chr.19: 45142243-45142243 on Build GRCh38
rs573794110 Chr.19: 45141964-45141990 on Build GRCh38
rs887088712 Chr.19: 45142130-45142130 on Build GRCh38
rs770557359 Chr.19: 45142189-45142189 on Build GRCh38
rs966107666 Chr.19: 45141968-45141968 on Build GRCh38
rs541184431 Chr.19: 45142123-45142123 on Build GRCh38
rs938722979 Chr.19: 45142089-45142089 on Build GRCh38
rs950022993 Chr.19: 45142288-45142288 on Build GRCh38
rs766123136 Chr.19: 45142095-45142095 on Build GRCh38
rs781192336 Chr.19: 45142378-45142378 on Build GRCh38
rs755923478 Chr.19: 45142413-45142413 on Build GRCh38
rs950604576 Chr.19: 45142000-45142000 on Build GRCh38
rs975707453 Chr.19: 45141990-45141990 on Build GRCh38
rs747665493 Chr.19: 45142257-45142257 on Build GRCh38
rs558677254 Chr.19: 45142113-45142113 on Build GRCh38
rs867751099 Chr.19: 45142131-45142131 on Build GRCh38
rs754082005 Chr.19: 45142323-45142323 on Build GRCh38
rs955616428 Chr.19: 45141969-45141969 on Build GRCh38
rs577814316 Chr.19: 45142294-45142294 on Build GRCh38
rs987040981 Chr.19: 45142194-45142194 on Build GRCh38
rs112401916 Chr.19: 45142295-45142295 on Build GRCh38
rs890003771 Chr.19: 45142305-45142305 on Build GRCh38
rs1019044368 Chr.19: 45142201-45142201 on Build GRCh38
rs947362102 Chr.19: 45142296-45142296 on Build GRCh38
rs1026043767 Chr.19: 45142394-45142394 on Build GRCh38
rs886117261 Chr.19: 45142408-45142408 on Build GRCh38
rs987543312 Chr.19: 45141974-45141974 on Build GRCh38
rs948554994 Chr.19: 45142053-45142053 on Build GRCh38
rs769262506 Chr.19: 45142132-45142132 on Build GRCh38
rs1043710665 Chr.19: 45142297-45142297 on Build GRCh38
rs909100195 Chr.19: 45142261-45142261 on Build GRCh38
rs1041207861 Chr.19: 45142352-45142352 on Build GRCh38
rs979960177 Chr.19: 45142267-45142267 on Build GRCh38
rs376218677 Chr.19: 45141991-45142017 on Build GRCh38
rs997217292 Chr.19: 45142165-45142165 on Build GRCh38
rs753795761 Chr.19: 45142028-45142028 on Build GRCh38
rs1005854242 Chr.19: 45142419-45142419 on Build GRCh38
rs919213429 Chr.19: 45141991-45141991 on Build GRCh38
rs772928659 Chr.19: 45142147-45142147 on Build GRCh38
rs751997536 Chr.19: 45142178-45142178 on Build GRCh38
rs754606231 Chr.19: 45142145-45142145 on Build GRCh38
rs114206393 Chr.19: 45142364-45142364 on Build GRCh38
rs746945972 Chr.19: 45142088-45142088 on Build GRCh38
rs915714279 Chr.19: 45142279-45142280 on Build GRCh38
rs938756488 Chr.19: 45142281-45142281 on Build GRCh38
rs984648903 Chr.19: 45142009-45142009 on Build GRCh38
rs754862826 Chr.19: 45142365-45142365 on Build GRCh38
rs940068116 Chr.19: 45142349-45142349 on Build GRCh38
rs768157731 Chr.19: 45142291-45142291 on Build GRCh38
rs1038328120 Chr.19: 45142351-45142351 on Build GRCh38
rs756805764 Chr.19: 45142226-45142226 on Build GRCh38
rs1007740425 Chr.19: 45141957-45141957 on Build GRCh38
rs889965139 Chr.19: 45142129-45142129 on Build GRCh38
rs898574847 Chr.19: 45142159-45142159 on Build GRCh38
rs765270436 Chr.19: 45142350-45142350 on Build GRCh38
rs764169190 Chr.19: 45142307-45142307 on Build GRCh38
rs573821392 Chr.19: 45142271-45142271 on Build GRCh38
rs935848847 Chr.19: 45142081-45142081 on Build GRCh38
rs545675471 Chr.19: 45142017-45142017 on Build GRCh38
rs967092950 Chr.19: 45142206-45142206 on Build GRCh38
rs868224590 Chr.19: 45142168-45142168 on Build GRCh38
rs1004725725 Chr.19: 45142140-45142140 on Build GRCh38
rs576335577 Chr.19: 45141967-45141967 on Build GRCh38
rs977072315 Chr.19: 45142210-45142210 on Build GRCh38
rs780944974 Chr.19: 45142096-45142096 on Build GRCh38

View Full Product Details