Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45885839-45886344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF2BP1
Entrez Gene ID
26145
Gene Name
interferon regulatory factor 2 binding protein 1
Gene Aliases
-
UniGene
Hs.515477
Gene Chromosome Location
Chr.19: 45883607-45886170 on Build GRCh38
Transcripts
NM_015649.2

SNP Detail

SNP ID Chromosome Location
rs897053013 Chr.19: 45886256-45886256 on Build GRCh38
rs1010703636 Chr.19: 45885890-45885890 on Build GRCh38
rs530358041 Chr.19: 45886234-45886234 on Build GRCh38
rs995145536 Chr.19: 45886257-45886257 on Build GRCh38
rs1050308447 Chr.19: 45886062-45886062 on Build GRCh38
rs567927203 Chr.19: 45885985-45885985 on Build GRCh38
rs1029204470 Chr.19: 45885892-45885892 on Build GRCh38
rs944272126 Chr.19: 45886008-45886008 on Build GRCh38
rs1023159125 Chr.19: 45886173-45886173 on Build GRCh38
rs944549125 Chr.19: 45886253-45886253 on Build GRCh38
rs59100610 Chr.19: 45886273-45886273 on Build GRCh38
rs961686420 Chr.19: 45885920-45885920 on Build GRCh38
rs948116546 Chr.19: 45886248-45886248 on Build GRCh38
rs563792252 Chr.19: 45886132-45886134 on Build GRCh38
rs981158884 Chr.19: 45885895-45885895 on Build GRCh38
rs1009054134 Chr.19: 45885917-45885917 on Build GRCh38
rs1013573387 Chr.19: 45886155-45886155 on Build GRCh38
rs946632239 Chr.19: 45886059-45886059 on Build GRCh38
rs888934096 Chr.19: 45886067-45886067 on Build GRCh38
rs932184045 Chr.19: 45886259-45886259 on Build GRCh38
rs978233086 Chr.19: 45886320-45886320 on Build GRCh38
rs961035282 Chr.19: 45886096-45886096 on Build GRCh38
rs1050752718 Chr.19: 45886247-45886247 on Build GRCh38
rs934675449 Chr.19: 45886045-45886045 on Build GRCh38
rs532872862 Chr.19: 45886284-45886284 on Build GRCh38
rs796573237 Chr.19: 45886258-45886258 on Build GRCh38
rs912824030 Chr.19: 45886007-45886007 on Build GRCh38
rs1024006000 Chr.19: 45886082-45886093 on Build GRCh38
rs559332959 Chr.19: 45886244-45886244 on Build GRCh38
rs993169532 Chr.19: 45886087-45886087 on Build GRCh38
rs990150074 Chr.19: 45886217-45886217 on Build GRCh38
rs758653330 Chr.19: 45886299-45886300 on Build GRCh38
rs1015246517 Chr.19: 45886091-45886091 on Build GRCh38
rs1044274968 Chr.19: 45886251-45886252 on Build GRCh38
rs956086478 Chr.19: 45886212-45886212 on Build GRCh38
rs888909276 Chr.19: 45885894-45885894 on Build GRCh38
rs916058863 Chr.19: 45885996-45885996 on Build GRCh38
rs60858628 Chr.19: 45886270-45886270 on Build GRCh38
rs573845734 Chr.19: 45886259-45886259 on Build GRCh38
rs1027919010 Chr.19: 45885937-45885937 on Build GRCh38
rs1040171013 Chr.19: 45886258-45886258 on Build GRCh38
rs545032810 Chr.19: 45886131-45886131 on Build GRCh38
rs1026707869 Chr.19: 45886103-45886103 on Build GRCh38
rs924676077 Chr.19: 45886042-45886042 on Build GRCh38
rs113663555 Chr.19: 45886068-45886068 on Build GRCh38
rs910445585 Chr.19: 45886250-45886250 on Build GRCh38
rs895209539 Chr.19: 45886063-45886063 on Build GRCh38
rs971697537 Chr.19: 45885921-45885921 on Build GRCh38
rs576706057 Chr.19: 45886080-45886080 on Build GRCh38
rs536841600 Chr.19: 45886028-45886028 on Build GRCh38
rs893696036 Chr.19: 45885882-45885882 on Build GRCh38
rs893727361 Chr.19: 45886055-45886055 on Build GRCh38
rs1012564045 Chr.19: 45886314-45886314 on Build GRCh38
rs952238487 Chr.19: 45885978-45885978 on Build GRCh38
rs961149632 Chr.19: 45885925-45885925 on Build GRCh38
rs534492619 Chr.19: 45886178-45886178 on Build GRCh38
rs1019126129 Chr.19: 45886074-45886074 on Build GRCh38
rs561183984 Chr.19: 45886230-45886230 on Build GRCh38
rs924707546 Chr.19: 45886194-45886194 on Build GRCh38
rs541515269 Chr.19: 45886179-45886179 on Build GRCh38
rs1022900415 Chr.19: 45885860-45885860 on Build GRCh38
rs952309805 Chr.19: 45886151-45886151 on Build GRCh38
rs1053877173 Chr.19: 45886060-45886060 on Build GRCh38
rs1054352251 Chr.19: 45886052-45886052 on Build GRCh38
rs549517077 Chr.19: 45886092-45886093 on Build GRCh38
rs950192282 Chr.19: 45886002-45886002 on Build GRCh38
rs973784713 Chr.19: 45885926-45885926 on Build GRCh38
rs912721946 Chr.19: 45886152-45886152 on Build GRCh38
rs1018641770 Chr.19: 45885918-45885918 on Build GRCh38
rs757705093 Chr.19: 45886304-45886304 on Build GRCh38
rs973668803 Chr.19: 45886102-45886103 on Build GRCh38
rs1027282884 Chr.19: 45886112-45886112 on Build GRCh38
rs59664371 Chr.19: 45886252-45886252 on Build GRCh38
rs1048790512 Chr.19: 45886292-45886292 on Build GRCh38

View Full Product Details