Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49171246-49171740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM4
Entrez Gene ID
54795
Gene Name
transient receptor potential cation channel subfamily M member 4
Gene Aliases
LTrpC4, PFHB1B, TRPM4B, hTRPM4
UniGene
Hs.467101
Gene Chromosome Location
Chr.19: 49157741-49211841 on Build GRCh38
Transcripts
NM_001195227.1 NM_001321281.1 NM_001321282.1 NM_001321283.1 NM_001321285.1 NM_017636.3 XM_005259017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4624063 c.909C>T p.G303G Chr.19: 49171628-49171628 on Build GRCh38
1304900 c.1014C>G p.F338L Chr.19: 49171733-49171733 on Build GRCh38
6809557 c.970delG p.G325fs*31 Chr.19: 49171689-49171689 on Build GRCh38
5981579 c.878A>G p.Q293R Chr.19: 49171597-49171597 on Build GRCh38
4634434 c.955C>A p.L319M Chr.19: 49171674-49171674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750752873 Chr.19: 49171448-49171448 on Build GRCh38
rs764449383 Chr.19: 49171326-49171326 on Build GRCh38
rs7250823 Chr.19: 49171306-49171306 on Build GRCh38
rs770028767 Chr.19: 49171667-49171667 on Build GRCh38
rs777637675 Chr.19: 49171626-49171626 on Build GRCh38
rs201932657 Chr.19: 49171715-49171715 on Build GRCh38
rs746448119 Chr.19: 49171383-49171383 on Build GRCh38
rs748192005 Chr.19: 49171663-49171663 on Build GRCh38
rs752367612 Chr.19: 49171703-49171703 on Build GRCh38
rs774242926 Chr.19: 49171375-49171376 on Build GRCh38
rs750086956 Chr.19: 49171372-49171372 on Build GRCh38
rs765745854 Chr.19: 49171353-49171353 on Build GRCh38
rs760622386 Chr.19: 49171418-49171418 on Build GRCh38
rs778825571 Chr.19: 49171639-49171639 on Build GRCh38
rs576085034 Chr.19: 49171344-49171344 on Build GRCh38
rs775497145 Chr.19: 49171323-49171323 on Build GRCh38
rs373245795 Chr.19: 49171439-49171439 on Build GRCh38
rs1016914454 Chr.19: 49171291-49171291 on Build GRCh38
rs760812370 Chr.19: 49171325-49171325 on Build GRCh38
rs754390829 Chr.19: 49171335-49171335 on Build GRCh38
rs767568075 Chr.19: 49171310-49171310 on Build GRCh38
rs746083145 Chr.19: 49171412-49171412 on Build GRCh38
rs201341162 Chr.19: 49171272-49171272 on Build GRCh38
rs774842165 Chr.19: 49171700-49171700 on Build GRCh38
rs760136632 Chr.19: 49171706-49171706 on Build GRCh38
rs758849474 Chr.19: 49171451-49171451 on Build GRCh38
rs138095759 Chr.19: 49171654-49171654 on Build GRCh38
rs779763844 Chr.19: 49171375-49171375 on Build GRCh38
rs988711796 Chr.19: 49171405-49171407 on Build GRCh38
rs376221055 Chr.19: 49171547-49171547 on Build GRCh38
rs753753488 Chr.19: 49171713-49171713 on Build GRCh38
rs770369257 Chr.19: 49171584-49171584 on Build GRCh38
rs172147855 Chr.19: 49171597-49171597 on Build GRCh38
rs762393084 Chr.19: 49171424-49171424 on Build GRCh38
rs765902184 Chr.19: 49171459-49171459 on Build GRCh38
rs771054449 Chr.19: 49171488-49171488 on Build GRCh38
rs373391017 Chr.19: 49171397-49171397 on Build GRCh38
rs747295565 Chr.19: 49171554-49171554 on Build GRCh38
rs1024691665 Chr.19: 49171564-49171564 on Build GRCh38
rs750176955 Chr.19: 49171719-49171719 on Build GRCh38
rs548064960 Chr.19: 49171689-49171689 on Build GRCh38
rs753873502 Chr.19: 49171274-49171274 on Build GRCh38
rs765694592 Chr.19: 49171447-49171447 on Build GRCh38
rs753802920 Chr.19: 49171629-49171629 on Build GRCh38
rs141997826 Chr.19: 49171589-49171589 on Build GRCh38
rs891211634 Chr.19: 49171429-49171429 on Build GRCh38
rs113664674 Chr.19: 49171465-49171465 on Build GRCh38
rs369997286 Chr.19: 49171365-49171365 on Build GRCh38
rs761013435 Chr.19: 49171632-49171632 on Build GRCh38
rs1004303871 Chr.19: 49171431-49171431 on Build GRCh38
rs775940376 Chr.19: 49171620-49171620 on Build GRCh38
rs772389167 Chr.19: 49171553-49171553 on Build GRCh38
rs756970522 Chr.19: 49171636-49171636 on Build GRCh38
rs369373685 Chr.19: 49171275-49171275 on Build GRCh38
rs774388263 Chr.19: 49171526-49171526 on Build GRCh38
rs759820855 Chr.19: 49171701-49171701 on Build GRCh38
rs573938815 Chr.19: 49171594-49171594 on Build GRCh38
rs144889775 Chr.19: 49171532-49171532 on Build GRCh38
rs773630362 Chr.19: 49171631-49171632 on Build GRCh38
rs757286692 Chr.19: 49171714-49171714 on Build GRCh38
rs776917960 Chr.19: 49171578-49171578 on Build GRCh38
rs748060223 Chr.19: 49171467-49171467 on Build GRCh38
rs764918057 Chr.19: 49171675-49171677 on Build GRCh38
rs777689047 Chr.19: 49171531-49171531 on Build GRCh38
rs775793134 Chr.19: 49171417-49171417 on Build GRCh38
rs769550907 Chr.19: 49171402-49171402 on Build GRCh38
rs747847726 Chr.19: 49171413-49171413 on Build GRCh38
rs112651228 Chr.19: 49171514-49171514 on Build GRCh38
rs925160533 Chr.19: 49171279-49171279 on Build GRCh38
rs145771389 Chr.19: 49171707-49171707 on Build GRCh38
rs754541695 Chr.19: 49171385-49171385 on Build GRCh38
rs567594620 Chr.19: 49171387-49171387 on Build GRCh38
rs150333519 Chr.19: 49171469-49171469 on Build GRCh38
rs762468500 Chr.19: 49171445-49171445 on Build GRCh38
rs936589853 Chr.19: 49171297-49171297 on Build GRCh38
rs151205002 Chr.19: 49171416-49171416 on Build GRCh38
rs773880484 Chr.19: 49171585-49171585 on Build GRCh38
rs200707657 Chr.19: 49171579-49171579 on Build GRCh38
rs754419823 Chr.19: 49171466-49171466 on Build GRCh38
rs577738761 Chr.19: 49171655-49171655 on Build GRCh38
rs369594025 Chr.19: 49171438-49171438 on Build GRCh38
rs866691706 Chr.19: 49171635-49171635 on Build GRCh38
rs150368509 Chr.19: 49171604-49171604 on Build GRCh38
rs370515188 Chr.19: 49171695-49171695 on Build GRCh38
rs1048005773 Chr.19: 49171515-49171515 on Build GRCh38
rs752132967 Chr.19: 49171612-49171612 on Build GRCh38
rs865856899 Chr.19: 49171684-49171684 on Build GRCh38
rs768606152 Chr.19: 49171572-49171572 on Build GRCh38
rs749503838 Chr.19: 49171477-49171477 on Build GRCh38
rs964057093 Chr.19: 49171299-49171299 on Build GRCh38
rs528764152 Chr.19: 49171410-49171410 on Build GRCh38
rs548983214 Chr.19: 49171339-49171339 on Build GRCh38
rs763568053 Chr.19: 49171693-49171693 on Build GRCh38
rs373649726 Chr.19: 49171546-49171546 on Build GRCh38
rs747192078 Chr.19: 49171642-49171642 on Build GRCh38

View Full Product Details