Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50376341-50376834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1H2
Entrez Gene ID
7376
Gene Name
nuclear receptor subfamily 1 group H member 2
Gene Aliases
LXR-b, LXRB, NER, NER-I, RIP15, UNR
UniGene
Hs.432976
Gene Chromosome Location
Chr.19: 50376423-50383028 on Build GRCh38
Transcripts
NM_001256647.1 NM_007121.5

SNP Detail

SNP ID Chromosome Location
rs894889414 Chr.19: 50376761-50376761 on Build GRCh38
rs957646964 Chr.19: 50376791-50376791 on Build GRCh38
rs1046029516 Chr.19: 50376765-50376765 on Build GRCh38
rs1013329531 Chr.19: 50376762-50376762 on Build GRCh38
rs954555956 Chr.19: 50376598-50376598 on Build GRCh38
rs753916555 Chr.19: 50376802-50376802 on Build GRCh38
rs149289202 Chr.19: 50376542-50376542 on Build GRCh38
rs1054890827 Chr.19: 50376756-50376756 on Build GRCh38
rs936747456 Chr.19: 50376657-50376657 on Build GRCh38
rs563738395 Chr.19: 50376715-50376715 on Build GRCh38
rs1024470514 Chr.19: 50376509-50376509 on Build GRCh38
rs2546569 Chr.19: 50376785-50376785 on Build GRCh38
rs1004010518 Chr.19: 50376453-50376453 on Build GRCh38
rs765227601 Chr.19: 50376391-50376391 on Build GRCh38
rs192908663 Chr.19: 50376532-50376532 on Build GRCh38
rs1016057670 Chr.19: 50376389-50376389 on Build GRCh38
rs966842559 Chr.19: 50376617-50376617 on Build GRCh38
rs545420000 Chr.19: 50376704-50376704 on Build GRCh38
rs73932483 Chr.19: 50376578-50376578 on Build GRCh38
rs73932484 Chr.19: 50376688-50376688 on Build GRCh38
rs971620953 Chr.19: 50376681-50376681 on Build GRCh38
rs1032317586 Chr.19: 50376790-50376790 on Build GRCh38
rs924815060 Chr.19: 50376526-50376526 on Build GRCh38
rs760745351 Chr.19: 50376780-50376780 on Build GRCh38
rs990670722 Chr.19: 50376541-50376541 on Build GRCh38
rs1004750664 Chr.19: 50376567-50376567 on Build GRCh38
rs921595427 Chr.19: 50376403-50376403 on Build GRCh38
rs891068143 Chr.19: 50376447-50376447 on Build GRCh38
rs1054461044 Chr.19: 50376401-50376401 on Build GRCh38
rs76217494 Chr.19: 50376796-50376796 on Build GRCh38
rs939970953 Chr.19: 50376574-50376574 on Build GRCh38
rs919870499 Chr.19: 50376751-50376751 on Build GRCh38
rs761728607 Chr.19: 50376372-50376372 on Build GRCh38
rs1055633175 Chr.19: 50376670-50376670 on Build GRCh38
rs41385146 Chr.19: 50376637-50376637 on Build GRCh38
rs936120755 Chr.19: 50376399-50376399 on Build GRCh38
rs756620240 Chr.19: 50376706-50376706 on Build GRCh38
rs1042448774 Chr.19: 50376448-50376448 on Build GRCh38
rs1050974291 Chr.19: 50376441-50376441 on Build GRCh38
rs999216296 Chr.19: 50376788-50376788 on Build GRCh38
rs55794952 Chr.19: 50376418-50376418 on Build GRCh38
rs368768174 Chr.19: 50376707-50376707 on Build GRCh38
rs1028628146 Chr.19: 50376595-50376595 on Build GRCh38
rs560275202 Chr.19: 50376749-50376749 on Build GRCh38
rs1024629181 Chr.19: 50376662-50376662 on Build GRCh38
rs556401763 Chr.19: 50376609-50376609 on Build GRCh38
rs73932486 Chr.19: 50376693-50376693 on Build GRCh38
rs531348269 Chr.19: 50376725-50376725 on Build GRCh38
rs990352757 Chr.19: 50376692-50376692 on Build GRCh38
rs1024927803 Chr.19: 50376521-50376521 on Build GRCh38
rs1016111330 Chr.19: 50376572-50376572 on Build GRCh38
rs995686179 Chr.19: 50376468-50376468 on Build GRCh38
rs963365400 Chr.19: 50376390-50376390 on Build GRCh38
rs931098986 Chr.19: 50376753-50376753 on Build GRCh38
rs986791914 Chr.19: 50376700-50376700 on Build GRCh38
rs776342065 Chr.19: 50376368-50376368 on Build GRCh38
rs569617957 Chr.19: 50376748-50376748 on Build GRCh38
rs73932482 Chr.19: 50376512-50376512 on Build GRCh38
rs772643788 Chr.19: 50376414-50376414 on Build GRCh38
rs1037326455 Chr.19: 50376740-50376740 on Build GRCh38
rs898242090 Chr.19: 50376466-50376466 on Build GRCh38
rs578051887 Chr.19: 50376469-50376470 on Build GRCh38
rs924683545 Chr.19: 50376393-50376393 on Build GRCh38
rs902207975 Chr.19: 50376779-50376779 on Build GRCh38
rs144745282 Chr.19: 50376632-50376632 on Build GRCh38
rs891014405 Chr.19: 50376571-50376571 on Build GRCh38
rs1019917081 Chr.19: 50376815-50376815 on Build GRCh38
rs545407351 Chr.19: 50376485-50376485 on Build GRCh38
rs1037514179 Chr.19: 50376763-50376763 on Build GRCh38
rs932731709 Chr.19: 50376695-50376695 on Build GRCh38
rs768198377 Chr.19: 50376616-50376631 on Build GRCh38
rs995364473 Chr.19: 50376600-50376600 on Build GRCh38
rs991804030 Chr.19: 50376506-50376506 on Build GRCh38
rs977612174 Chr.19: 50376513-50376513 on Build GRCh38
rs1033337055 Chr.19: 50376476-50376476 on Build GRCh38
rs535784543 Chr.19: 50376404-50376404 on Build GRCh38
rs903981009 Chr.19: 50376452-50376452 on Build GRCh38
rs978103468 Chr.19: 50376682-50376682 on Build GRCh38
rs1036848209 Chr.19: 50376577-50376577 on Build GRCh38
rs932616863 Chr.19: 50376544-50376544 on Build GRCh38
rs1051098655 Chr.19: 50376563-50376563 on Build GRCh38
rs1013925235 Chr.19: 50376364-50376364 on Build GRCh38
rs35447988 Chr.19: 50376727-50376728 on Build GRCh38
rs565962754 Chr.19: 50376478-50376478 on Build GRCh38
rs963119396 Chr.19: 50376573-50376573 on Build GRCh38
rs541778966 Chr.19: 50376683-50376683 on Build GRCh38
rs983188327 Chr.19: 50376387-50376387 on Build GRCh38
rs1015877047 Chr.19: 50376458-50376458 on Build GRCh38

View Full Product Details