Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55294076-55294484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRSK1
Entrez Gene ID
84446
Gene Name
BR serine/threonine kinase 1
Gene Aliases
hSAD1
UniGene
Hs.182081
Gene Chromosome Location
Chr.19: 55284101-55312562 on Build GRCh38
Transcripts
NM_032430.1 XM_005259327.3 XM_011527395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960534 c.609+7G>C p.? Chr.19: 55294254-55294254 on Build GRCh38
3539416 c.537C>T p.S179S Chr.19: 55294095-55294095 on Build GRCh38
6612994 c.534G>A p.A178A Chr.19: 55294092-55294092 on Build GRCh38
5883182 c.548G>A p.G183E Chr.19: 55294106-55294106 on Build GRCh38
1001462 c.610G>A p.G204R Chr.19: 55294329-55294329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767100726 Chr.19: 55294318-55294318 on Build GRCh38
rs774421200 Chr.19: 55294273-55294273 on Build GRCh38
rs760033577 Chr.19: 55294411-55294411 on Build GRCh38
rs761004264 Chr.19: 55294171-55294171 on Build GRCh38
rs768373650 Chr.19: 55294416-55294416 on Build GRCh38
rs757218974 Chr.19: 55294128-55294128 on Build GRCh38
rs763779659 Chr.19: 55294311-55294311 on Build GRCh38
rs755209419 Chr.19: 55294358-55294358 on Build GRCh38
rs369642683 Chr.19: 55294359-55294359 on Build GRCh38
rs372316579 Chr.19: 55294306-55294306 on Build GRCh38
rs368835496 Chr.19: 55294299-55294299 on Build GRCh38
rs758019189 Chr.19: 55294196-55294196 on Build GRCh38
rs753707281 Chr.19: 55294443-55294443 on Build GRCh38
rs375079381 Chr.19: 55294275-55294275 on Build GRCh38
rs762342387 Chr.19: 55294164-55294164 on Build GRCh38
rs142824115 Chr.19: 55294260-55294260 on Build GRCh38
rs766761915 Chr.19: 55294176-55294176 on Build GRCh38
rs781137543 Chr.19: 55294131-55294131 on Build GRCh38
rs373604846 Chr.19: 55294413-55294413 on Build GRCh38
rs377470339 Chr.19: 55294125-55294125 on Build GRCh38
rs374051558 Chr.19: 55294229-55294229 on Build GRCh38
rs774369769 Chr.19: 55294159-55294159 on Build GRCh38
rs199858665 Chr.19: 55294264-55294264 on Build GRCh38
rs371273066 Chr.19: 55294261-55294261 on Build GRCh38
rs371209071 Chr.19: 55294150-55294150 on Build GRCh38
rs201059652 Chr.19: 55294155-55294155 on Build GRCh38
rs771401240 Chr.19: 55294405-55294405 on Build GRCh38
rs142326820 Chr.19: 55294454-55294454 on Build GRCh38
rs752381874 Chr.19: 55294101-55294101 on Build GRCh38
rs370327413 Chr.19: 55294199-55294199 on Build GRCh38
rs776872208 Chr.19: 55294410-55294410 on Build GRCh38
rs749913408 Chr.19: 55294431-55294431 on Build GRCh38
rs1011452467 Chr.19: 55294337-55294337 on Build GRCh38
rs761532010 Chr.19: 55294423-55294423 on Build GRCh38
rs753605137 Chr.19: 55294352-55294352 on Build GRCh38
rs955676508 Chr.19: 55294456-55294456 on Build GRCh38
rs754836262 Chr.19: 55294447-55294447 on Build GRCh38
rs751045755 Chr.19: 55294214-55294214 on Build GRCh38
rs999577153 Chr.19: 55294165-55294165 on Build GRCh38
rs200918462 Chr.19: 55294278-55294278 on Build GRCh38
rs777887552 Chr.19: 55294402-55294402 on Build GRCh38
rs201801123 Chr.19: 55294154-55294154 on Build GRCh38
rs773660197 Chr.19: 55294298-55294298 on Build GRCh38
rs887560380 Chr.19: 55294305-55294305 on Build GRCh38
rs757914003 Chr.19: 55294104-55294104 on Build GRCh38
rs766285203 Chr.19: 55294438-55294438 on Build GRCh38
rs200460715 Chr.19: 55294326-55294326 on Build GRCh38
rs375155431 Chr.19: 55294166-55294166 on Build GRCh38
rs748366809 Chr.19: 55294394-55294394 on Build GRCh38
rs778425492 Chr.19: 55294207-55294207 on Build GRCh38
rs752332196 Chr.19: 55294186-55294186 on Build GRCh38
rs766954327 Chr.19: 55294428-55294428 on Build GRCh38
rs777398882 Chr.19: 55294109-55294109 on Build GRCh38
rs779748376 Chr.19: 55294351-55294351 on Build GRCh38
rs1044827065 Chr.19: 55294304-55294304 on Build GRCh38
rs747502391 Chr.19: 55294404-55294404 on Build GRCh38
rs772127628 Chr.19: 55294276-55294276 on Build GRCh38
rs911216897 Chr.19: 55294217-55294217 on Build GRCh38
rs551181362 Chr.19: 55294347-55294347 on Build GRCh38
rs751241925 Chr.19: 55294312-55294312 on Build GRCh38
rs568960841 Chr.19: 55294302-55294302 on Build GRCh38
rs756033784 Chr.19: 55294334-55294334 on Build GRCh38
rs548024845 Chr.19: 55294269-55294269 on Build GRCh38
rs930101165 Chr.19: 55294280-55294280 on Build GRCh38
rs749498500 Chr.19: 55294266-55294266 on Build GRCh38
rs1055904619 Chr.19: 55294307-55294307 on Build GRCh38
rs147671102 Chr.19: 55294451-55294451 on Build GRCh38
rs368368382 Chr.19: 55294170-55294170 on Build GRCh38
rs201444523 Chr.19: 55294313-55294313 on Build GRCh38
rs935215861 Chr.19: 55294250-55294250 on Build GRCh38
rs757271976 Chr.19: 55294228-55294228 on Build GRCh38
rs745811034 Chr.19: 55294257-55294257 on Build GRCh38
rs35068852 Chr.19: 55294332-55294332 on Build GRCh38
rs868048846 Chr.19: 55294455-55294455 on Build GRCh38
rs376778077 Chr.19: 55294435-55294435 on Build GRCh38
rs779166281 Chr.19: 55294385-55294385 on Build GRCh38
rs762516603 Chr.19: 55294310-55294310 on Build GRCh38

View Full Product Details