Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55383957-55384456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM238
Entrez Gene ID
388564
Gene Name
transmembrane protein 238
Gene Aliases
-
UniGene
Hs.534672
Gene Chromosome Location
Chr.19: 55379244-55384259 on Build GRCh38
Transcripts
NM_001190764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278061 c.172G>A p.A58T Chr.19: 55384088-55384088 on Build GRCh38
6049611 c.105G>C p.R35R Chr.19: 55384155-55384155 on Build GRCh38
7289020 c.298G>T p.E100* Chr.19: 55383962-55383962 on Build GRCh38
6419585 c.110C>A p.A37E Chr.19: 55384150-55384150 on Build GRCh38
6035148 c.225C>G p.G75G Chr.19: 55384035-55384035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016239907 Chr.19: 55384375-55384375 on Build GRCh38
rs754143591 Chr.19: 55384101-55384101 on Build GRCh38
rs113093748 Chr.19: 55384391-55384391 on Build GRCh38
rs890030314 Chr.19: 55384412-55384412 on Build GRCh38
rs766136378 Chr.19: 55384097-55384097 on Build GRCh38
rs545709932 Chr.19: 55384155-55384155 on Build GRCh38
rs367637151 Chr.19: 55384408-55384408 on Build GRCh38
rs760500389 Chr.19: 55384006-55384006 on Build GRCh38
rs959342928 Chr.19: 55384428-55384428 on Build GRCh38
rs146031178 Chr.19: 55384335-55384335 on Build GRCh38
rs933548190 Chr.19: 55384312-55384312 on Build GRCh38
rs937666999 Chr.19: 55384201-55384201 on Build GRCh38
rs1036674003 Chr.19: 55384379-55384379 on Build GRCh38
rs1016292409 Chr.19: 55384064-55384064 on Build GRCh38
rs892302995 Chr.19: 55384078-55384078 on Build GRCh38
rs542716879 Chr.19: 55384290-55384290 on Build GRCh38
rs909638887 Chr.19: 55384350-55384350 on Build GRCh38
rs148442876 Chr.19: 55384338-55384338 on Build GRCh38
rs983916981 Chr.19: 55384341-55384341 on Build GRCh38
rs778466254 Chr.19: 55384210-55384210 on Build GRCh38
rs559419682 Chr.19: 55384332-55384332 on Build GRCh38
rs1034001291 Chr.19: 55384411-55384412 on Build GRCh38
rs1041879605 Chr.19: 55384282-55384282 on Build GRCh38
rs989123942 Chr.19: 55384434-55384434 on Build GRCh38
rs755093459 Chr.19: 55384117-55384117 on Build GRCh38
rs575843500 Chr.19: 55384173-55384173 on Build GRCh38
rs189206666 Chr.19: 55384171-55384171 on Build GRCh38
rs779066781 Chr.19: 55384143-55384143 on Build GRCh38
rs767269394 Chr.19: 55384330-55384330 on Build GRCh38
rs982442049 Chr.19: 55384178-55384178 on Build GRCh38
rs939743824 Chr.19: 55384376-55384376 on Build GRCh38
rs953504381 Chr.19: 55383986-55383986 on Build GRCh38
rs889407706 Chr.19: 55384359-55384359 on Build GRCh38
rs970639335 Chr.19: 55384177-55384177 on Build GRCh38
rs1870074 Chr.19: 55384220-55384220 on Build GRCh38
rs544484361 Chr.19: 55384213-55384213 on Build GRCh38
rs572871340 Chr.19: 55384017-55384017 on Build GRCh38
rs868745292 Chr.19: 55384025-55384025 on Build GRCh38
rs3745272 Chr.19: 55384395-55384395 on Build GRCh38
rs879301055 Chr.19: 55384216-55384216 on Build GRCh38
rs374241331 Chr.19: 55384287-55384287 on Build GRCh38
rs1007596155 Chr.19: 55384057-55384057 on Build GRCh38
rs926602149 Chr.19: 55384198-55384198 on Build GRCh38
rs970966407 Chr.19: 55384397-55384397 on Build GRCh38

View Full Product Details