Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55482830-55483338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF628
Entrez Gene ID
89887
Gene Name
zinc finger protein 628
Gene Aliases
ZEC, Zfp628
UniGene
Hs.525209
Gene Chromosome Location
Chr.19: 55476332-55484487 on Build GRCh38
Transcripts
NM_033113.2 XM_005259371.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7225683 c.1835A>G p.H612R Chr.19: 55483040-55483040 on Build GRCh38
5432137 c.1637C>T p.T546M Chr.19: 55482842-55482842 on Build GRCh38
4413365 c.1909C>G p.L637V Chr.19: 55483102-55483102 on Build GRCh38
6499607 c.2003C>T p.P668L Chr.19: 55483208-55483208 on Build GRCh38
6021893 c.1855G>A p.E619K Chr.19: 55483048-55483048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995407626 Chr.19: 55483272-55483272 on Build GRCh38
rs964570889 Chr.19: 55483204-55483204 on Build GRCh38
rs866949540 Chr.19: 55482919-55482919 on Build GRCh38
rs761377966 Chr.19: 55482966-55482966 on Build GRCh38
rs775220906 Chr.19: 55483309-55483309 on Build GRCh38
rs367866791 Chr.19: 55482985-55482985 on Build GRCh38
rs756660858 Chr.19: 55483201-55483201 on Build GRCh38
rs748239369 Chr.19: 55482903-55482903 on Build GRCh38
rs750471793 Chr.19: 55483198-55483198 on Build GRCh38
rs139724904 Chr.19: 55482977-55482977 on Build GRCh38
rs752898034 Chr.19: 55483130-55483130 on Build GRCh38
rs764379159 Chr.19: 55483169-55483169 on Build GRCh38
rs767545312 Chr.19: 55483177-55483177 on Build GRCh38
rs767004725 Chr.19: 55483056-55483056 on Build GRCh38
rs868350072 Chr.19: 55482889-55482889 on Build GRCh38
rs769831272 Chr.19: 55483307-55483307 on Build GRCh38
rs751725945 Chr.19: 55483133-55483133 on Build GRCh38
rs778452703 Chr.19: 55483095-55483095 on Build GRCh38
rs773054424 Chr.19: 55483060-55483060 on Build GRCh38
rs766395275 Chr.19: 55483061-55483061 on Build GRCh38
rs765562671 Chr.19: 55483068-55483068 on Build GRCh38
rs756861789 Chr.19: 55483280-55483280 on Build GRCh38
rs758434074 Chr.19: 55483074-55483074 on Build GRCh38
rs766235765 Chr.19: 55482981-55482981 on Build GRCh38
rs949751649 Chr.19: 55483291-55483291 on Build GRCh38
rs771813788 Chr.19: 55483154-55483154 on Build GRCh38
rs978583793 Chr.19: 55483254-55483254 on Build GRCh38
rs770081794 Chr.19: 55483094-55483094 on Build GRCh38
rs758438294 Chr.19: 55482995-55482995 on Build GRCh38
rs750876337 Chr.19: 55483144-55483144 on Build GRCh38
rs757813058 Chr.19: 55483082-55483082 on Build GRCh38
rs748897137 Chr.19: 55483075-55483075 on Build GRCh38
rs113592428 Chr.19: 55483131-55483131 on Build GRCh38
rs756501494 Chr.19: 55483151-55483151 on Build GRCh38
rs749592612 Chr.19: 55483153-55483153 on Build GRCh38
rs988572983 Chr.19: 55483277-55483277 on Build GRCh38
rs1032790087 Chr.19: 55483262-55483262 on Build GRCh38
rs149796895 Chr.19: 55482983-55482983 on Build GRCh38
rs779265539 Chr.19: 55482987-55482987 on Build GRCh38
rs777495635 Chr.19: 55483273-55483273 on Build GRCh38
rs767356947 Chr.19: 55482968-55482968 on Build GRCh38
rs752906914 Chr.19: 55483070-55483070 on Build GRCh38
rs773906342 Chr.19: 55482963-55482963 on Build GRCh38
rs774494338 Chr.19: 55483170-55483170 on Build GRCh38
rs200905523 Chr.19: 55482878-55482885 on Build GRCh38
rs762555311 Chr.19: 55482948-55482948 on Build GRCh38
rs577993609 Chr.19: 55483203-55483203 on Build GRCh38
rs375761117 Chr.19: 55483050-55483050 on Build GRCh38
rs375880107 Chr.19: 55482942-55482942 on Build GRCh38
rs761900516 Chr.19: 55483173-55483173 on Build GRCh38
rs764202274 Chr.19: 55483132-55483132 on Build GRCh38
rs750249037 Chr.19: 55482857-55482857 on Build GRCh38
rs771592419 Chr.19: 55483101-55483101 on Build GRCh38
rs964516226 Chr.19: 55482959-55482959 on Build GRCh38
rs1017430499 Chr.19: 55482952-55482952 on Build GRCh38
rs565700401 Chr.19: 55483086-55483086 on Build GRCh38
rs201469671 Chr.19: 55483065-55483065 on Build GRCh38
rs962589524 Chr.19: 55483263-55483263 on Build GRCh38
rs771320119 Chr.19: 55483020-55483020 on Build GRCh38
rs12981044 Chr.19: 55482873-55482873 on Build GRCh38
rs755138882 Chr.19: 55482878-55482878 on Build GRCh38
rs987396187 Chr.19: 55482934-55482934 on Build GRCh38
rs776733178 Chr.19: 55483164-55483164 on Build GRCh38
rs376033958 Chr.19: 55482856-55482856 on Build GRCh38
rs746611968 Chr.19: 55483276-55483276 on Build GRCh38
rs954816539 Chr.19: 55482880-55482880 on Build GRCh38
rs780894245 Chr.19: 55483292-55483292 on Build GRCh38
rs370949939 Chr.19: 55482940-55482940 on Build GRCh38
rs759511441 Chr.19: 55483166-55483166 on Build GRCh38
rs777023117 Chr.19: 55483021-55483021 on Build GRCh38
rs746216385 Chr.19: 55483038-55483038 on Build GRCh38
rs368625084 Chr.19: 55482915-55482915 on Build GRCh38
rs755232674 Chr.19: 55483241-55483241 on Build GRCh38
rs770589755 Chr.19: 55483308-55483308 on Build GRCh38
rs759401484 Chr.19: 55483066-55483066 on Build GRCh38
rs780436601 Chr.19: 55482868-55482868 on Build GRCh38
rs773983704 Chr.19: 55483049-55483049 on Build GRCh38
rs775406992 Chr.19: 55483168-55483168 on Build GRCh38
rs985318618 Chr.19: 55482971-55482971 on Build GRCh38
rs766781614 Chr.19: 55483221-55483221 on Build GRCh38
rs769591940 Chr.19: 55483167-55483167 on Build GRCh38
rs765079958 Chr.19: 55483129-55483129 on Build GRCh38
rs746523993 Chr.19: 55483160-55483160 on Build GRCh38
rs867236606 Chr.19: 55483259-55483259 on Build GRCh38
rs371736358 Chr.19: 55483011-55483011 on Build GRCh38
rs1010875151 Chr.19: 55482907-55482907 on Build GRCh38
rs776644070 Chr.19: 55483126-55483126 on Build GRCh38
rs746232658 Chr.19: 55483092-55483092 on Build GRCh38
rs550689648 Chr.19: 55482972-55482972 on Build GRCh38
rs956013312 Chr.19: 55483264-55483264 on Build GRCh38
rs1000040544 Chr.19: 55482939-55482939 on Build GRCh38
rs958523259 Chr.19: 55483300-55483300 on Build GRCh38
rs759417772 Chr.19: 55483127-55483127 on Build GRCh38
rs1043971363 Chr.19: 55483315-55483315 on Build GRCh38
rs745352238 Chr.19: 55483305-55483305 on Build GRCh38
rs747339144 Chr.19: 55482938-55482938 on Build GRCh38
rs777289113 Chr.19: 55483157-55483157 on Build GRCh38
rs772825368 Chr.19: 55483113-55483113 on Build GRCh38
rs78991146 Chr.19: 55483046-55483046 on Build GRCh38
rs777873726 Chr.19: 55483078-55483078 on Build GRCh38
rs780507059 Chr.19: 55483152-55483152 on Build GRCh38
rs999543676 Chr.19: 55483165-55483165 on Build GRCh38
rs779171454 Chr.19: 55482891-55482891 on Build GRCh38
rs530772643 Chr.19: 55482918-55482918 on Build GRCh38
rs771060228 Chr.19: 55483125-55483125 on Build GRCh38
rs200785885 Chr.19: 55483163-55483163 on Build GRCh38
rs763711566 Chr.19: 55482958-55482958 on Build GRCh38
rs371420206 Chr.19: 55483137-55483137 on Build GRCh38
rs200197263 Chr.19: 55483119-55483119 on Build GRCh38
rs367631740 Chr.19: 55483108-55483108 on Build GRCh38
rs751564703 Chr.19: 55483080-55483080 on Build GRCh38
rs1017482348 Chr.19: 55483202-55483202 on Build GRCh38
rs755866837 Chr.19: 55482862-55482862 on Build GRCh38
rs902594112 Chr.19: 55482931-55482931 on Build GRCh38
rs537391720 Chr.19: 55483224-55483224 on Build GRCh38
rs778358728 Chr.19: 55483311-55483311 on Build GRCh38
rs748244490 Chr.19: 55482992-55482992 on Build GRCh38
rs781252239 Chr.19: 55482852-55482853 on Build GRCh38
rs955353170 Chr.19: 55482969-55482969 on Build GRCh38

View Full Product Details