Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57499821-57500325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF773
Entrez Gene ID
374928
Gene Name
zinc finger protein 773
Gene Aliases
ZNF419B
UniGene
Hs.579576
Gene Chromosome Location
Chr.19: 57499854-57516423 on Build GRCh38
Transcripts
NM_001304334.1 NM_001304335.1 NM_001304336.1 NM_001304337.1 NM_198542.2 NR_130705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6847856 c.102+2T>G p.? Chr.19: 57500115-57500115 on Build GRCh38
5092181 c.28C>T p.R10W Chr.19: 57500039-57500039 on Build GRCh38
5092182 c.57G>A p.P19P Chr.19: 57500068-57500068 on Build GRCh38
6832956 c.56C>T p.P19L Chr.19: 57500067-57500067 on Build GRCh38
7399661 c.100C>T p.Q34* Chr.19: 57500111-57500111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901266089 Chr.19: 57500029-57500029 on Build GRCh38
rs1033029119 Chr.19: 57499912-57499912 on Build GRCh38
rs367895171 Chr.19: 57500297-57500297 on Build GRCh38
rs748664324 Chr.19: 57500061-57500061 on Build GRCh38
rs1013901148 Chr.19: 57500288-57500288 on Build GRCh38
rs368836011 Chr.19: 57500045-57500045 on Build GRCh38
rs1047166043 Chr.19: 57500236-57500236 on Build GRCh38
rs986091566 Chr.19: 57499949-57499949 on Build GRCh38
rs894738417 Chr.19: 57500280-57500280 on Build GRCh38
rs994333694 Chr.19: 57499854-57499854 on Build GRCh38
rs757305978 Chr.19: 57500165-57500165 on Build GRCh38
rs371511968 Chr.19: 57499856-57499856 on Build GRCh38
rs747220315 Chr.19: 57500063-57500063 on Build GRCh38
rs758415435 Chr.19: 57500062-57500062 on Build GRCh38
rs772183662 Chr.19: 57499990-57499990 on Build GRCh38
rs540161828 Chr.19: 57499945-57499945 on Build GRCh38
rs149399810 Chr.19: 57499877-57499877 on Build GRCh38
rs376086087 Chr.19: 57499893-57499893 on Build GRCh38
rs551019855 Chr.19: 57500057-57500057 on Build GRCh38
rs930069219 Chr.19: 57500224-57500224 on Build GRCh38
rs976727563 Chr.19: 57500143-57500143 on Build GRCh38
rs558277082 Chr.19: 57499903-57499903 on Build GRCh38
rs867597947 Chr.19: 57500156-57500156 on Build GRCh38
rs760886432 Chr.19: 57500003-57500003 on Build GRCh38
rs894144791 Chr.19: 57500056-57500056 on Build GRCh38
rs190821966 Chr.19: 57500007-57500007 on Build GRCh38
rs751477551 Chr.19: 57500044-57500044 on Build GRCh38
rs184177703 Chr.19: 57499922-57499922 on Build GRCh38
rs560449833 Chr.19: 57499968-57499968 on Build GRCh38
rs1056412152 Chr.19: 57500272-57500272 on Build GRCh38
rs779527773 Chr.19: 57499978-57499978 on Build GRCh38
rs775217745 Chr.19: 57500016-57500016 on Build GRCh38
rs750055477 Chr.19: 57500049-57500049 on Build GRCh38
rs739586 Chr.19: 57500142-57500142 on Build GRCh38
rs762910500 Chr.19: 57500039-57500039 on Build GRCh38
rs758893174 Chr.19: 57500145-57500145 on Build GRCh38
rs1033243241 Chr.19: 57500080-57500080 on Build GRCh38
rs766178824 Chr.19: 57500105-57500105 on Build GRCh38
rs761750002 Chr.19: 57500086-57500086 on Build GRCh38
rs773377975 Chr.19: 57500001-57500001 on Build GRCh38
rs374358853 Chr.19: 57500209-57500209 on Build GRCh38
rs771147362 Chr.19: 57500064-57500064 on Build GRCh38
rs1047688759 Chr.19: 57500042-57500042 on Build GRCh38
rs146373125 Chr.19: 57499906-57499906 on Build GRCh38
rs774574311 Chr.19: 57499972-57499972 on Build GRCh38
rs753524530 Chr.19: 57500108-57500108 on Build GRCh38
rs748283891 Chr.19: 57499981-57499981 on Build GRCh38
rs749833019 Chr.19: 57499967-57499967 on Build GRCh38
rs941578032 Chr.19: 57500247-57500247 on Build GRCh38
rs977667191 Chr.19: 57499934-57499934 on Build GRCh38
rs766879579 Chr.19: 57500052-57500052 on Build GRCh38
rs764058968 Chr.19: 57500043-57500043 on Build GRCh38
rs138317120 Chr.19: 57500079-57500079 on Build GRCh38
rs528537582 Chr.19: 57499996-57499996 on Build GRCh38
rs377637832 Chr.19: 57499964-57499964 on Build GRCh38
rs142896117 Chr.19: 57500093-57500093 on Build GRCh38
rs910505775 Chr.19: 57500133-57500133 on Build GRCh38
rs12463137 Chr.19: 57500053-57500053 on Build GRCh38
rs368071146 Chr.19: 57500132-57500132 on Build GRCh38
rs776803796 Chr.19: 57500067-57500067 on Build GRCh38
rs910028584 Chr.19: 57500243-57500243 on Build GRCh38
rs992570493 Chr.19: 57499909-57499910 on Build GRCh38
rs538273377 Chr.19: 57500173-57500173 on Build GRCh38
rs755464072 Chr.19: 57500124-57500124 on Build GRCh38
rs565656361 Chr.19: 57499851-57499851 on Build GRCh38
rs772735204 Chr.19: 57500095-57500095 on Build GRCh38
rs942772724 Chr.19: 57499847-57499847 on Build GRCh38
rs574858028 Chr.19: 57500304-57500304 on Build GRCh38
rs558111663 Chr.19: 57500233-57500233 on Build GRCh38
rs754054196 Chr.19: 57500137-57500138 on Build GRCh38
rs529873176 Chr.19: 57499941-57499941 on Build GRCh38
rs560938157 Chr.19: 57499940-57499940 on Build GRCh38
rs777748485 Chr.19: 57500152-57500152 on Build GRCh38
rs888702004 Chr.19: 57499859-57499859 on Build GRCh38
rs376340455 Chr.19: 57499915-57499915 on Build GRCh38
rs925865894 Chr.19: 57499935-57499935 on Build GRCh38
rs942734975 Chr.19: 57499998-57499998 on Build GRCh38
rs901339671 Chr.19: 57499853-57499853 on Build GRCh38
rs779625870 Chr.19: 57500060-57500060 on Build GRCh38
rs780406880 Chr.19: 57499962-57499962 on Build GRCh38
rs553584456 Chr.19: 57499873-57499873 on Build GRCh38
rs749132050 Chr.19: 57500070-57500070 on Build GRCh38
rs747154752 Chr.19: 57500154-57500154 on Build GRCh38

View Full Product Details