Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10129122-10129666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM2
Entrez Gene ID
6241
Gene Name
ribonucleotide reductase regulatory subunit M2
Gene Aliases
R2, RR2, RR2M
UniGene
Hs.226390
Gene Chromosome Location
Chr.2: 10122568-10131419 on Build GRCh38
Transcripts
NM_001034.3 NM_001165931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305480 c.1030G>C p.E344Q Chr.2: 10129246-10129246 on Build GRCh38
1305479 c.1210G>C p.E404Q Chr.2: 10129246-10129246 on Build GRCh38
4083849 c.1214A>C p.N405T Chr.2: 10129250-10129250 on Build GRCh38
6290817 c.1123A>G p.M375V Chr.2: 10129339-10129339 on Build GRCh38
1004617 c.996G>A p.M332I Chr.2: 10129133-10129133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142939100 Chr.2: 10129174-10129174 on Build GRCh38
rs62129882 Chr.2: 10129204-10129204 on Build GRCh38
rs577803252 Chr.2: 10129576-10129576 on Build GRCh38
rs371603045 Chr.2: 10129417-10129418 on Build GRCh38
rs749647333 Chr.2: 10129401-10129401 on Build GRCh38
rs765011415 Chr.2: 10129247-10129247 on Build GRCh38
rs773615566 Chr.2: 10129178-10129178 on Build GRCh38
rs373279736 Chr.2: 10129157-10129157 on Build GRCh38
rs764059345 Chr.2: 10129418-10129418 on Build GRCh38
rs781580850 Chr.2: 10129165-10129165 on Build GRCh38
rs1013158466 Chr.2: 10129214-10129214 on Build GRCh38
rs373151401 Chr.2: 10129234-10129234 on Build GRCh38
rs557947933 Chr.2: 10129558-10129558 on Build GRCh38
rs756637379 Chr.2: 10129387-10129387 on Build GRCh38
rs774957221 Chr.2: 10129624-10129624 on Build GRCh38
rs79162203 Chr.2: 10129418-10129418 on Build GRCh38
rs756710024 Chr.2: 10129418-10129419 on Build GRCh38
rs780697028 Chr.2: 10129351-10129351 on Build GRCh38
rs771246264 Chr.2: 10129202-10129202 on Build GRCh38
rs748663501 Chr.2: 10129368-10129368 on Build GRCh38
rs371457678 Chr.2: 10129356-10129356 on Build GRCh38
rs370125026 Chr.2: 10129195-10129195 on Build GRCh38
rs567302711 Chr.2: 10129435-10129435 on Build GRCh38
rs778037879 Chr.2: 10129396-10129396 on Build GRCh38
rs78311808 Chr.2: 10129417-10129417 on Build GRCh38
rs761749812 Chr.2: 10129242-10129242 on Build GRCh38
rs5030757 Chr.2: 10129421-10129421 on Build GRCh38
rs771373014 Chr.2: 10129402-10129402 on Build GRCh38
rs768110234 Chr.2: 10129164-10129164 on Build GRCh38
rs772288724 Chr.2: 10129430-10129430 on Build GRCh38
rs975301006 Chr.2: 10129459-10129459 on Build GRCh38
rs151112936 Chr.2: 10129486-10129486 on Build GRCh38
rs535862183 Chr.2: 10129464-10129464 on Build GRCh38
rs776799796 Chr.2: 10129515-10129516 on Build GRCh38
rs751173166 Chr.2: 10129317-10129317 on Build GRCh38
rs142631876 Chr.2: 10129240-10129240 on Build GRCh38
rs376458388 Chr.2: 10129407-10129407 on Build GRCh38
rs778621406 Chr.2: 10129275-10129275 on Build GRCh38
rs15516 Chr.2: 10129354-10129354 on Build GRCh38
rs186654277 Chr.2: 10129485-10129485 on Build GRCh38
rs534347219 Chr.2: 10129601-10129601 on Build GRCh38
rs759707613 Chr.2: 10129230-10129230 on Build GRCh38
rs758099243 Chr.2: 10129254-10129254 on Build GRCh38
rs923485611 Chr.2: 10129468-10129468 on Build GRCh38
rs774622230 Chr.2: 10129216-10129216 on Build GRCh38
rs750203926 Chr.2: 10129248-10129248 on Build GRCh38
rs978160437 Chr.2: 10129541-10129541 on Build GRCh38
rs934549855 Chr.2: 10129177-10129177 on Build GRCh38
rs538123832 Chr.2: 10129532-10129532 on Build GRCh38

View Full Product Details