Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11194100-11194596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROCK2
Entrez Gene ID
9475
Gene Name
Rho associated coiled-coil containing protein kinase 2
Gene Aliases
ROCK-II
UniGene
Hs.681743
Gene Chromosome Location
Chr.2: 11179759-11345407 on Build GRCh38
Transcripts
NM_001321643.1 NM_004850.4 XM_005246190.3 XM_011510417.2 XM_017005378.1 XM_017005379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1005575 c.3574G>T p.E1192* Chr.2: 11194290-11194290 on Build GRCh38
6768407 c.3529G>A p.V1177I Chr.2: 11194335-11194335 on Build GRCh38
441004 c.3560G>A p.S1187N Chr.2: 11194304-11194304 on Build GRCh38
21111 c.3580T>C p.S1194P Chr.2: 11194284-11194284 on Build GRCh38
263412 c.3547C>T p.L1183F Chr.2: 11194317-11194317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914992023 Chr.2: 11194530-11194530 on Build GRCh38
rs776237815 Chr.2: 11194412-11194412 on Build GRCh38
rs745316302 Chr.2: 11194224-11194224 on Build GRCh38
rs55839233 Chr.2: 11194258-11194258 on Build GRCh38
rs927824946 Chr.2: 11194489-11194489 on Build GRCh38
rs763206938 Chr.2: 11194320-11194322 on Build GRCh38
rs368885239 Chr.2: 11194324-11194324 on Build GRCh38
rs190290058 Chr.2: 11194164-11194164 on Build GRCh38
rs761936331 Chr.2: 11194296-11194296 on Build GRCh38
rs1040827600 Chr.2: 11194537-11194537 on Build GRCh38
rs770546446 Chr.2: 11194207-11194207 on Build GRCh38
rs778287254 Chr.2: 11194223-11194223 on Build GRCh38
rs969329840 Chr.2: 11194474-11194474 on Build GRCh38
rs956365638 Chr.2: 11194510-11194510 on Build GRCh38
rs752545390 Chr.2: 11194365-11194366 on Build GRCh38
rs1001123245 Chr.2: 11194357-11194357 on Build GRCh38
rs901952608 Chr.2: 11194555-11194555 on Build GRCh38
rs755796496 Chr.2: 11194393-11194395 on Build GRCh38
rs1004136751 Chr.2: 11194378-11194378 on Build GRCh38
rs1032546061 Chr.2: 11194391-11194391 on Build GRCh38
rs918254100 Chr.2: 11194469-11194469 on Build GRCh38
rs755998768 Chr.2: 11194363-11194363 on Build GRCh38
rs995103997 Chr.2: 11194399-11194399 on Build GRCh38
rs762981787 Chr.2: 11194343-11194343 on Build GRCh38
rs1048305102 Chr.2: 11194488-11194488 on Build GRCh38
rs766535916 Chr.2: 11194358-11194359 on Build GRCh38
rs1051085519 Chr.2: 11194280-11194280 on Build GRCh38
rs753606831 Chr.2: 11194388-11194388 on Build GRCh38
rs946887000 Chr.2: 11194133-11194133 on Build GRCh38
rs972831547 Chr.2: 11194455-11194455 on Build GRCh38
rs566450656 Chr.2: 11194485-11194485 on Build GRCh38
rs946467547 Chr.2: 11194548-11194548 on Build GRCh38
rs889606976 Chr.2: 11194189-11194189 on Build GRCh38
rs917693135 Chr.2: 11194403-11194403 on Build GRCh38
rs776098469 Chr.2: 11194249-11194249 on Build GRCh38
rs921430781 Chr.2: 11194562-11194562 on Build GRCh38
rs772726360 Chr.2: 11194243-11194243 on Build GRCh38
rs186002617 Chr.2: 11194521-11194521 on Build GRCh38
rs199722144 Chr.2: 11194392-11194392 on Build GRCh38
rs562300215 Chr.2: 11194475-11194475 on Build GRCh38
rs934121877 Chr.2: 11194571-11194571 on Build GRCh38
rs765442638 Chr.2: 11194315-11194315 on Build GRCh38
rs372690238 Chr.2: 11194196-11194196 on Build GRCh38
rs763796245 Chr.2: 11194381-11194381 on Build GRCh38
rs767484397 Chr.2: 11194354-11194354 on Build GRCh38
rs369007584 Chr.2: 11194266-11194266 on Build GRCh38
rs752611113 Chr.2: 11194356-11194356 on Build GRCh38
rs976015454 Chr.2: 11194557-11194557 on Build GRCh38
rs997592792 Chr.2: 11194571-11194571 on Build GRCh38
rs371363409 Chr.2: 11194516-11194516 on Build GRCh38

View Full Product Details