Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 14634546-14635044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84A
Entrez Gene ID
151354
Gene Name
family with sequence similarity 84 member A
Gene Aliases
NSE1, PP11517
UniGene
Hs.260855
Gene Chromosome Location
Chr.2: 14632686-14640046 on Build GRCh38
Transcripts
NM_145175.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660282 c.876G>A p.E292E Chr.2: 14634855-14634855 on Build GRCh38
1613459 c.628A>C p.T210P Chr.2: 14634607-14634607 on Build GRCh38
4966899 c.686G>C p.G229A Chr.2: 14634665-14634665 on Build GRCh38
7322621 c.816C>G p.I272M Chr.2: 14634795-14634795 on Build GRCh38
7316770 c.645C>T p.F215F Chr.2: 14634624-14634624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772554241 Chr.2: 14634675-14634675 on Build GRCh38
rs775440654 Chr.2: 14634698-14634698 on Build GRCh38
rs151113082 Chr.2: 14634677-14634677 on Build GRCh38
rs554290117 Chr.2: 14634660-14634660 on Build GRCh38
rs764123791 Chr.2: 14634703-14634703 on Build GRCh38
rs751639770 Chr.2: 14634790-14634790 on Build GRCh38
rs148059470 Chr.2: 14634857-14634857 on Build GRCh38
rs761739824 Chr.2: 14634860-14634860 on Build GRCh38
rs1030438993 Chr.2: 14634936-14634936 on Build GRCh38
rs558105455 Chr.2: 14634762-14634762 on Build GRCh38
rs758473044 Chr.2: 14634709-14634709 on Build GRCh38
rs752679216 Chr.2: 14634588-14634588 on Build GRCh38
rs759331922 Chr.2: 14634695-14634695 on Build GRCh38
rs779767755 Chr.2: 14634910-14634910 on Build GRCh38
rs755342021 Chr.2: 14634752-14634752 on Build GRCh38
rs752962434 Chr.2: 14634808-14634808 on Build GRCh38
rs1012848097 Chr.2: 14634994-14634994 on Build GRCh38
rs767464077 Chr.2: 14634575-14634575 on Build GRCh38
rs752964073 Chr.2: 14634879-14634879 on Build GRCh38
rs767627527 Chr.2: 14634874-14634874 on Build GRCh38
rs781465760 Chr.2: 14634725-14634725 on Build GRCh38
rs778070458 Chr.2: 14634820-14634820 on Build GRCh38
rs1016426961 Chr.2: 14634680-14634680 on Build GRCh38
rs760541856 Chr.2: 14634772-14634772 on Build GRCh38
rs947214273 Chr.2: 14634877-14634877 on Build GRCh38
rs997576494 Chr.2: 14634923-14634923 on Build GRCh38
rs760866834 Chr.2: 14634880-14634880 on Build GRCh38
rs138567856 Chr.2: 14634833-14634833 on Build GRCh38
rs757976936 Chr.2: 14634909-14634909 on Build GRCh38
rs1005127584 Chr.2: 14634926-14634926 on Build GRCh38
rs748540195 Chr.2: 14634737-14634737 on Build GRCh38
rs767744002 Chr.2: 14634805-14634805 on Build GRCh38
rs528575711 Chr.2: 14635024-14635024 on Build GRCh38
rs763892256 Chr.2: 14634884-14634884 on Build GRCh38
rs753768386 Chr.2: 14634708-14634708 on Build GRCh38
rs755330174 Chr.2: 14634666-14634666 on Build GRCh38
rs747270798 Chr.2: 14634663-14634663 on Build GRCh38
rs757721901 Chr.2: 14634835-14634835 on Build GRCh38
rs770473121 Chr.2: 14634748-14634748 on Build GRCh38
rs759711572 Chr.2: 14634801-14634801 on Build GRCh38
rs542000572 Chr.2: 14634919-14634919 on Build GRCh38
rs779300890 Chr.2: 14634592-14634592 on Build GRCh38
rs545568492 Chr.2: 14634916-14634916 on Build GRCh38
rs536808118 Chr.2: 14634651-14634651 on Build GRCh38
rs17852410 Chr.2: 14634849-14634849 on Build GRCh38
rs374702675 Chr.2: 14634674-14634674 on Build GRCh38
rs757745054 Chr.2: 14634889-14634889 on Build GRCh38
rs371981437 Chr.2: 14634907-14634907 on Build GRCh38
rs756525535 Chr.2: 14634739-14634739 on Build GRCh38
rs971248522 Chr.2: 14635007-14635007 on Build GRCh38
rs746612514 Chr.2: 14634914-14634914 on Build GRCh38
rs374854100 Chr.2: 14634869-14634869 on Build GRCh38
rs749747903 Chr.2: 14634592-14634594 on Build GRCh38
rs779585397 Chr.2: 14634892-14634892 on Build GRCh38
rs773098737 Chr.2: 14634945-14634945 on Build GRCh38
rs576744264 Chr.2: 14634778-14634778 on Build GRCh38
rs769622207 Chr.2: 14634938-14634938 on Build GRCh38
rs769752404 Chr.2: 14634864-14634864 on Build GRCh38
rs760623968 Chr.2: 14634699-14634699 on Build GRCh38
rs768483128 Chr.2: 14634853-14634853 on Build GRCh38
rs750521675 Chr.2: 14634639-14634639 on Build GRCh38
rs866628333 Chr.2: 14634669-14634669 on Build GRCh38
rs770401072 Chr.2: 14634679-14634679 on Build GRCh38
rs745403644 Chr.2: 14634841-14634841 on Build GRCh38
rs147914327 Chr.2: 14634750-14634750 on Build GRCh38
rs970813784 Chr.2: 14634771-14634771 on Build GRCh38
rs766082550 Chr.2: 14634568-14634568 on Build GRCh38
rs745460850 Chr.2: 14634760-14634760 on Build GRCh38
rs761713256 Chr.2: 14634786-14634786 on Build GRCh38
rs767409304 Chr.2: 14634696-14634696 on Build GRCh38
rs771110896 Chr.2: 14634898-14634899 on Build GRCh38
rs773972787 Chr.2: 14634570-14634570 on Build GRCh38
rs774119229 Chr.2: 14634689-14634689 on Build GRCh38
rs551391652 Chr.2: 14634779-14634779 on Build GRCh38
rs751017634 Chr.2: 14634893-14634893 on Build GRCh38
rs190859188 Chr.2: 14634747-14634747 on Build GRCh38
rs562761110 Chr.2: 14634888-14634888 on Build GRCh38
rs766433831 Chr.2: 14634722-14634722 on Build GRCh38
rs1053020358 Chr.2: 14634832-14634832 on Build GRCh38
rs1051403518 Chr.2: 14634895-14634895 on Build GRCh38
rs868759608 Chr.2: 14634913-14634913 on Build GRCh38
rs781592536 Chr.2: 14634667-14634667 on Build GRCh38
rs367691817 Chr.2: 14634723-14634723 on Build GRCh38
rs549933261 Chr.2: 14634954-14634954 on Build GRCh38
rs561402930 Chr.2: 14634995-14634995 on Build GRCh38
rs756473710 Chr.2: 14634815-14634815 on Build GRCh38
rs544194083 Chr.2: 14634847-14634847 on Build GRCh38
rs1005075292 Chr.2: 14634678-14634678 on Build GRCh38
rs934586499 Chr.2: 14634814-14634814 on Build GRCh38
rs1023775814 Chr.2: 14634996-14634996 on Build GRCh38
rs759386804 Chr.2: 14634572-14634572 on Build GRCh38
rs778500896 Chr.2: 14634837-14634837 on Build GRCh38
rs747915125 Chr.2: 14634925-14634925 on Build GRCh38
rs145966184 Chr.2: 14634788-14634788 on Build GRCh38
rs759586743 Chr.2: 14634873-14634873 on Build GRCh38
rs775448280 Chr.2: 14635005-14635005 on Build GRCh38
rs963624247 Chr.2: 14634960-14634960 on Build GRCh38

View Full Product Details