Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 20005557-20006082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATN3
Entrez Gene ID
4148
Gene Name
matrilin 3
Gene Aliases
DIPOA, EDM5, HOA, OADIP, OS2
UniGene
Hs.656199
Gene Chromosome Location
Chr.2: 19992052-20012694 on Build GRCh38
Transcripts
NM_002381.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4508654 c.782C>T p.T261I Chr.2: 20005752-20005752 on Build GRCh38
1014825 c.648G>T p.E216D Chr.2: 20005886-20005886 on Build GRCh38
1014826 c.634G>T p.A212S Chr.2: 20005900-20005900 on Build GRCh38
7376498 c.617T>G p.V206G Chr.2: 20005917-20005917 on Build GRCh38
4572138 c.657T>A p.A219A Chr.2: 20005877-20005877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774059189 Chr.2: 20005736-20005736 on Build GRCh38
rs773642745 Chr.2: 20005841-20005841 on Build GRCh38
rs182164052 Chr.2: 20005801-20005801 on Build GRCh38
rs765225021 Chr.2: 20005950-20005950 on Build GRCh38
rs141336290 Chr.2: 20005660-20005661 on Build GRCh38
rs759213910 Chr.2: 20005743-20005743 on Build GRCh38
rs5829721 Chr.2: 20005662-20005663 on Build GRCh38
rs755185596 Chr.2: 20005880-20005880 on Build GRCh38
rs571353547 Chr.2: 20006021-20006021 on Build GRCh38
rs961591361 Chr.2: 20005777-20005777 on Build GRCh38
rs370903228 Chr.2: 20005702-20005702 on Build GRCh38
rs766686702 Chr.2: 20005883-20005883 on Build GRCh38
rs900858544 Chr.2: 20005976-20005976 on Build GRCh38
rs767267280 Chr.2: 20005749-20005749 on Build GRCh38
rs570227171 Chr.2: 20005604-20005604 on Build GRCh38
rs975351292 Chr.2: 20005683-20005683 on Build GRCh38
rs751625952 Chr.2: 20005876-20005876 on Build GRCh38
rs201491595 Chr.2: 20005863-20005863 on Build GRCh38
rs752508536 Chr.2: 20005762-20005762 on Build GRCh38
rs2077676 Chr.2: 20005591-20005591 on Build GRCh38
rs761650716 Chr.2: 20006038-20006038 on Build GRCh38
rs756307320 Chr.2: 20005981-20005981 on Build GRCh38
rs200722440 Chr.2: 20005657-20005658 on Build GRCh38
rs1025828837 Chr.2: 20005605-20005605 on Build GRCh38
rs1057116161 Chr.2: 20005620-20005620 on Build GRCh38
rs938817136 Chr.2: 20005811-20005811 on Build GRCh38
rs527641507 Chr.2: 20005582-20005582 on Build GRCh38
rs1019356302 Chr.2: 20005984-20005984 on Build GRCh38
rs371740669 Chr.2: 20005802-20005802 on Build GRCh38
rs376123290 Chr.2: 20005864-20005864 on Build GRCh38
rs778098828 Chr.2: 20005907-20005907 on Build GRCh38
rs747373453 Chr.2: 20005590-20005590 on Build GRCh38
rs72538221 Chr.2: 20005659-20005660 on Build GRCh38
rs374827048 Chr.2: 20005847-20005847 on Build GRCh38
rs201329777 Chr.2: 20005993-20005993 on Build GRCh38
rs748505019 Chr.2: 20005824-20005824 on Build GRCh38
rs557780276 Chr.2: 20005829-20005829 on Build GRCh38
rs751612128 Chr.2: 20005956-20005956 on Build GRCh38
rs368478331 Chr.2: 20005782-20005782 on Build GRCh38
rs778152107 Chr.2: 20005992-20005992 on Build GRCh38
rs749791991 Chr.2: 20005842-20005842 on Build GRCh38
rs368500408 Chr.2: 20005694-20005694 on Build GRCh38
rs777038966 Chr.2: 20005708-20005708 on Build GRCh38
rs749845872 Chr.2: 20005908-20005908 on Build GRCh38
rs977737529 Chr.2: 20006020-20006020 on Build GRCh38
rs52826764 Chr.2: 20005780-20005780 on Build GRCh38
rs773830850 Chr.2: 20005848-20005848 on Build GRCh38
rs756895540 Chr.2: 20005581-20005581 on Build GRCh38
rs3731668 Chr.2: 20005594-20005594 on Build GRCh38
rs776543565 Chr.2: 20005933-20005933 on Build GRCh38
rs546499777 Chr.2: 20005766-20005766 on Build GRCh38
rs147178840 Chr.2: 20005607-20005608 on Build GRCh38
rs375487489 Chr.2: 20005909-20005909 on Build GRCh38
rs963693880 Chr.2: 20005670-20005670 on Build GRCh38
rs1008312162 Chr.2: 20005613-20005613 on Build GRCh38
rs776597111 Chr.2: 20006024-20006024 on Build GRCh38
rs191039473 Chr.2: 20005593-20005593 on Build GRCh38
rs1031771923 Chr.2: 20005675-20005676 on Build GRCh38
rs999084190 Chr.2: 20005671-20005671 on Build GRCh38
rs894678354 Chr.2: 20005633-20005633 on Build GRCh38
rs202118156 Chr.2: 20005844-20005844 on Build GRCh38
rs767490056 Chr.2: 20005980-20005980 on Build GRCh38
rs529531121 Chr.2: 20005610-20005610 on Build GRCh38
rs1013021352 Chr.2: 20005643-20005643 on Build GRCh38
rs769937419 Chr.2: 20005743-20005744 on Build GRCh38
rs145597203 Chr.2: 20006023-20006023 on Build GRCh38
rs104893645 Chr.2: 20005953-20005953 on Build GRCh38
rs3832086 Chr.2: 20005663-20005664 on Build GRCh38
rs779422089 Chr.2: 20005997-20005997 on Build GRCh38
rs370381391 Chr.2: 20006011-20006011 on Build GRCh38
rs187943382 Chr.2: 20005875-20005875 on Build GRCh38
rs769632737 Chr.2: 20006044-20006044 on Build GRCh38
rs528598406 Chr.2: 20005913-20005913 on Build GRCh38
rs73920488 Chr.2: 20005851-20005851 on Build GRCh38
rs762787929 Chr.2: 20006054-20006054 on Build GRCh38
rs773422621 Chr.2: 20005834-20005834 on Build GRCh38
rs753889985 Chr.2: 20005870-20005870 on Build GRCh38
rs1019323796 Chr.2: 20005619-20005619 on Build GRCh38
rs28939677 Chr.2: 20005878-20005878 on Build GRCh38
rs997848702 Chr.2: 20005987-20005987 on Build GRCh38
rs779413744 Chr.2: 20006016-20006016 on Build GRCh38
rs869208593 Chr.2: 20005664-20005665 on Build GRCh38
rs759410098 Chr.2: 20006032-20006032 on Build GRCh38
rs759154288 Chr.2: 20005850-20005850 on Build GRCh38
rs761879729 Chr.2: 20005940-20005940 on Build GRCh38
rs78069341 Chr.2: 20005662-20005662 on Build GRCh38
rs766025586 Chr.2: 20005724-20005724 on Build GRCh38
rs1043215096 Chr.2: 20005904-20005904 on Build GRCh38
rs369658011 Chr.2: 20005871-20005871 on Build GRCh38
rs764078454 Chr.2: 20005775-20005775 on Build GRCh38
rs550378911 Chr.2: 20005603-20005603 on Build GRCh38
rs372346575 Chr.2: 20005706-20005706 on Build GRCh38
rs748649427 Chr.2: 20005717-20005717 on Build GRCh38
rs1043330162 Chr.2: 20005669-20005669 on Build GRCh38
rs1057019976 Chr.2: 20005589-20005589 on Build GRCh38
rs768627357 Chr.2: 20005921-20005921 on Build GRCh38
rs200762092 Chr.2: 20006008-20006008 on Build GRCh38
rs373064792 Chr.2: 20005893-20005893 on Build GRCh38
rs773268514 Chr.2: 20006053-20006053 on Build GRCh38
rs756538311 Chr.2: 20005898-20005898 on Build GRCh38
rs780273268 Chr.2: 20005796-20005796 on Build GRCh38
rs775138248 Chr.2: 20005914-20005914 on Build GRCh38
rs28401180 Chr.2: 20005919-20005919 on Build GRCh38
rs770303469 Chr.2: 20005718-20005718 on Build GRCh38

View Full Product Details