Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25165544-25166040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMC
Entrez Gene ID
5443
Gene Name
proopiomelanocortin
Gene Aliases
ACTH, CLIP, LPH, MSH, NPP, POC
UniGene
Hs.1897
Gene Chromosome Location
Chr.2: 25160853-25168851 on Build GRCh38
Transcripts
NM_000939.3 NM_001035256.2 NM_001319204.1 NM_001319205.1

SNP Detail

SNP ID Chromosome Location
rs923260454 Chr.2: 25165824-25165824 on Build GRCh38
rs888160248 Chr.2: 25165968-25165968 on Build GRCh38
rs1034929966 Chr.2: 25165595-25165595 on Build GRCh38
rs1011873082 Chr.2: 25165626-25165626 on Build GRCh38
rs1006393031 Chr.2: 25165986-25165986 on Build GRCh38
rs1016534190 Chr.2: 25165775-25165775 on Build GRCh38
rs577747120 Chr.2: 25165997-25165997 on Build GRCh38
rs913537892 Chr.2: 25165660-25165660 on Build GRCh38
rs985024093 Chr.2: 25165785-25165794 on Build GRCh38
rs946399225 Chr.2: 25165673-25165673 on Build GRCh38
rs375857744 Chr.2: 25165842-25165842 on Build GRCh38
rs534227454 Chr.2: 25165848-25165848 on Build GRCh38
rs187140257 Chr.2: 25165574-25165574 on Build GRCh38
rs746089425 Chr.2: 25165789-25165789 on Build GRCh38
rs910234699 Chr.2: 25165923-25165923 on Build GRCh38
rs929522791 Chr.2: 25165964-25165964 on Build GRCh38
rs960878462 Chr.2: 25165608-25165608 on Build GRCh38
rs1035241304 Chr.2: 25165817-25165817 on Build GRCh38
rs550717647 Chr.2: 25165688-25165688 on Build GRCh38
rs1025409812 Chr.2: 25165958-25165958 on Build GRCh38
rs1047981733 Chr.2: 25165755-25165755 on Build GRCh38
rs929507011 Chr.2: 25165750-25165750 on Build GRCh38
rs988079090 Chr.2: 25165613-25165613 on Build GRCh38
rs141471328 Chr.2: 25165809-25165809 on Build GRCh38
rs770063188 Chr.2: 25165841-25165841 on Build GRCh38
rs774077650 Chr.2: 25165865-25165865 on Build GRCh38
rs372707356 Chr.2: 25165647-25165648 on Build GRCh38
rs547421846 Chr.2: 25165889-25165889 on Build GRCh38
rs985340329 Chr.2: 25165762-25165762 on Build GRCh38
rs184500955 Chr.2: 25165922-25165922 on Build GRCh38
rs950894790 Chr.2: 25165734-25165734 on Build GRCh38
rs964794789 Chr.2: 25165940-25165940 on Build GRCh38
rs936528716 Chr.2: 25165856-25165856 on Build GRCh38
rs1006215671 Chr.2: 25165763-25165763 on Build GRCh38
rs993922887 Chr.2: 25165712-25165712 on Build GRCh38
rs547116193 Chr.2: 25165825-25165825 on Build GRCh38
rs886055854 Chr.2: 25165696-25165696 on Build GRCh38
rs565459347 Chr.2: 25165952-25165952 on Build GRCh38
rs535397453 Chr.2: 25165636-25165636 on Build GRCh38
rs905985972 Chr.2: 25165788-25165788 on Build GRCh38
rs191630457 Chr.2: 25165900-25165900 on Build GRCh38
rs1024555715 Chr.2: 25165698-25165698 on Build GRCh38
rs371624218 Chr.2: 25165795-25165795 on Build GRCh38
rs776736472 Chr.2: 25165727-25165727 on Build GRCh38
rs1047961711 Chr.2: 25165985-25165985 on Build GRCh38
rs886055855 Chr.2: 25165698-25165698 on Build GRCh38
rs555984755 Chr.2: 25165857-25165857 on Build GRCh38
rs1026926947 Chr.2: 25165718-25165718 on Build GRCh38
rs906330004 Chr.2: 25165702-25165702 on Build GRCh38
rs902030542 Chr.2: 25165735-25165735 on Build GRCh38
rs573381390 Chr.2: 25165830-25165830 on Build GRCh38
rs906194287 Chr.2: 25165950-25165950 on Build GRCh38
rs974781111 Chr.2: 25165790-25165790 on Build GRCh38
rs569002054 Chr.2: 25165786-25165786 on Build GRCh38
rs975857446 Chr.2: 25165951-25165951 on Build GRCh38

View Full Product Details